Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161689558-161690065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA6
Entrez Gene ID
2559
Gene Name
gamma-aminobutyric acid type A receptor alpha6 subunit
Gene Aliases
-
UniGene
Hs.90791
Gene Chromosome Location
Chr.5: 161685652-161702592 on Build GRCh38
Transcripts
NM_000811.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139626 c.564G>A p.W188* Chr.5: 161689670-161689670 on Build GRCh38
7104204 c.530-8A>G p.? Chr.5: 161689628-161689628 on Build GRCh38
3715031 c.630G>T p.L210L Chr.5: 161689736-161689736 on Build GRCh38
5950794 c.564G>C p.W188C Chr.5: 161689670-161689670 on Build GRCh38
5023725 c.566A>T p.K189I Chr.5: 161689672-161689672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548355913 Chr.5: 161689888-161689888 on Build GRCh38
rs563480640 Chr.5: 161689914-161689914 on Build GRCh38
rs754107619 Chr.5: 161689753-161689753 on Build GRCh38
rs762734884 Chr.5: 161689610-161689610 on Build GRCh38
rs3811993 Chr.5: 161689666-161689666 on Build GRCh38
rs764380787 Chr.5: 161689639-161689639 on Build GRCh38
rs529032834 Chr.5: 161689975-161689975 on Build GRCh38
rs899567409 Chr.5: 161689950-161689950 on Build GRCh38
rs752120603 Chr.5: 161689780-161689780 on Build GRCh38
rs922360474 Chr.5: 161690008-161690008 on Build GRCh38
rs1052904661 Chr.5: 161689999-161689999 on Build GRCh38
rs1005723214 Chr.5: 161689895-161689895 on Build GRCh38
rs752671384 Chr.5: 161689621-161689621 on Build GRCh38
rs762057483 Chr.5: 161689759-161689759 on Build GRCh38
rs367733338 Chr.5: 161689756-161689756 on Build GRCh38
rs762829158 Chr.5: 161689724-161689724 on Build GRCh38
rs969972108 Chr.5: 161689965-161689965 on Build GRCh38
rs34826485 Chr.5: 161689745-161689745 on Build GRCh38
rs958805244 Chr.5: 161689856-161689856 on Build GRCh38
rs368306346 Chr.5: 161689595-161689595 on Build GRCh38
rs768261411 Chr.5: 161689588-161689588 on Build GRCh38
rs541145520 Chr.5: 161689804-161689804 on Build GRCh38
rs10043518 Chr.5: 161689940-161689940 on Build GRCh38
rs760867520 Chr.5: 161689632-161689632 on Build GRCh38
rs547453071 Chr.5: 161689594-161689594 on Build GRCh38
rs574955747 Chr.5: 161689644-161689644 on Build GRCh38
rs768208279 Chr.5: 161689677-161689677 on Build GRCh38
rs183133713 Chr.5: 161689792-161689792 on Build GRCh38
rs4921402 Chr.5: 161689897-161689897 on Build GRCh38
rs774791692 Chr.5: 161689582-161689582 on Build GRCh38
rs890630067 Chr.5: 161689974-161689975 on Build GRCh38
rs768545890 Chr.5: 161689611-161689614 on Build GRCh38
rs916545688 Chr.5: 161689956-161689956 on Build GRCh38
rs780406601 Chr.5: 161689779-161689779 on Build GRCh38
rs774140490 Chr.5: 161689821-161689821 on Build GRCh38
rs111753385 Chr.5: 161689827-161689827 on Build GRCh38
rs747671960 Chr.5: 161689794-161689794 on Build GRCh38
rs757682550 Chr.5: 161689658-161689658 on Build GRCh38
rs868382842 Chr.5: 161689669-161689669 on Build GRCh38
rs991245182 Chr.5: 161689952-161689952 on Build GRCh38
rs928024636 Chr.5: 161689627-161689627 on Build GRCh38
rs759611967 Chr.5: 161689824-161689824 on Build GRCh38
rs747570774 Chr.5: 161689727-161689727 on Build GRCh38
rs574652664 Chr.5: 161689989-161689989 on Build GRCh38
rs111768483 Chr.5: 161689992-161689992 on Build GRCh38
rs899147489 Chr.5: 161690004-161690004 on Build GRCh38
rs925816395 Chr.5: 161689975-161689975 on Build GRCh38
rs60087947 Chr.5: 161689862-161689862 on Build GRCh38
rs769441572 Chr.5: 161689704-161689704 on Build GRCh38
rs754160838 Chr.5: 161689656-161689656 on Build GRCh38
rs41311631 Chr.5: 161690035-161690035 on Build GRCh38
rs113538260 Chr.5: 161689972-161689972 on Build GRCh38
rs933759358 Chr.5: 161689951-161689951 on Build GRCh38
rs900861590 Chr.5: 161689806-161689806 on Build GRCh38
rs746474290 Chr.5: 161689672-161689672 on Build GRCh38
rs772826373 Chr.5: 161689714-161689714 on Build GRCh38
rs925576667 Chr.5: 161689906-161689906 on Build GRCh38
rs973429856 Chr.5: 161689833-161689833 on Build GRCh38
rs749266935 Chr.5: 161689590-161689598 on Build GRCh38
rs746538862 Chr.5: 161689586-161689586 on Build GRCh38
rs1042913786 Chr.5: 161689719-161689719 on Build GRCh38
rs200968260 Chr.5: 161689769-161689769 on Build GRCh38
rs534671781 Chr.5: 161690016-161690016 on Build GRCh38
rs376905968 Chr.5: 161689863-161689863 on Build GRCh38
rs765078307 Chr.5: 161689600-161689600 on Build GRCh38
rs147064708 Chr.5: 161689754-161689754 on Build GRCh38
rs368251992 Chr.5: 161689798-161689798 on Build GRCh38
rs371369081 Chr.5: 161689817-161689817 on Build GRCh38
rs200341002 Chr.5: 161689782-161689782 on Build GRCh38
rs760815978 Chr.5: 161689740-161689740 on Build GRCh38
rs765506471 Chr.5: 161689768-161689768 on Build GRCh38
rs774386607 Chr.5: 161689737-161689737 on Build GRCh38
rs748105776 Chr.5: 161689691-161689696 on Build GRCh38
rs900645002 Chr.5: 161689749-161689749 on Build GRCh38
rs927399334 Chr.5: 161689842-161689842 on Build GRCh38
rs770808265 Chr.5: 161689818-161689818 on Build GRCh38
rs933734174 Chr.5: 161690015-161690015 on Build GRCh38
rs960793811 Chr.5: 161689949-161689949 on Build GRCh38
rs3841130 Chr.5: 161690042-161690043 on Build GRCh38
rs945951590 Chr.5: 161689878-161689878 on Build GRCh38
rs3811992 Chr.5: 161689936-161689936 on Build GRCh38
rs889448790 Chr.5: 161690020-161690020 on Build GRCh38
rs138428207 Chr.5: 161689771-161689771 on Build GRCh38
rs145469537 Chr.5: 161689731-161689731 on Build GRCh38
rs934464003 Chr.5: 161689982-161689982 on Build GRCh38
rs200710979 Chr.5: 161689667-161689667 on Build GRCh38
rs1038568153 Chr.5: 161690036-161690036 on Build GRCh38
rs780812038 Chr.5: 161689691-161689691 on Build GRCh38
rs143830239 Chr.5: 161689700-161689700 on Build GRCh38
rs779209499 Chr.5: 161689662-161689662 on Build GRCh38
rs766354679 Chr.5: 161689619-161689619 on Build GRCh38
rs925743412 Chr.5: 161689728-161689728 on Build GRCh38
rs961520539 Chr.5: 161689903-161689903 on Build GRCh38
rs774441339 Chr.5: 161689652-161689657 on Build GRCh38
rs770734349 Chr.5: 161689729-161689729 on Build GRCh38

View Full Product Details