Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177459015-177459522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBN1
Entrez Gene ID
1627
Gene Name
drebrin 1
Gene Aliases
D0S117E
UniGene
Hs.130316
Gene Chromosome Location
Chr.5: 177456612-177480368 on Build GRCh38
Transcripts
NM_004395.3 NM_080881.2 XM_005265827.3 XM_006714826.2 XM_011534446.2 XM_011534447.2 XM_017009139.1 XM_017009140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640243 c.1006C>T p.P336S Chr.5: 177459224-177459224 on Build GRCh38
6496496 c.1086G>T p.S362S Chr.5: 177459138-177459138 on Build GRCh38
4905053 c.1099C>T p.P367S Chr.5: 177459131-177459131 on Build GRCh38
6640247 c.1056G>A p.E352E Chr.5: 177459174-177459174 on Build GRCh38
3373817 c.1011C>T p.S337S Chr.5: 177459213-177459213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759829739 Chr.5: 177459237-177459237 on Build GRCh38
rs748335051 Chr.5: 177459220-177459220 on Build GRCh38
rs778827333 Chr.5: 177459216-177459216 on Build GRCh38
rs767872072 Chr.5: 177459238-177459238 on Build GRCh38
rs906538142 Chr.5: 177459314-177459314 on Build GRCh38
rs1005732278 Chr.5: 177459480-177459480 on Build GRCh38
rs760768420 Chr.5: 177459086-177459086 on Build GRCh38
rs781411729 Chr.5: 177459383-177459383 on Build GRCh38
rs769703884 Chr.5: 177459065-177459065 on Build GRCh38
rs749173170 Chr.5: 177459150-177459150 on Build GRCh38
rs113667169 Chr.5: 177459211-177459211 on Build GRCh38
rs754541844 Chr.5: 177459050-177459050 on Build GRCh38
rs896066143 Chr.5: 177459436-177459436 on Build GRCh38
rs777223635 Chr.5: 177459204-177459204 on Build GRCh38
rs1043385874 Chr.5: 177459146-177459146 on Build GRCh38
rs762442289 Chr.5: 177459115-177459115 on Build GRCh38
rs753333141 Chr.5: 177459281-177459281 on Build GRCh38
rs374420181 Chr.5: 177459191-177459191 on Build GRCh38
rs747305657 Chr.5: 177459156-177459156 on Build GRCh38
rs374027433 Chr.5: 177459168-177459168 on Build GRCh38
rs569806768 Chr.5: 177459155-177459155 on Build GRCh38
rs774852138 Chr.5: 177459235-177459235 on Build GRCh38
rs770029000 Chr.5: 177459225-177459225 on Build GRCh38
rs887253457 Chr.5: 177459477-177459477 on Build GRCh38
rs904881260 Chr.5: 177459159-177459159 on Build GRCh38
rs534580030 Chr.5: 177459195-177459195 on Build GRCh38
rs751259597 Chr.5: 177459128-177459128 on Build GRCh38
rs34692220 Chr.5: 177459118-177459118 on Build GRCh38
rs201656197 Chr.5: 177459079-177459079 on Build GRCh38
rs758849423 Chr.5: 177459127-177459127 on Build GRCh38
rs913765291 Chr.5: 177459274-177459274 on Build GRCh38
rs574208563 Chr.5: 177459456-177459456 on Build GRCh38
rs750318364 Chr.5: 177459297-177459297 on Build GRCh38
rs770982243 Chr.5: 177459320-177459320 on Build GRCh38
rs951485393 Chr.5: 177459184-177459184 on Build GRCh38
rs537160653 Chr.5: 177459166-177459166 on Build GRCh38
rs145842585 Chr.5: 177459138-177459138 on Build GRCh38
rs763518397 Chr.5: 177459179-177459179 on Build GRCh38
rs773055413 Chr.5: 177459068-177459068 on Build GRCh38
rs199742703 Chr.5: 177459063-177459063 on Build GRCh38
rs757449081 Chr.5: 177459362-177459362 on Build GRCh38
rs112618456 Chr.5: 177459234-177459234 on Build GRCh38
rs531788215 Chr.5: 177459058-177459058 on Build GRCh38
rs763642188 Chr.5: 177459264-177459264 on Build GRCh38
rs760288037 Chr.5: 177459251-177459251 on Build GRCh38
rs953696331 Chr.5: 177459468-177459468 on Build GRCh38
rs770123909 Chr.5: 177459311-177459311 on Build GRCh38
rs987708993 Chr.5: 177459489-177459489 on Build GRCh38
rs781287215 Chr.5: 177459219-177459219 on Build GRCh38
rs780533135 Chr.5: 177459053-177459053 on Build GRCh38
rs370415928 Chr.5: 177459303-177459303 on Build GRCh38
rs201033708 Chr.5: 177459194-177459194 on Build GRCh38
rs775803342 Chr.5: 177459085-177459085 on Build GRCh38
rs755552376 Chr.5: 177459144-177459144 on Build GRCh38
rs149274225 Chr.5: 177459210-177459210 on Build GRCh38
rs753006714 Chr.5: 177459164-177459164 on Build GRCh38
rs763607968 Chr.5: 177459229-177459229 on Build GRCh38
rs750463326 Chr.5: 177459412-177459412 on Build GRCh38
rs922526617 Chr.5: 177459089-177459089 on Build GRCh38
rs374073663 Chr.5: 177459309-177459309 on Build GRCh38
rs755736671 Chr.5: 177459183-177459184 on Build GRCh38
rs562104269 Chr.5: 177459346-177459346 on Build GRCh38
rs144418852 Chr.5: 177459217-177459217 on Build GRCh38
rs963459667 Chr.5: 177459418-177459418 on Build GRCh38
rs200475259 Chr.5: 177459218-177459218 on Build GRCh38
rs375373491 Chr.5: 177459139-177459139 on Build GRCh38
rs567476563 Chr.5: 177459180-177459180 on Build GRCh38
rs377045843 Chr.5: 177459248-177459248 on Build GRCh38
rs116548591 Chr.5: 177459228-177459228 on Build GRCh38
rs1004880457 Chr.5: 177459345-177459345 on Build GRCh38
rs776551029 Chr.5: 177459101-177459101 on Build GRCh38
rs755642391 Chr.5: 177459197-177459197 on Build GRCh38
rs35186664 Chr.5: 177459491-177459491 on Build GRCh38
rs757252967 Chr.5: 177459290-177459290 on Build GRCh38
rs199808119 Chr.5: 177459243-177459243 on Build GRCh38
rs778722477 Chr.5: 177459295-177459295 on Build GRCh38
rs879246127 Chr.5: 177459087-177459087 on Build GRCh38
rs139033012 Chr.5: 177459117-177459117 on Build GRCh38
rs995340951 Chr.5: 177459451-177459451 on Build GRCh38
rs749085048 Chr.5: 177459072-177459072 on Build GRCh38
rs199917938 Chr.5: 177459147-177459147 on Build GRCh38
rs937637580 Chr.5: 177459401-177459401 on Build GRCh38
rs139683766 Chr.5: 177459178-177459178 on Build GRCh38
rs1014879790 Chr.5: 177459349-177459349 on Build GRCh38
rs1014490582 Chr.5: 177459444-177459444 on Build GRCh38
rs764275212 Chr.5: 177459088-177459088 on Build GRCh38
rs201408921 Chr.5: 177459244-177459244 on Build GRCh38
rs549405976 Chr.5: 177459325-177459325 on Build GRCh38
rs541340235 Chr.5: 177459472-177459472 on Build GRCh38
rs750160145 Chr.5: 177459139-177459141 on Build GRCh38
rs754561090 Chr.5: 177459129-177459129 on Build GRCh38
rs183235683 Chr.5: 177459312-177459312 on Build GRCh38
rs749380469 Chr.5: 177459313-177459313 on Build GRCh38

View Full Product Details