Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179032880-179033381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF879
Entrez Gene ID
345462
Gene Name
zinc finger protein 879
Gene Aliases
DKFZp686E2433
UniGene
Hs.445740
Gene Chromosome Location
Chr.5: 179023759-179034387 on Build GRCh38
Transcripts
NM_001136116.1 XM_005265908.3 XM_011534550.2 XM_011534551.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7399790 c.968G>A p.R323K Chr.5: 179032916-179032916 on Build GRCh38
1066603 c.1093A>C p.I365L Chr.5: 179033041-179033041 on Build GRCh38
7327995 c.1028C>T p.P343L Chr.5: 179032976-179032976 on Build GRCh38
1066602 c.1091G>A p.R364Q Chr.5: 179033039-179033039 on Build GRCh38
5466402 c.1424G>A p.R475Q Chr.5: 179033372-179033372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973408174 Chr.5: 179033302-179033302 on Build GRCh38
rs760674313 Chr.5: 179033272-179033272 on Build GRCh38
rs201080388 Chr.5: 179032983-179032983 on Build GRCh38
rs770031204 Chr.5: 179033110-179033110 on Build GRCh38
rs750137753 Chr.5: 179033330-179033330 on Build GRCh38
rs116786294 Chr.5: 179033040-179033040 on Build GRCh38
rs761994175 Chr.5: 179032976-179032976 on Build GRCh38
rs771342608 Chr.5: 179033345-179033345 on Build GRCh38
rs748711219 Chr.5: 179033197-179033197 on Build GRCh38
rs759349535 Chr.5: 179033145-179033145 on Build GRCh38
rs780406331 Chr.5: 179032916-179032916 on Build GRCh38
rs774129557 Chr.5: 179033124-179033124 on Build GRCh38
rs753431703 Chr.5: 179033159-179033159 on Build GRCh38
rs962903402 Chr.5: 179032925-179032925 on Build GRCh38
rs1047452834 Chr.5: 179033300-179033300 on Build GRCh38
rs780539539 Chr.5: 179033058-179033058 on Build GRCh38
rs756132792 Chr.5: 179033201-179033201 on Build GRCh38
rs751437598 Chr.5: 179033008-179033008 on Build GRCh38
rs865979653 Chr.5: 179033288-179033288 on Build GRCh38
rs747612026 Chr.5: 179032924-179032924 on Build GRCh38
rs543177356 Chr.5: 179033310-179033310 on Build GRCh38
rs766647489 Chr.5: 179033161-179033162 on Build GRCh38
rs773282146 Chr.5: 179033111-179033111 on Build GRCh38
rs1011214040 Chr.5: 179033128-179033128 on Build GRCh38
rs375911965 Chr.5: 179033273-179033273 on Build GRCh38
rs566103156 Chr.5: 179033096-179033096 on Build GRCh38
rs767445646 Chr.5: 179033019-179033019 on Build GRCh38
rs759535880 Chr.5: 179033018-179033018 on Build GRCh38
rs775395836 Chr.5: 179033271-179033271 on Build GRCh38
rs771126982 Chr.5: 179033229-179033229 on Build GRCh38
rs373329459 Chr.5: 179033151-179033151 on Build GRCh38
rs769332422 Chr.5: 179033110-179033112 on Build GRCh38
rs575817184 Chr.5: 179033284-179033284 on Build GRCh38
rs749639156 Chr.5: 179032994-179033001 on Build GRCh38
rs986737354 Chr.5: 179032989-179032989 on Build GRCh38
rs749331918 Chr.5: 179033228-179033228 on Build GRCh38
rs748206740 Chr.5: 179033023-179033023 on Build GRCh38
rs766244526 Chr.5: 179032992-179032992 on Build GRCh38
rs919291218 Chr.5: 179033319-179033319 on Build GRCh38
rs1030480915 Chr.5: 179033188-179033188 on Build GRCh38
rs951272907 Chr.5: 179033321-179033321 on Build GRCh38
rs568627602 Chr.5: 179033038-179033038 on Build GRCh38
rs771904927 Chr.5: 179033243-179033243 on Build GRCh38
rs752155214 Chr.5: 179033021-179033021 on Build GRCh38
rs910541761 Chr.5: 179032984-179032984 on Build GRCh38
rs773460949 Chr.5: 179033115-179033115 on Build GRCh38
rs758680623 Chr.5: 179033044-179033044 on Build GRCh38
rs954815342 Chr.5: 179032969-179032969 on Build GRCh38
rs553838683 Chr.5: 179033065-179033065 on Build GRCh38
rs767404275 Chr.5: 179033148-179033148 on Build GRCh38
rs111486809 Chr.5: 179032977-179032977 on Build GRCh38
rs776788653 Chr.5: 179032955-179032955 on Build GRCh38
rs751918879 Chr.5: 179033181-179033181 on Build GRCh38
rs963335810 Chr.5: 179033280-179033280 on Build GRCh38
rs764814824 Chr.5: 179033176-179033176 on Build GRCh38
rs371754812 Chr.5: 179032965-179032965 on Build GRCh38
rs903439539 Chr.5: 179033155-179033155 on Build GRCh38
rs759955520 Chr.5: 179033344-179033344 on Build GRCh38
rs973964041 Chr.5: 179033014-179033014 on Build GRCh38
rs778869843 Chr.5: 179033020-179033020 on Build GRCh38
rs781769103 Chr.5: 179033088-179033088 on Build GRCh38
rs773511634 Chr.5: 179032979-179032979 on Build GRCh38
rs1030491207 Chr.5: 179032940-179032943 on Build GRCh38
rs879817972 Chr.5: 179032974-179032974 on Build GRCh38
rs763721076 Chr.5: 179033154-179033154 on Build GRCh38
rs746581394 Chr.5: 179033256-179033258 on Build GRCh38
rs764900886 Chr.5: 179033311-179033311 on Build GRCh38
rs772213908 Chr.5: 179033073-179033073 on Build GRCh38
rs755652706 Chr.5: 179033028-179033028 on Build GRCh38
rs770496225 Chr.5: 179033286-179033286 on Build GRCh38
rs749581718 Chr.5: 179033112-179033112 on Build GRCh38
rs777974953 Chr.5: 179033215-179033215 on Build GRCh38
rs376464480 Chr.5: 179033239-179033239 on Build GRCh38
rs975478173 Chr.5: 179032962-179032962 on Build GRCh38
rs772528683 Chr.5: 179032912-179032912 on Build GRCh38
rs527743804 Chr.5: 179033240-179033240 on Build GRCh38
rs372355840 Chr.5: 179033144-179033144 on Build GRCh38
rs896208852 Chr.5: 179033126-179033126 on Build GRCh38
rs557857712 Chr.5: 179033195-179033195 on Build GRCh38
rs756871165 Chr.5: 179033175-179033175 on Build GRCh38

View Full Product Details