Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 149931168-149931663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
UniGene
Hs.425144
Gene Chromosome Location
Chr.1: 149928651-149936898 on Build GRCh38
Transcripts
NM_001145862.1 NM_181873.3 XM_006711137.1 XM_011509099.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3862622 c.1191C>T p.A397A Chr.1: 149931359-149931359 on Build GRCh38
4021903 c.1073A>T p.E358V Chr.1: 149931261-149931261 on Build GRCh38
462892 c.1249C>G p.P417A Chr.1: 149931301-149931301 on Build GRCh38
5688638 c.1167C>A p.V389V Chr.1: 149931383-149931383 on Build GRCh38
1472564 c.1251C>A p.P417P Chr.1: 149931299-149931299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782160730 Chr.1: 149931288-149931288 on Build GRCh38
rs782501862 Chr.1: 149931232-149931232 on Build GRCh38
rs782790149 Chr.1: 149931423-149931423 on Build GRCh38
rs782655361 Chr.1: 149931372-149931372 on Build GRCh38
rs72692819 Chr.1: 149931230-149931230 on Build GRCh38
rs375222367 Chr.1: 149931236-149931236 on Build GRCh38
rs782539192 Chr.1: 149931460-149931460 on Build GRCh38
rs781810386 Chr.1: 149931308-149931308 on Build GRCh38
rs782446121 Chr.1: 149931365-149931365 on Build GRCh38
rs587614222 Chr.1: 149931461-149931461 on Build GRCh38
rs587664270 Chr.1: 149931550-149931550 on Build GRCh38
rs782088294 Chr.1: 149931320-149931320 on Build GRCh38
rs369746257 Chr.1: 149931246-149931246 on Build GRCh38
rs587759324 Chr.1: 149931464-149931464 on Build GRCh38
rs587724284 Chr.1: 149931333-149931333 on Build GRCh38
rs782534836 Chr.1: 149931351-149931351 on Build GRCh38
rs1012662641 Chr.1: 149931572-149931572 on Build GRCh38
rs782492859 Chr.1: 149931265-149931265 on Build GRCh38
rs373958464 Chr.1: 149931271-149931271 on Build GRCh38
rs781873910 Chr.1: 149931450-149931450 on Build GRCh38
rs989517022 Chr.1: 149931414-149931414 on Build GRCh38
rs587764831 Chr.1: 149931529-149931529 on Build GRCh38
rs890338851 Chr.1: 149931192-149931192 on Build GRCh38
rs916756655 Chr.1: 149931267-149931267 on Build GRCh38
rs183984411 Chr.1: 149931462-149931462 on Build GRCh38
rs377265000 Chr.1: 149931235-149931235 on Build GRCh38
rs75910903 Chr.1: 149931391-149931391 on Build GRCh38
rs781789146 Chr.1: 149931364-149931364 on Build GRCh38
rs782430686 Chr.1: 149931415-149931415 on Build GRCh38
rs200897487 Chr.1: 149931475-149931475 on Build GRCh38
rs141696447 Chr.1: 149931504-149931504 on Build GRCh38
rs890285805 Chr.1: 149931457-149931457 on Build GRCh38
rs782708821 Chr.1: 149931289-149931289 on Build GRCh38
rs782025362 Chr.1: 149931287-149931287 on Build GRCh38
rs915273926 Chr.1: 149931419-149931419 on Build GRCh38
rs961996358 Chr.1: 149931231-149931231 on Build GRCh38
rs782400204 Chr.1: 149931285-149931285 on Build GRCh38
rs782128829 Chr.1: 149931422-149931422 on Build GRCh38
rs782434910 Chr.1: 149931220-149931220 on Build GRCh38
rs587740865 Chr.1: 149931445-149931445 on Build GRCh38
rs782673843 Chr.1: 149931358-149931358 on Build GRCh38
rs781907037 Chr.1: 149931555-149931555 on Build GRCh38
rs782228227 Chr.1: 149931472-149931472 on Build GRCh38
rs782554129 Chr.1: 149931247-149931247 on Build GRCh38
rs1024098181 Chr.1: 149931617-149931617 on Build GRCh38
rs970865356 Chr.1: 149931621-149931621 on Build GRCh38
rs782759787 Chr.1: 149931325-149931325 on Build GRCh38
rs376034416 Chr.1: 149931405-149931405 on Build GRCh38
rs782757993 Chr.1: 149931449-149931449 on Build GRCh38
rs782726457 Chr.1: 149931429-149931429 on Build GRCh38
rs377020825 Chr.1: 149931503-149931503 on Build GRCh38
rs587673875 Chr.1: 149931261-149931261 on Build GRCh38
rs587743913 Chr.1: 149931608-149931608 on Build GRCh38
rs1027324050 Chr.1: 149931505-149931505 on Build GRCh38
rs149091972 Chr.1: 149931404-149931404 on Build GRCh38
rs781982143 Chr.1: 149931421-149931421 on Build GRCh38
rs781927053 Chr.1: 149931416-149931416 on Build GRCh38
rs374325191 Chr.1: 149931620-149931620 on Build GRCh38
rs924210809 Chr.1: 149931299-149931299 on Build GRCh38
rs781977650 Chr.1: 149931277-149931277 on Build GRCh38
rs782070877 Chr.1: 149931420-149931420 on Build GRCh38
rs782283078 Chr.1: 149931411-149931411 on Build GRCh38
rs894180484 Chr.1: 149931506-149931506 on Build GRCh38

View Full Product Details