Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150262964-150263459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA14
Entrez Gene ID
23632
Gene Name
carbonic anhydrase 14
Gene Aliases
CAXiV
UniGene
Hs.528988
Gene Chromosome Location
Chr.1: 150257255-150265078 on Build GRCh38
Transcripts
NM_012113.2 XM_005245059.2 XM_005245060.2 XM_006711259.2 XM_006711261.2 XM_011509379.2 XM_017000893.1 XM_017000894.1 XM_017000895.1 XM_017000896.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6343169 c.720+7G>T p.? Chr.1: 150263206-150263206 on Build GRCh38
1199138 c.710C>A p.S237* Chr.1: 150263189-150263189 on Build GRCh38
895476 c.830C>A p.S277Y Chr.1: 150263408-150263408 on Build GRCh38
6024793 c.627C>A p.Y209* Chr.1: 150263106-150263106 on Build GRCh38
895475 c.822C>A p.V274V Chr.1: 150263400-150263400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781854917 Chr.1: 150263171-150263171 on Build GRCh38
rs371804274 Chr.1: 150263212-150263212 on Build GRCh38
rs781851841 Chr.1: 150263320-150263320 on Build GRCh38
rs199642489 Chr.1: 150263123-150263123 on Build GRCh38
rs782289969 Chr.1: 150263004-150263004 on Build GRCh38
rs782007977 Chr.1: 150263319-150263319 on Build GRCh38
rs369770952 Chr.1: 150263393-150263393 on Build GRCh38
rs782118148 Chr.1: 150263236-150263236 on Build GRCh38
rs782430716 Chr.1: 150263350-150263350 on Build GRCh38
rs993679392 Chr.1: 150263426-150263426 on Build GRCh38
rs782090209 Chr.1: 150263026-150263026 on Build GRCh38
rs147099110 Chr.1: 150263392-150263392 on Build GRCh38
rs782376644 Chr.1: 150263360-150263360 on Build GRCh38
rs782635141 Chr.1: 150263207-150263207 on Build GRCh38
rs147108777 Chr.1: 150263058-150263058 on Build GRCh38
rs587720730 Chr.1: 150263209-150263209 on Build GRCh38
rs370307632 Chr.1: 150262994-150262994 on Build GRCh38
rs782468374 Chr.1: 150263272-150263272 on Build GRCh38
rs1052955326 Chr.1: 150262987-150262987 on Build GRCh38
rs782370173 Chr.1: 150263369-150263369 on Build GRCh38
rs782579828 Chr.1: 150263285-150263285 on Build GRCh38
rs781844460 Chr.1: 150263185-150263185 on Build GRCh38
rs200952346 Chr.1: 150263163-150263163 on Build GRCh38
rs368797497 Chr.1: 150263121-150263121 on Build GRCh38
rs373055861 Chr.1: 150263293-150263293 on Build GRCh38
rs782711012 Chr.1: 150263284-150263284 on Build GRCh38
rs781917865 Chr.1: 150263039-150263039 on Build GRCh38
rs889456995 Chr.1: 150263370-150263370 on Build GRCh38
rs149155658 Chr.1: 150263337-150263337 on Build GRCh38
rs374781042 Chr.1: 150263147-150263147 on Build GRCh38
rs372825966 Chr.1: 150263140-150263140 on Build GRCh38
rs782152998 Chr.1: 150263164-150263164 on Build GRCh38
rs782171606 Chr.1: 150263107-150263107 on Build GRCh38
rs782364436 Chr.1: 150263030-150263030 on Build GRCh38
rs782077787 Chr.1: 150263159-150263159 on Build GRCh38
rs3738323 Chr.1: 150263124-150263124 on Build GRCh38
rs782433764 Chr.1: 150263164-150263165 on Build GRCh38
rs970584562 Chr.1: 150263247-150263247 on Build GRCh38
rs782546928 Chr.1: 150263204-150263204 on Build GRCh38
rs369293943 Chr.1: 150263233-150263233 on Build GRCh38
rs782320990 Chr.1: 150263125-150263125 on Build GRCh38
rs782734595 Chr.1: 150263193-150263193 on Build GRCh38
rs587755689 Chr.1: 150263327-150263327 on Build GRCh38
rs781960899 Chr.1: 150263267-150263267 on Build GRCh38
rs781964610 Chr.1: 150263221-150263221 on Build GRCh38
rs182810500 Chr.1: 150263052-150263052 on Build GRCh38
rs782216582 Chr.1: 150263240-150263240 on Build GRCh38
rs782094375 Chr.1: 150263409-150263409 on Build GRCh38
rs139457338 Chr.1: 150263313-150263313 on Build GRCh38
rs183068071 Chr.1: 150263388-150263388 on Build GRCh38
rs537082825 Chr.1: 150263098-150263098 on Build GRCh38
rs928501994 Chr.1: 150263261-150263261 on Build GRCh38
rs782635287 Chr.1: 150263238-150263238 on Build GRCh38
rs782581132 Chr.1: 150263194-150263194 on Build GRCh38
rs142693414 Chr.1: 150263111-150263111 on Build GRCh38
rs914503836 Chr.1: 150263341-150263341 on Build GRCh38
rs782203061 Chr.1: 150263223-150263223 on Build GRCh38
rs981540344 Chr.1: 150263259-150263259 on Build GRCh38
rs782341031 Chr.1: 150263217-150263217 on Build GRCh38
rs185727898 Chr.1: 150263330-150263330 on Build GRCh38
rs782113059 Chr.1: 150263063-150263063 on Build GRCh38
rs782651671 Chr.1: 150263361-150263361 on Build GRCh38
rs782544136 Chr.1: 150263117-150263117 on Build GRCh38
rs1007939345 Chr.1: 150263081-150263081 on Build GRCh38
rs368149759 Chr.1: 150263006-150263006 on Build GRCh38
rs144552698 Chr.1: 150263431-150263431 on Build GRCh38
rs200048032 Chr.1: 150263012-150263012 on Build GRCh38
rs150554080 Chr.1: 150263133-150263133 on Build GRCh38
rs782442582 Chr.1: 150263192-150263192 on Build GRCh38
rs782742200 Chr.1: 150263162-150263162 on Build GRCh38
rs782776922 Chr.1: 150263075-150263075 on Build GRCh38
rs782010133 Chr.1: 150263126-150263126 on Build GRCh38
rs782658910 Chr.1: 150263001-150263001 on Build GRCh38
rs375722927 Chr.1: 150263076-150263076 on Build GRCh38
rs782187217 Chr.1: 150262991-150262991 on Build GRCh38
rs373907752 Chr.1: 150263394-150263394 on Build GRCh38
rs782282293 Chr.1: 150263128-150263128 on Build GRCh38
rs781893937 Chr.1: 150263059-150263059 on Build GRCh38
rs376799889 Chr.1: 150263229-150263229 on Build GRCh38
rs782785318 Chr.1: 150263266-150263266 on Build GRCh38
rs782274683 Chr.1: 150263201-150263201 on Build GRCh38
rs587643737 Chr.1: 150263110-150263110 on Build GRCh38
rs190973369 Chr.1: 150263355-150263355 on Build GRCh38
rs587769091 Chr.1: 150263173-150263173 on Build GRCh38
rs782676908 Chr.1: 150263352-150263352 on Build GRCh38
rs782815220 Chr.1: 150263165-150263165 on Build GRCh38
rs782303556 Chr.1: 150263351-150263351 on Build GRCh38
rs782773744 Chr.1: 150263429-150263429 on Build GRCh38
rs377358154 Chr.1: 150263316-150263316 on Build GRCh38
rs199956011 Chr.1: 150263179-150263179 on Build GRCh38
rs782101140 Chr.1: 150263283-150263283 on Build GRCh38
rs587602115 Chr.1: 150263368-150263368 on Build GRCh38
rs148420100 Chr.1: 150263432-150263432 on Build GRCh38
rs782443282 Chr.1: 150263077-150263077 on Build GRCh38

View Full Product Details