Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181155245-181155741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2V2
Entrez Gene ID
285659
Gene Name
olfactory receptor family 2 subfamily V member 2
Gene Aliases
OR2V3, OST713
UniGene
Hs.553681
Gene Chromosome Location
Chr.5: 181154943-181155890 on Build GRCh38
Transcripts
NM_206880.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5320409 c.466G>A p.D156N Chr.5: 181155408-181155408 on Build GRCh38
364025 c.736C>A p.L246M Chr.5: 181155678-181155678 on Build GRCh38
1311065 c.685A>G p.M229V Chr.5: 181155627-181155627 on Build GRCh38
3744819 c.446C>T p.S149F Chr.5: 181155388-181155388 on Build GRCh38
163128 c.408C>T p.L136L Chr.5: 181155350-181155350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544530511 Chr.5: 181155514-181155514 on Build GRCh38
rs369495119 Chr.5: 181155705-181155705 on Build GRCh38
rs765758694 Chr.5: 181155369-181155369 on Build GRCh38
rs751302973 Chr.5: 181155709-181155709 on Build GRCh38
rs201817098 Chr.5: 181155412-181155412 on Build GRCh38
rs746254883 Chr.5: 181155289-181155289 on Build GRCh38
rs528232543 Chr.5: 181155588-181155588 on Build GRCh38
rs868856438 Chr.5: 181155674-181155674 on Build GRCh38
rs745838099 Chr.5: 181155603-181155603 on Build GRCh38
rs146384535 Chr.5: 181155587-181155587 on Build GRCh38
rs753980331 Chr.5: 181155598-181155598 on Build GRCh38
rs865981525 Chr.5: 181155342-181155342 on Build GRCh38
rs770800753 Chr.5: 181155404-181155404 on Build GRCh38
rs190605759 Chr.5: 181155629-181155629 on Build GRCh38
rs537379075 Chr.5: 181155330-181155330 on Build GRCh38
rs757812634 Chr.5: 181155269-181155269 on Build GRCh38
rs866667348 Chr.5: 181155437-181155437 on Build GRCh38
rs749288116 Chr.5: 181155401-181155401 on Build GRCh38
rs968497778 Chr.5: 181155475-181155475 on Build GRCh38
rs749197962 Chr.5: 181155480-181155480 on Build GRCh38
rs754870897 Chr.5: 181155718-181155718 on Build GRCh38
rs201187190 Chr.5: 181155317-181155317 on Build GRCh38
rs747655556 Chr.5: 181155626-181155626 on Build GRCh38
rs760468337 Chr.5: 181155361-181155361 on Build GRCh38
rs147476494 Chr.5: 181155465-181155465 on Build GRCh38
rs769833872 Chr.5: 181155395-181155395 on Build GRCh38
rs757469535 Chr.5: 181155601-181155601 on Build GRCh38
rs750955578 Chr.5: 181155375-181155375 on Build GRCh38
rs200123627 Chr.5: 181155408-181155408 on Build GRCh38
rs942179929 Chr.5: 181155621-181155621 on Build GRCh38
rs201302859 Chr.5: 181155688-181155688 on Build GRCh38
rs756322798 Chr.5: 181155591-181155591 on Build GRCh38
rs12108779 Chr.5: 181155314-181155314 on Build GRCh38
rs762975612 Chr.5: 181155482-181155494 on Build GRCh38
rs57951228 Chr.5: 181155321-181155321 on Build GRCh38
rs371453705 Chr.5: 181155537-181155537 on Build GRCh38
rs746610207 Chr.5: 181155610-181155610 on Build GRCh38
rs774125205 Chr.5: 181155334-181155334 on Build GRCh38
rs561264667 Chr.5: 181155532-181155532 on Build GRCh38
rs767884326 Chr.5: 181155656-181155656 on Build GRCh38
rs774819263 Chr.5: 181155632-181155632 on Build GRCh38
rs573289347 Chr.5: 181155409-181155409 on Build GRCh38
rs751937981 Chr.5: 181155450-181155450 on Build GRCh38
rs780442071 Chr.5: 181155380-181155380 on Build GRCh38
rs774600605 Chr.5: 181155711-181155713 on Build GRCh38
rs900238640 Chr.5: 181155671-181155671 on Build GRCh38
rs754959139 Chr.5: 181155623-181155623 on Build GRCh38
rs759284677 Chr.5: 181155340-181155340 on Build GRCh38
rs1012044538 Chr.5: 181155512-181155512 on Build GRCh38
rs530541842 Chr.5: 181155667-181155667 on Build GRCh38
rs140710127 Chr.5: 181155278-181155278 on Build GRCh38
rs375774612 Chr.5: 181155556-181155556 on Build GRCh38
rs975497946 Chr.5: 181155704-181155704 on Build GRCh38
rs758728218 Chr.5: 181155379-181155379 on Build GRCh38
rs778791176 Chr.5: 181155495-181155495 on Build GRCh38
rs777736840 Chr.5: 181155398-181155398 on Build GRCh38
rs756197727 Chr.5: 181155471-181155471 on Build GRCh38
rs763486987 Chr.5: 181155432-181155432 on Build GRCh38
rs746616699 Chr.5: 181155613-181155613 on Build GRCh38
rs992549696 Chr.5: 181155561-181155561 on Build GRCh38
rs771825912 Chr.5: 181155524-181155524 on Build GRCh38
rs766860954 Chr.5: 181155564-181155564 on Build GRCh38
rs764124851 Chr.5: 181155676-181155676 on Build GRCh38
rs762804978 Chr.5: 181155333-181155333 on Build GRCh38
rs775061312 Chr.5: 181155410-181155410 on Build GRCh38
rs748249492 Chr.5: 181155394-181155394 on Build GRCh38
rs779732026 Chr.5: 181155609-181155609 on Build GRCh38
rs542253999 Chr.5: 181155719-181155719 on Build GRCh38
rs200000066 Chr.5: 181155385-181155385 on Build GRCh38
rs762167301 Chr.5: 181155363-181155363 on Build GRCh38
rs766061723 Chr.5: 181155513-181155513 on Build GRCh38
rs529325416 Chr.5: 181155310-181155310 on Build GRCh38
rs147854327 Chr.5: 181155309-181155309 on Build GRCh38
rs754179181 Chr.5: 181155677-181155677 on Build GRCh38
rs201743552 Chr.5: 181155573-181155573 on Build GRCh38
rs777738257 Chr.5: 181155476-181155476 on Build GRCh38
rs550860049 Chr.5: 181155680-181155680 on Build GRCh38
rs759984585 Chr.5: 181155642-181155642 on Build GRCh38
rs143315247 Chr.5: 181155460-181155460 on Build GRCh38
rs956619573 Chr.5: 181155352-181155352 on Build GRCh38
rs201466646 Chr.5: 181155411-181155411 on Build GRCh38
rs766666780 Chr.5: 181155434-181155434 on Build GRCh38
rs200506840 Chr.5: 181155362-181155362 on Build GRCh38
rs113166181 Chr.5: 181155557-181155557 on Build GRCh38
rs779843904 Chr.5: 181155706-181155706 on Build GRCh38
rs767094570 Chr.5: 181155343-181155343 on Build GRCh38
rs200724682 Chr.5: 181155681-181155681 on Build GRCh38
rs751745753 Chr.5: 181155382-181155382 on Build GRCh38
rs896147465 Chr.5: 181155498-181155498 on Build GRCh38
rs139903393 Chr.5: 181155568-181155568 on Build GRCh38
rs764207975 Chr.5: 181155675-181155675 on Build GRCh38
rs760894124 Chr.5: 181155669-181155669 on Build GRCh38
rs150156274 Chr.5: 181155306-181155306 on Build GRCh38
rs750915586 Chr.5: 181155428-181155428 on Build GRCh38
rs764168620 Chr.5: 181155592-181155592 on Build GRCh38
rs149585637 Chr.5: 181155407-181155407 on Build GRCh38
rs757263867 Chr.5: 181155485-181155485 on Build GRCh38
rs781078736 Chr.5: 181155386-181155386 on Build GRCh38
rs2546423 Chr.5: 181155604-181155604 on Build GRCh38
rs201633301 Chr.5: 181155387-181155387 on Build GRCh38
rs758818035 Chr.5: 181155294-181155294 on Build GRCh38
rs780246198 Chr.5: 181155299-181155299 on Build GRCh38
rs887376019 Chr.5: 181155464-181155464 on Build GRCh38
rs375556036 Chr.5: 181155619-181155619 on Build GRCh38
rs752904239 Chr.5: 181155470-181155470 on Build GRCh38

View Full Product Details