Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 2903096-2903588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB9
Entrez Gene ID
5272
Gene Name
serpin family B member 9
Gene Aliases
CAP-3, CAP3, PI-9, PI9
UniGene
Hs.104879
Gene Chromosome Location
Chr.6: 2887265-2903318 on Build GRCh38
Transcripts
NM_004155.5 XM_017010942.1

SNP Detail

SNP ID Chromosome Location
rs143439160 Chr.6: 2903487-2903487 on Build GRCh38
rs924529057 Chr.6: 2903457-2903457 on Build GRCh38
rs116695846 Chr.6: 2903346-2903346 on Build GRCh38
rs529001848 Chr.6: 2903400-2903400 on Build GRCh38
rs893097769 Chr.6: 2903471-2903471 on Build GRCh38
rs1054904283 Chr.6: 2903245-2903245 on Build GRCh38
rs1016537190 Chr.6: 2903552-2903552 on Build GRCh38
rs12192148 Chr.6: 2903278-2903278 on Build GRCh38
rs2236275 Chr.6: 2903347-2903347 on Build GRCh38
rs915703865 Chr.6: 2903140-2903140 on Build GRCh38
rs116210820 Chr.6: 2903275-2903275 on Build GRCh38
rs1039489346 Chr.6: 2903486-2903486 on Build GRCh38
rs966585859 Chr.6: 2903286-2903286 on Build GRCh38
rs766274024 Chr.6: 2903259-2903259 on Build GRCh38
rs949908193 Chr.6: 2903162-2903162 on Build GRCh38
rs115205633 Chr.6: 2903117-2903117 on Build GRCh38
rs113302547 Chr.6: 2903236-2903236 on Build GRCh38
rs541706285 Chr.6: 2903290-2903290 on Build GRCh38
rs903117541 Chr.6: 2903190-2903190 on Build GRCh38
rs991625389 Chr.6: 2903399-2903399 on Build GRCh38
rs1039718155 Chr.6: 2903343-2903343 on Build GRCh38
rs949854387 Chr.6: 2903420-2903420 on Build GRCh38
rs551633931 Chr.6: 2903473-2903473 on Build GRCh38
rs1022867330 Chr.6: 2903470-2903490 on Build GRCh38
rs867324645 Chr.6: 2903180-2903180 on Build GRCh38
rs548959886 Chr.6: 2903417-2903417 on Build GRCh38
rs556762114 Chr.6: 2903534-2903534 on Build GRCh38
rs111874467 Chr.6: 2903475-2903475 on Build GRCh38
rs934602406 Chr.6: 2903464-2903464 on Build GRCh38
rs576147176 Chr.6: 2903218-2903218 on Build GRCh38
rs986982964 Chr.6: 2903199-2903199 on Build GRCh38
rs2236276 Chr.6: 2903350-2903350 on Build GRCh38
rs998805933 Chr.6: 2903244-2903244 on Build GRCh38
rs950925274 Chr.6: 2903541-2903541 on Build GRCh38
rs1026521790 Chr.6: 2903512-2903512 on Build GRCh38
rs369250170 Chr.6: 2903369-2903369 on Build GRCh38
rs893203106 Chr.6: 2903269-2903269 on Build GRCh38
rs148566942 Chr.6: 2903229-2903240 on Build GRCh38
rs997675758 Chr.6: 2903509-2903509 on Build GRCh38
rs117052396 Chr.6: 2903432-2903432 on Build GRCh38
rs528768901 Chr.6: 2903279-2903279 on Build GRCh38
rs1045586645 Chr.6: 2903179-2903179 on Build GRCh38
rs1006504491 Chr.6: 2903546-2903546 on Build GRCh38
rs564652300 Chr.6: 2903337-2903337 on Build GRCh38
rs569739894 Chr.6: 2903388-2903388 on Build GRCh38
rs1026533132 Chr.6: 2903294-2903294 on Build GRCh38
rs1052951458 Chr.6: 2903425-2903425 on Build GRCh38
rs534369477 Chr.6: 2903437-2903437 on Build GRCh38

View Full Product Details