Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4117205-4117750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf201
Entrez Gene ID
404220
Gene Name
chromosome 6 open reading frame 201
Gene Aliases
dJ1013A10.5
UniGene
Hs.716731
Gene Chromosome Location
Chr.6: 4079197-4130788 on Build GRCh38
Transcripts
NM_001085401.2 NR_104463.1 NR_104464.1 XM_011514613.2 XM_017010861.1 NM_001166010.1 NM_006117.2 NM_206836.2 NR_028588.1 XM_006714957.1 XM_017010170.1

Gene Symbol
ECI2
Entrez Gene ID
10455
Gene Name
enoyl-CoA delta isomerase 2
Gene Aliases
ACBD2, DRS-1, DRS1, HCA88, PECI, dJ1013A10.3
UniGene
Hs.15250
Gene Chromosome Location
Chr.6: 4115693-4135597 on Build GRCh38
Transcripts
NM_001085401.2 NR_104463.1 NR_104464.1 XM_011514613.2 XM_017010861.1 NM_001166010.1 NM_006117.2 NM_206836.2 NR_028588.1 XM_006714957.1 XM_017010170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7270147 c.833C>T p.A278V Chr.6: 4117414-4117414 on Build GRCh38
4680084 c.939+2T>C p.? Chr.6: 4117306-4117306 on Build GRCh38
375089 c.894A>T p.K298N Chr.6: 4117353-4117353 on Build GRCh38
1719966 c.932C>T p.P311L Chr.6: 4117315-4117315 on Build GRCh38
6300337 c.977T>C p.F326S Chr.6: 4117360-4117360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984398252 Chr.6: 4117659-4117659 on Build GRCh38
rs781757435 Chr.6: 4117244-4117244 on Build GRCh38
rs769909134 Chr.6: 4117463-4117463 on Build GRCh38
rs1043229297 Chr.6: 4117251-4117251 on Build GRCh38
rs537845031 Chr.6: 4117650-4117650 on Build GRCh38
rs539719453 Chr.6: 4117250-4117250 on Build GRCh38
rs745779761 Chr.6: 4117394-4117394 on Build GRCh38
rs909811344 Chr.6: 4117403-4117403 on Build GRCh38
rs771936826 Chr.6: 4117398-4117398 on Build GRCh38
rs1044891649 Chr.6: 4117476-4117476 on Build GRCh38
rs760088765 Chr.6: 4117440-4117440 on Build GRCh38
rs1000779375 Chr.6: 4117515-4117515 on Build GRCh38
rs139209867 Chr.6: 4117408-4117408 on Build GRCh38
rs34743600 Chr.6: 4117570-4117571 on Build GRCh38
rs778930664 Chr.6: 4117482-4117482 on Build GRCh38
rs201522132 Chr.6: 4117442-4117442 on Build GRCh38
rs534479908 Chr.6: 4117675-4117675 on Build GRCh38
rs1051444200 Chr.6: 4117600-4117600 on Build GRCh38
rs950051354 Chr.6: 4117454-4117454 on Build GRCh38
rs756363416 Chr.6: 4117466-4117466 on Build GRCh38
rs369831173 Chr.6: 4117345-4117345 on Build GRCh38
rs768920826 Chr.6: 4117546-4117546 on Build GRCh38
rs754045004 Chr.6: 4117474-4117474 on Build GRCh38
rs527578254 Chr.6: 4117407-4117407 on Build GRCh38
rs976895519 Chr.6: 4117356-4117356 on Build GRCh38
rs147139055 Chr.6: 4117329-4117329 on Build GRCh38
rs142875686 Chr.6: 4117400-4117400 on Build GRCh38
rs7771538 Chr.6: 4117404-4117404 on Build GRCh38
rs775254989 Chr.6: 4117267-4117267 on Build GRCh38
rs978617134 Chr.6: 4117503-4117503 on Build GRCh38
rs968992555 Chr.6: 4117484-4117484 on Build GRCh38
rs749297618 Chr.6: 4117384-4117384 on Build GRCh38
rs753241882 Chr.6: 4117297-4117297 on Build GRCh38
rs752163718 Chr.6: 4117429-4117429 on Build GRCh38
rs555195689 Chr.6: 4117608-4117608 on Build GRCh38
rs768683178 Chr.6: 4117238-4117238 on Build GRCh38
rs1028495843 Chr.6: 4117645-4117645 on Build GRCh38
rs755971060 Chr.6: 4117431-4117431 on Build GRCh38
rs1010427089 Chr.6: 4117555-4117555 on Build GRCh38
rs1045201527 Chr.6: 4117706-4117706 on Build GRCh38
rs538442638 Chr.6: 4117595-4117595 on Build GRCh38
rs368401698 Chr.6: 4117274-4117274 on Build GRCh38
rs757373246 Chr.6: 4117475-4117475 on Build GRCh38
rs377338010 Chr.6: 4117363-4117363 on Build GRCh38
rs905299566 Chr.6: 4117708-4117708 on Build GRCh38
rs763694679 Chr.6: 4117289-4117289 on Build GRCh38
rs201392950 Chr.6: 4117360-4117360 on Build GRCh38
rs763571320 Chr.6: 4117323-4117323 on Build GRCh38
rs569690399 Chr.6: 4117545-4117545 on Build GRCh38
rs376879467 Chr.6: 4117263-4117263 on Build GRCh38
rs987801609 Chr.6: 4117264-4117264 on Build GRCh38
rs778877292 Chr.6: 4117386-4117386 on Build GRCh38
rs12557 Chr.6: 4117413-4117413 on Build GRCh38
rs3177264 Chr.6: 4117306-4117306 on Build GRCh38
rs752108872 Chr.6: 4117342-4117342 on Build GRCh38
rs569960938 Chr.6: 4117606-4117606 on Build GRCh38
rs764287981 Chr.6: 4117468-4117468 on Build GRCh38
rs549634942 Chr.6: 4117530-4117530 on Build GRCh38
rs1020574623 Chr.6: 4117298-4117298 on Build GRCh38
rs965432514 Chr.6: 4117607-4117607 on Build GRCh38
rs745832688 Chr.6: 4117490-4117490 on Build GRCh38
rs35202579 Chr.6: 4117373-4117373 on Build GRCh38
rs1020607320 Chr.6: 4117596-4117596 on Build GRCh38
rs776514196 Chr.6: 4117282-4117282 on Build GRCh38
rs997839584 Chr.6: 4117629-4117629 on Build GRCh38
rs554988683 Chr.6: 4117695-4117695 on Build GRCh38
rs114531537 Chr.6: 4117514-4117514 on Build GRCh38
rs972656881 Chr.6: 4117725-4117725 on Build GRCh38

View Full Product Details