Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7228916-7229420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RREB1
Entrez Gene ID
6239
Gene Name
ras responsive element binding protein 1
Gene Aliases
FINB, HNT, LZ321, RREB-1, Zep-1
UniGene
Hs.298248
Gene Chromosome Location
Chr.6: 7107597-7251980 on Build GRCh38
Transcripts
NM_001003698.3 NM_001003699.3 NM_001003700.1 NM_001168344.1 XM_006715157.3 XM_011514794.2 XM_011514795.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1137559 c.1048G>A p.A350T Chr.6: 7229147-7229147 on Build GRCh38
4722851 c.1123G>A p.V375I Chr.6: 7229222-7229222 on Build GRCh38
5996935 c.1318C>T p.Q440* Chr.6: 7229417-7229417 on Build GRCh38
1596475 c.1022C>T p.A341V Chr.6: 7229121-7229121 on Build GRCh38
5989719 c.1174C>G p.Q392E Chr.6: 7229273-7229273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769099559 Chr.6: 7229352-7229352 on Build GRCh38
rs140206154 Chr.6: 7229042-7229042 on Build GRCh38
rs751985080 Chr.6: 7229032-7229032 on Build GRCh38
rs376997025 Chr.6: 7228961-7228961 on Build GRCh38
rs750493154 Chr.6: 7229284-7229284 on Build GRCh38
rs748318952 Chr.6: 7229068-7229068 on Build GRCh38
rs779786535 Chr.6: 7229156-7229156 on Build GRCh38
rs373563472 Chr.6: 7228960-7228960 on Build GRCh38
rs9502563 Chr.6: 7228957-7228957 on Build GRCh38
rs773730596 Chr.6: 7229112-7229112 on Build GRCh38
rs751432487 Chr.6: 7229372-7229372 on Build GRCh38
rs758793624 Chr.6: 7229040-7229040 on Build GRCh38
rs757352942 Chr.6: 7229211-7229211 on Build GRCh38
rs766241536 Chr.6: 7229363-7229363 on Build GRCh38
rs761029336 Chr.6: 7229189-7229189 on Build GRCh38
rs146024995 Chr.6: 7229095-7229095 on Build GRCh38
rs745684594 Chr.6: 7229151-7229151 on Build GRCh38
rs761104833 Chr.6: 7229266-7229266 on Build GRCh38
rs562969847 Chr.6: 7229054-7229054 on Build GRCh38
rs756205550 Chr.6: 7229070-7229070 on Build GRCh38
rs747760972 Chr.6: 7229313-7229313 on Build GRCh38
rs759915310 Chr.6: 7229253-7229253 on Build GRCh38
rs142150270 Chr.6: 7229311-7229311 on Build GRCh38
rs149913845 Chr.6: 7229119-7229119 on Build GRCh38
rs1052194129 Chr.6: 7229216-7229216 on Build GRCh38
rs371245632 Chr.6: 7229385-7229385 on Build GRCh38
rs755285816 Chr.6: 7229118-7229118 on Build GRCh38
rs370083804 Chr.6: 7229168-7229168 on Build GRCh38
rs774662855 Chr.6: 7229173-7229173 on Build GRCh38
rs755270177 Chr.6: 7229043-7229043 on Build GRCh38
rs368606725 Chr.6: 7229384-7229384 on Build GRCh38
rs146744494 Chr.6: 7229312-7229312 on Build GRCh38
rs186056777 Chr.6: 7228992-7228992 on Build GRCh38
rs761989344 Chr.6: 7229358-7229358 on Build GRCh38
rs1046347213 Chr.6: 7228956-7228956 on Build GRCh38
rs943641761 Chr.6: 7229317-7229317 on Build GRCh38
rs371245191 Chr.6: 7229254-7229254 on Build GRCh38
rs935091827 Chr.6: 7228972-7228972 on Build GRCh38
rs2714315 Chr.6: 7229113-7229113 on Build GRCh38
rs750346771 Chr.6: 7229273-7229273 on Build GRCh38
rs150994044 Chr.6: 7229360-7229360 on Build GRCh38
rs746772631 Chr.6: 7229226-7229226 on Build GRCh38
rs758335458 Chr.6: 7229345-7229345 on Build GRCh38
rs770786364 Chr.6: 7228987-7228987 on Build GRCh38
rs763554481 Chr.6: 7229107-7229107 on Build GRCh38
rs1039941599 Chr.6: 7229383-7229383 on Build GRCh38
rs780035681 Chr.6: 7229308-7229308 on Build GRCh38
rs775760837 Chr.6: 7229262-7229262 on Build GRCh38
rs749360071 Chr.6: 7229137-7229137 on Build GRCh38
rs1016767690 Chr.6: 7229209-7229209 on Build GRCh38
rs1029421003 Chr.6: 7229154-7229154 on Build GRCh38
rs749202273 Chr.6: 7229074-7229074 on Build GRCh38
rs766731211 Chr.6: 7229019-7229019 on Build GRCh38
rs140980354 Chr.6: 7229362-7229362 on Build GRCh38
rs747378425 Chr.6: 7229351-7229351 on Build GRCh38
rs138825803 Chr.6: 7229390-7229390 on Build GRCh38
rs779762492 Chr.6: 7229224-7229224 on Build GRCh38
rs754042440 Chr.6: 7229270-7229270 on Build GRCh38
rs780735125 Chr.6: 7229399-7229399 on Build GRCh38
rs113114064 Chr.6: 7229123-7229123 on Build GRCh38
rs386696677 Chr.6: 7229324-7229326 on Build GRCh38
rs774340195 Chr.6: 7229078-7229078 on Build GRCh38
rs369785247 Chr.6: 7229080-7229080 on Build GRCh38
rs12192672 Chr.6: 7229386-7229386 on Build GRCh38
rs765181105 Chr.6: 7229278-7229278 on Build GRCh38
rs369853780 Chr.6: 7228980-7228980 on Build GRCh38
rs200496428 Chr.6: 7229380-7229380 on Build GRCh38
rs775130340 Chr.6: 7229005-7229005 on Build GRCh38
rs189495748 Chr.6: 7229240-7229240 on Build GRCh38
rs116483452 Chr.6: 7229071-7229071 on Build GRCh38
rs373354349 Chr.6: 7229222-7229222 on Build GRCh38
rs767737169 Chr.6: 7229181-7229181 on Build GRCh38
rs902058950 Chr.6: 7229335-7229335 on Build GRCh38
rs1016637229 Chr.6: 7229131-7229131 on Build GRCh38
rs147528961 Chr.6: 7229033-7229033 on Build GRCh38
rs115374113 Chr.6: 7229316-7229316 on Build GRCh38
rs758306467 Chr.6: 7229286-7229286 on Build GRCh38
rs748805448 Chr.6: 7229324-7229324 on Build GRCh38
rs774954588 Chr.6: 7229245-7229245 on Build GRCh38
rs764383122 Chr.6: 7229200-7229200 on Build GRCh38
rs114551633 Chr.6: 7229130-7229130 on Build GRCh38
rs778911018 Chr.6: 7229213-7229213 on Build GRCh38
rs771812048 Chr.6: 7229089-7229089 on Build GRCh38
rs754026145 Chr.6: 7229210-7229210 on Build GRCh38
rs778755255 Chr.6: 7229143-7229143 on Build GRCh38
rs770874252 Chr.6: 7229075-7229075 on Build GRCh38
rs773269265 Chr.6: 7228979-7228979 on Build GRCh38
rs376341337 Chr.6: 7229171-7229171 on Build GRCh38
rs139799223 Chr.6: 7229167-7229167 on Build GRCh38
rs774180539 Chr.6: 7228991-7228991 on Build GRCh38
rs747673605 Chr.6: 7229237-7229237 on Build GRCh38
rs376403896 Chr.6: 7229236-7229236 on Build GRCh38
rs769839254 Chr.6: 7228976-7228976 on Build GRCh38
rs756715537 Chr.6: 7229393-7229393 on Build GRCh38
rs750284070 Chr.6: 7229221-7229221 on Build GRCh38
rs369396664 Chr.6: 7228950-7228950 on Build GRCh38
rs375591269 Chr.6: 7229274-7229274 on Build GRCh38
rs964168526 Chr.6: 7229006-7229006 on Build GRCh38
rs201518822 Chr.6: 7228993-7228993 on Build GRCh38
rs899514750 Chr.6: 7229147-7229147 on Build GRCh38
rs368120167 Chr.6: 7229296-7229296 on Build GRCh38
rs752990584 Chr.6: 7229186-7229186 on Build GRCh38
rs764421899 Chr.6: 7229267-7229267 on Build GRCh38
rs759950901 Chr.6: 7229178-7229178 on Build GRCh38
rs781269289 Chr.6: 7229053-7229053 on Build GRCh38
rs772525626 Chr.6: 7229342-7229342 on Build GRCh38
rs752029956 Chr.6: 7229117-7229117 on Build GRCh38
rs751011488 Chr.6: 7228960-7228961 on Build GRCh38
rs774942973 Chr.6: 7229094-7229094 on Build GRCh38
rs768363513 Chr.6: 7229233-7229233 on Build GRCh38
rs761309732 Chr.6: 7229170-7229170 on Build GRCh38
rs114528151 Chr.6: 7229015-7229015 on Build GRCh38
rs550430739 Chr.6: 7229252-7229252 on Build GRCh38
rs747213952 Chr.6: 7228952-7228952 on Build GRCh38
rs771693035 Chr.6: 7229152-7229152 on Build GRCh38
rs113454423 Chr.6: 7229322-7229322 on Build GRCh38
rs765466558 Chr.6: 7229359-7229359 on Build GRCh38
rs527505375 Chr.6: 7229356-7229356 on Build GRCh38
rs757328415 Chr.6: 7229142-7229142 on Build GRCh38
rs756297366 Chr.6: 7229127-7229127 on Build GRCh38

View Full Product Details