Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10527516-10528019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT2
Entrez Gene ID
2651
Gene Name
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
Gene Aliases
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1
UniGene
Hs.519884
Gene Chromosome Location
Chr.6: 10521283-10629368 on Build GRCh38
Transcripts
NM_145649.4 XM_005248999.2 XM_006715052.3 XM_011514468.2

SNP Detail

SNP ID Chromosome Location
rs932109569 Chr.6: 10527799-10527799 on Build GRCh38
rs1040714197 Chr.6: 10527847-10527847 on Build GRCh38
rs1049760637 Chr.6: 10527884-10527884 on Build GRCh38
rs760129146 Chr.6: 10527845-10527845 on Build GRCh38
rs948386811 Chr.6: 10527696-10527696 on Build GRCh38
rs996845020 Chr.6: 10527742-10527742 on Build GRCh38
rs548483308 Chr.6: 10527792-10527792 on Build GRCh38
rs920376199 Chr.6: 10527839-10527839 on Build GRCh38
rs944513437 Chr.6: 10527871-10527871 on Build GRCh38
rs934908365 Chr.6: 10527680-10527680 on Build GRCh38
rs978740786 Chr.6: 10527986-10527986 on Build GRCh38
rs1014407748 Chr.6: 10527922-10527922 on Build GRCh38
rs927306827 Chr.6: 10527541-10527541 on Build GRCh38
rs993210693 Chr.6: 10527729-10527729 on Build GRCh38
rs201378348 Chr.6: 10527808-10527808 on Build GRCh38
rs1043331624 Chr.6: 10527776-10527776 on Build GRCh38
rs907731227 Chr.6: 10527857-10527857 on Build GRCh38
rs904485623 Chr.6: 10527885-10527885 on Build GRCh38
rs565354840 Chr.6: 10527612-10527612 on Build GRCh38
rs522587 Chr.6: 10527720-10527720 on Build GRCh38
rs1031847321 Chr.6: 10527765-10527765 on Build GRCh38
rs992953011 Chr.6: 10527964-10527964 on Build GRCh38
rs550513996 Chr.6: 10527564-10527564 on Build GRCh38
rs917497002 Chr.6: 10527796-10527796 on Build GRCh38
rs975920604 Chr.6: 10527585-10527585 on Build GRCh38
rs692986 Chr.6: 10527985-10527985 on Build GRCh38
rs142300856 Chr.6: 10527745-10527745 on Build GRCh38
rs913235084 Chr.6: 10527976-10527976 on Build GRCh38
rs544060820 Chr.6: 10527591-10527591 on Build GRCh38
rs373960822 Chr.6: 10527719-10527720 on Build GRCh38
rs896863009 Chr.6: 10527898-10527898 on Build GRCh38
rs930401077 Chr.6: 10527841-10527841 on Build GRCh38
rs1057132504 Chr.6: 10527573-10527573 on Build GRCh38
rs761168993 Chr.6: 10527876-10527879 on Build GRCh38
rs532698832 Chr.6: 10527664-10527664 on Build GRCh38
rs55823938 Chr.6: 10527949-10527949 on Build GRCh38
rs916921263 Chr.6: 10527693-10527693 on Build GRCh38
rs956222876 Chr.6: 10527910-10527910 on Build GRCh38
rs993501932 Chr.6: 10527793-10527793 on Build GRCh38
rs536829793 Chr.6: 10527876-10527876 on Build GRCh38
rs867183678 Chr.6: 10527779-10527779 on Build GRCh38
rs1026868800 Chr.6: 10527914-10527914 on Build GRCh38
rs997289670 Chr.6: 10527883-10527883 on Build GRCh38
rs181322189 Chr.6: 10527583-10527583 on Build GRCh38
rs374051478 Chr.6: 10527804-10527804 on Build GRCh38
rs772564185 Chr.6: 10527913-10527913 on Build GRCh38
rs952556432 Chr.6: 10527705-10527705 on Build GRCh38
rs1006017365 Chr.6: 10527555-10527569 on Build GRCh38
rs1047545050 Chr.6: 10527711-10527711 on Build GRCh38
rs994076255 Chr.6: 10527900-10527900 on Build GRCh38
rs913266356 Chr.6: 10527732-10527732 on Build GRCh38
rs142638998 Chr.6: 10527556-10527556 on Build GRCh38
rs1008270205 Chr.6: 10527606-10527606 on Build GRCh38
rs994618944 Chr.6: 10527568-10527568 on Build GRCh38
rs900398509 Chr.6: 10527880-10527880 on Build GRCh38
rs979792933 Chr.6: 10527834-10527834 on Build GRCh38
rs780031396 Chr.6: 10527576-10527576 on Build GRCh38
rs74971297 Chr.6: 10527706-10527706 on Build GRCh38
rs889551695 Chr.6: 10527604-10527604 on Build GRCh38
rs892022041 Chr.6: 10527574-10527574 on Build GRCh38
rs778081868 Chr.6: 10527621-10527621 on Build GRCh38
rs960388955 Chr.6: 10527960-10527960 on Build GRCh38
rs188858652 Chr.6: 10527582-10527582 on Build GRCh38
rs938646474 Chr.6: 10527562-10527562 on Build GRCh38
rs1001749303 Chr.6: 10527944-10527944 on Build GRCh38
rs775894734 Chr.6: 10527928-10527928 on Build GRCh38
rs1008770547 Chr.6: 10527721-10527722 on Build GRCh38
rs141470231 Chr.6: 10527717-10527718 on Build GRCh38
rs1043195333 Chr.6: 10527577-10527577 on Build GRCh38
rs948478583 Chr.6: 10527538-10527538 on Build GRCh38
rs547801975 Chr.6: 10527691-10527691 on Build GRCh38
rs1000301027 Chr.6: 10527888-10527888 on Build GRCh38
rs760976080 Chr.6: 10527929-10527929 on Build GRCh38
rs924892172 Chr.6: 10527665-10527665 on Build GRCh38
rs1018838495 Chr.6: 10527733-10527733 on Build GRCh38
rs147641128 Chr.6: 10527707-10527707 on Build GRCh38
rs951690573 Chr.6: 10527983-10527986 on Build GRCh38
rs543307917 Chr.6: 10527557-10527557 on Build GRCh38
rs569886446 Chr.6: 10527852-10527852 on Build GRCh38
rs978923771 Chr.6: 10527641-10527643 on Build GRCh38
rs1019303932 Chr.6: 10527617-10527617 on Build GRCh38
rs1044569358 Chr.6: 10527705-10527705 on Build GRCh38
rs901181998 Chr.6: 10527736-10527738 on Build GRCh38
rs978807515 Chr.6: 10527761-10527761 on Build GRCh38
rs968782133 Chr.6: 10527753-10527753 on Build GRCh38
rs1026815846 Chr.6: 10527698-10527698 on Build GRCh38

View Full Product Details