Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 12120984-12121494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP1
Entrez Gene ID
3096
Gene Name
human immunodeficiency virus type I enhancer binding protein 1
Gene Aliases
CIRIP, CRYBP1, GAAP, MBP-1, PRDII-BF1, Schnurri-1, ZAS1, ZNF40, ZNF40A
UniGene
Hs.567284
Gene Chromosome Location
Chr.6: 12002096-12212034 on Build GRCh38
Transcripts
NM_002114.3 XM_011514546.2 XM_011514547.2 XM_011514548.2 XM_011514549.2 XM_011514550.2 XM_011514551.2 XM_011514552.2 XM_011514553.1 XM_011514555.2 XM_017010800.1 XM_017010801.1 XM_017010802.1 XM_017010803.1 XM_017010804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5021491 c.1231T>C p.C411R Chr.6: 12121026-12121026 on Build GRCh38
1642839 c.1200_1201insA p.Q403fs*7 Chr.6: 12120995-12120996 on Build GRCh38
7313225 c.1276C>A p.R426S Chr.6: 12121071-12121071 on Build GRCh38
5003285 c.1241C>A p.A414E Chr.6: 12121036-12121036 on Build GRCh38
4635318 c.1342A>G p.N448D Chr.6: 12121137-12121137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766116990 Chr.6: 12121074-12121074 on Build GRCh38
rs769623031 Chr.6: 12121388-12121388 on Build GRCh38
rs760466347 Chr.6: 12121069-12121069 on Build GRCh38
rs374499754 Chr.6: 12121226-12121226 on Build GRCh38
rs1001160729 Chr.6: 12121269-12121269 on Build GRCh38
rs745487145 Chr.6: 12121386-12121386 on Build GRCh38
rs749709698 Chr.6: 12121358-12121358 on Build GRCh38
rs746586060 Chr.6: 12121285-12121285 on Build GRCh38
rs373677100 Chr.6: 12121030-12121030 on Build GRCh38
rs761828844 Chr.6: 12121445-12121445 on Build GRCh38
rs759356350 Chr.6: 12121294-12121294 on Build GRCh38
rs144854234 Chr.6: 12121160-12121160 on Build GRCh38
rs373558322 Chr.6: 12121259-12121259 on Build GRCh38
rs147927050 Chr.6: 12121322-12121322 on Build GRCh38
rs754148064 Chr.6: 12121467-12121467 on Build GRCh38
rs1040365078 Chr.6: 12121144-12121144 on Build GRCh38
rs781516905 Chr.6: 12121172-12121172 on Build GRCh38
rs775396723 Chr.6: 12121304-12121304 on Build GRCh38
rs746447641 Chr.6: 12121368-12121368 on Build GRCh38
rs41273116 Chr.6: 12121436-12121436 on Build GRCh38
rs776437287 Chr.6: 12121293-12121293 on Build GRCh38
rs879549638 Chr.6: 12121362-12121362 on Build GRCh38
rs757411103 Chr.6: 12121248-12121248 on Build GRCh38
rs753138437 Chr.6: 12121068-12121068 on Build GRCh38
rs756304931 Chr.6: 12121463-12121463 on Build GRCh38
rs977076204 Chr.6: 12121154-12121154 on Build GRCh38
rs377112392 Chr.6: 12121350-12121350 on Build GRCh38
rs767665966 Chr.6: 12121326-12121326 on Build GRCh38
rs370387597 Chr.6: 12121257-12121257 on Build GRCh38
rs576536946 Chr.6: 12121341-12121341 on Build GRCh38
rs542090941 Chr.6: 12121405-12121405 on Build GRCh38
rs746294103 Chr.6: 12121173-12121173 on Build GRCh38
rs1017189825 Chr.6: 12121460-12121460 on Build GRCh38
rs371106253 Chr.6: 12121163-12121163 on Build GRCh38
rs751711898 Chr.6: 12121246-12121246 on Build GRCh38
rs761377051 Chr.6: 12121051-12121051 on Build GRCh38
rs901941886 Chr.6: 12121196-12121196 on Build GRCh38
rs755374488 Chr.6: 12121472-12121472 on Build GRCh38
rs199639832 Chr.6: 12121260-12121260 on Build GRCh38
rs775976599 Chr.6: 12121179-12121179 on Build GRCh38
rs199702924 Chr.6: 12121231-12121231 on Build GRCh38
rs758421701 Chr.6: 12121151-12121151 on Build GRCh38
rs767127377 Chr.6: 12121055-12121055 on Build GRCh38
rs367826618 Chr.6: 12121227-12121227 on Build GRCh38
rs771500900 Chr.6: 12121206-12121206 on Build GRCh38
rs750539459 Chr.6: 12121462-12121462 on Build GRCh38
rs762779995 Chr.6: 12121316-12121316 on Build GRCh38
rs760211750 Chr.6: 12121211-12121211 on Build GRCh38
rs780487144 Chr.6: 12121263-12121263 on Build GRCh38
rs754944617 Chr.6: 12121109-12121109 on Build GRCh38
rs749788697 Chr.6: 12121266-12121266 on Build GRCh38
rs752758843 Chr.6: 12121142-12121142 on Build GRCh38
rs779540772 Chr.6: 12121365-12121365 on Build GRCh38
rs761326630 Chr.6: 12121197-12121197 on Build GRCh38
rs753671085 Chr.6: 12121220-12121220 on Build GRCh38
rs770339091 Chr.6: 12121013-12121013 on Build GRCh38
rs2228217 Chr.6: 12121380-12121380 on Build GRCh38
rs376161362 Chr.6: 12121105-12121105 on Build GRCh38
rs766104822 Chr.6: 12121217-12121217 on Build GRCh38
rs199542147 Chr.6: 12121464-12121464 on Build GRCh38
rs779050740 Chr.6: 12121129-12121129 on Build GRCh38
rs777787146 Chr.6: 12121280-12121280 on Build GRCh38
rs201586281 Chr.6: 12121364-12121364 on Build GRCh38
rs148587775 Chr.6: 12121420-12121420 on Build GRCh38
rs971010260 Chr.6: 12121100-12121100 on Build GRCh38
rs368028074 Chr.6: 12121300-12121300 on Build GRCh38
rs907790729 Chr.6: 12121433-12121433 on Build GRCh38
rs757366527 Chr.6: 12121323-12121323 on Build GRCh38
rs552930353 Chr.6: 12121369-12121369 on Build GRCh38
rs770702579 Chr.6: 12121292-12121292 on Build GRCh38
rs189758383 Chr.6: 12121441-12121441 on Build GRCh38
rs747228633 Chr.6: 12121164-12121164 on Build GRCh38
rs777687225 Chr.6: 12121240-12121240 on Build GRCh38
rs756206724 Chr.6: 12121071-12121071 on Build GRCh38
rs769243742 Chr.6: 12121267-12121267 on Build GRCh38
rs371593450 Chr.6: 12121346-12121346 on Build GRCh38
rs759409658 Chr.6: 12121223-12121223 on Build GRCh38
rs930468571 Chr.6: 12121395-12121395 on Build GRCh38
rs910335204 Chr.6: 12121250-12121250 on Build GRCh38
rs917006148 Chr.6: 12121094-12121094 on Build GRCh38
rs771227503 Chr.6: 12121167-12121167 on Build GRCh38

View Full Product Details