Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28325215-28325726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN31
Entrez Gene ID
64288
Gene Name
zinc finger and SCAN domain containing 31
Gene Aliases
ZNF20-Lp, ZNF310P, ZNF323
UniGene
Hs.656413
Gene Chromosome Location
Chr.6: 28324737-28358202 on Build GRCh38
Transcripts
NM_001135215.1 NM_001135216.1 NM_001243241.1 NM_001243242.1 NM_001243243.1 NM_001243244.1 NM_030899.4 NM_145909.2 XM_005249295.1 XM_005249296.4 XM_011514807.2 XM_011514808.2 XM_011514809.1 XM_011514811.2 XM_011514812.2 XM_011514813.2 XM_017011196.1

SNP Detail

SNP ID Chromosome Location
rs550898373 Chr.6: 28325647-28325647 on Build GRCh38
rs995870639 Chr.6: 28325341-28325341 on Build GRCh38
rs200897420 Chr.6: 28325633-28325633 on Build GRCh38
rs184360572 Chr.6: 28325255-28325255 on Build GRCh38
rs187765860 Chr.6: 28325310-28325310 on Build GRCh38
rs755291743 Chr.6: 28325595-28325595 on Build GRCh38
rs201699922 Chr.6: 28325472-28325473 on Build GRCh38
rs1045697183 Chr.6: 28325360-28325360 on Build GRCh38
rs917938061 Chr.6: 28325624-28325624 on Build GRCh38
rs963222775 Chr.6: 28325556-28325556 on Build GRCh38
rs758832486 Chr.6: 28325287-28325287 on Build GRCh38
rs1017923719 Chr.6: 28325502-28325502 on Build GRCh38
rs936856819 Chr.6: 28325665-28325665 on Build GRCh38
rs983249512 Chr.6: 28325611-28325611 on Build GRCh38
rs1026613284 Chr.6: 28325674-28325674 on Build GRCh38
rs565384029 Chr.6: 28325574-28325574 on Build GRCh38
rs191471537 Chr.6: 28325657-28325657 on Build GRCh38
rs973204101 Chr.6: 28325561-28325561 on Build GRCh38
rs943810946 Chr.6: 28325629-28325629 on Build GRCh38
rs758468944 Chr.6: 28325592-28325592 on Build GRCh38
rs922193162 Chr.6: 28325537-28325537 on Build GRCh38
rs536779296 Chr.6: 28325342-28325345 on Build GRCh38
rs545434920 Chr.6: 28325487-28325487 on Build GRCh38
rs866993250 Chr.6: 28325501-28325501 on Build GRCh38
rs759954727 Chr.6: 28325390-28325390 on Build GRCh38
rs1009286752 Chr.6: 28325443-28325443 on Build GRCh38
rs559746972 Chr.6: 28325386-28325386 on Build GRCh38
rs959242603 Chr.6: 28325350-28325350 on Build GRCh38
rs978187574 Chr.6: 28325470-28325470 on Build GRCh38
rs200096835 Chr.6: 28325470-28325470 on Build GRCh38
rs780294185 Chr.6: 28325385-28325385 on Build GRCh38
rs367764221 Chr.6: 28325471-28325472 on Build GRCh38
rs143618681 Chr.6: 28325631-28325632 on Build GRCh38
rs1051895463 Chr.6: 28325585-28325585 on Build GRCh38
rs140200831 Chr.6: 28325470-28325471 on Build GRCh38
rs896439631 Chr.6: 28325306-28325306 on Build GRCh38
rs917733913 Chr.6: 28325448-28325448 on Build GRCh38
rs1007950691 Chr.6: 28325265-28325265 on Build GRCh38
rs573264048 Chr.6: 28325437-28325437 on Build GRCh38
rs905440902 Chr.6: 28325663-28325663 on Build GRCh38
rs752019613 Chr.6: 28325397-28325397 on Build GRCh38
rs532149816 Chr.6: 28325575-28325575 on Build GRCh38
rs954500742 Chr.6: 28325490-28325490 on Build GRCh38
rs949402987 Chr.6: 28325642-28325642 on Build GRCh38
rs1032132744 Chr.6: 28325392-28325392 on Build GRCh38

View Full Product Details