Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 37316612-37317118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D22B
Entrez Gene ID
55633
Gene Name
TBC1 domain family member 22B
Gene Aliases
C6orf197
UniGene
Hs.731702
Gene Chromosome Location
Chr.6: 37257704-37332970 on Build GRCh38
Transcripts
NM_017772.3 NR_130108.1 XM_011514738.2 XM_011514739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5756849 c.1271T>C p.I424T Chr.6: 37316808-37316808 on Build GRCh38
5743132 c.1222C>T p.R408C Chr.6: 37316759-37316759 on Build GRCh38
4447290 c.1166-3C>T p.? Chr.6: 37316700-37316700 on Build GRCh38
3076529 c.1290T>A p.Y430* Chr.6: 37316827-37316827 on Build GRCh38
6228175 c.1274G>A p.R425H Chr.6: 37316811-37316811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576288878 Chr.6: 37316731-37316731 on Build GRCh38
rs749666077 Chr.6: 37316802-37316802 on Build GRCh38
rs919063339 Chr.6: 37316741-37316741 on Build GRCh38
rs746172442 Chr.6: 37316658-37316658 on Build GRCh38
rs775227536 Chr.6: 37316883-37316884 on Build GRCh38
rs780453792 Chr.6: 37316799-37316799 on Build GRCh38
rs763593173 Chr.6: 37317076-37317076 on Build GRCh38
rs765205125 Chr.6: 37316754-37316754 on Build GRCh38
rs748187893 Chr.6: 37316839-37316839 on Build GRCh38
rs754925073 Chr.6: 37316749-37316749 on Build GRCh38
rs993179150 Chr.6: 37317002-37317002 on Build GRCh38
rs1026047057 Chr.6: 37317030-37317030 on Build GRCh38
rs928679375 Chr.6: 37316773-37316773 on Build GRCh38
rs750171819 Chr.6: 37316686-37316686 on Build GRCh38
rs751088542 Chr.6: 37317079-37317079 on Build GRCh38
rs1007512742 Chr.6: 37316899-37316899 on Build GRCh38
rs773500330 Chr.6: 37316845-37316845 on Build GRCh38
rs553271265 Chr.6: 37317093-37317093 on Build GRCh38
rs199852239 Chr.6: 37316674-37316674 on Build GRCh38
rs760943446 Chr.6: 37316846-37316846 on Build GRCh38
rs748909156 Chr.6: 37316907-37316907 on Build GRCh38
rs904215838 Chr.6: 37316882-37316882 on Build GRCh38
rs777352451 Chr.6: 37316791-37316791 on Build GRCh38
rs767805885 Chr.6: 37316711-37316711 on Build GRCh38
rs762024925 Chr.6: 37316700-37316700 on Build GRCh38
rs1051426159 Chr.6: 37316781-37316781 on Build GRCh38
rs1009742633 Chr.6: 37316976-37316976 on Build GRCh38
rs3734330 Chr.6: 37317060-37317060 on Build GRCh38
rs187851344 Chr.6: 37316684-37316684 on Build GRCh38
rs773588312 Chr.6: 37316727-37316727 on Build GRCh38
rs762520820 Chr.6: 37317092-37317093 on Build GRCh38
rs1025189491 Chr.6: 37317007-37317007 on Build GRCh38
rs901477345 Chr.6: 37316969-37316969 on Build GRCh38
rs879069650 Chr.6: 37316788-37316788 on Build GRCh38
rs775797679 Chr.6: 37316666-37316666 on Build GRCh38
rs916131177 Chr.6: 37316645-37316645 on Build GRCh38
rs777437282 Chr.6: 37316654-37316654 on Build GRCh38
rs772936513 Chr.6: 37316823-37316823 on Build GRCh38
rs775027321 Chr.6: 37316804-37316804 on Build GRCh38
rs563711711 Chr.6: 37316875-37316875 on Build GRCh38
rs746682000 Chr.6: 37316794-37316794 on Build GRCh38
rs139229363 Chr.6: 37316961-37316961 on Build GRCh38
rs984996427 Chr.6: 37316640-37316640 on Build GRCh38
rs112831664 Chr.6: 37316993-37316993 on Build GRCh38
rs926599278 Chr.6: 37317013-37317013 on Build GRCh38
rs752165608 Chr.6: 37316760-37316760 on Build GRCh38
rs970796634 Chr.6: 37317008-37317008 on Build GRCh38
rs952163626 Chr.6: 37316638-37316638 on Build GRCh38
rs769256729 Chr.6: 37316803-37316803 on Build GRCh38
rs753638924 Chr.6: 37316744-37316744 on Build GRCh38
rs576887062 Chr.6: 37316717-37316717 on Build GRCh38
rs144137553 Chr.6: 37316774-37316774 on Build GRCh38
rs765113337 Chr.6: 37316881-37316881 on Build GRCh38
rs774552787 Chr.6: 37316692-37316692 on Build GRCh38
rs577596047 Chr.6: 37316950-37316950 on Build GRCh38
rs889022317 Chr.6: 37316861-37316861 on Build GRCh38
rs199901438 Chr.6: 37316847-37316847 on Build GRCh38
rs776462437 Chr.6: 37316850-37316850 on Build GRCh38
rs779796731 Chr.6: 37317010-37317010 on Build GRCh38
rs377540623 Chr.6: 37316843-37316843 on Build GRCh38
rs1039100364 Chr.6: 37316858-37316858 on Build GRCh38
rs770211621 Chr.6: 37316661-37316661 on Build GRCh38
rs756927511 Chr.6: 37316798-37316798 on Build GRCh38

View Full Product Details