Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41587219-41587717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP4
Entrez Gene ID
116113
Gene Name
forkhead box P4
Gene Aliases
hFKHLA
UniGene
Hs.131436
Gene Chromosome Location
Chr.6: 41546426-41602384 on Build GRCh38
Transcripts
NM_001012426.1 NM_001012427.1 NM_138457.2 XM_006714991.3 XM_011514289.2 XM_011514290.2 XM_011514291.2 XM_011514292.2 XM_011514293.2 XM_017010233.1 XM_017010234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1079220 c.805C>T p.P269S Chr.6: 41587448-41587448 on Build GRCh38
6889004 c.814C>T p.L272F Chr.6: 41587454-41587454 on Build GRCh38
5756862 c.811_812insA p.L271fs*44 Chr.6: 41587454-41587455 on Build GRCh38
6988910 c.793C>T p.P265S Chr.6: 41587433-41587433 on Build GRCh38
3783986 c.862C>T p.R288W Chr.6: 41587502-41587502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961561112 Chr.6: 41587473-41587473 on Build GRCh38
rs768344957 Chr.6: 41587452-41587452 on Build GRCh38
rs200300359 Chr.6: 41587340-41587340 on Build GRCh38
rs567186191 Chr.6: 41587537-41587537 on Build GRCh38
rs751465280 Chr.6: 41587343-41587343 on Build GRCh38
rs911613561 Chr.6: 41587689-41587689 on Build GRCh38
rs762339292 Chr.6: 41587410-41587410 on Build GRCh38
rs570902052 Chr.6: 41587460-41587460 on Build GRCh38
rs896923412 Chr.6: 41587621-41587621 on Build GRCh38
rs765089602 Chr.6: 41587310-41587310 on Build GRCh38
rs530916537 Chr.6: 41587431-41587431 on Build GRCh38
rs557410453 Chr.6: 41587619-41587619 on Build GRCh38
rs116862966 Chr.6: 41587655-41587655 on Build GRCh38
rs1007168980 Chr.6: 41587543-41587543 on Build GRCh38
rs139784428 Chr.6: 41587433-41587433 on Build GRCh38
rs747127172 Chr.6: 41587263-41587263 on Build GRCh38
rs1009687571 Chr.6: 41587579-41587579 on Build GRCh38
rs200703463 Chr.6: 41587363-41587363 on Build GRCh38
rs140659341 Chr.6: 41587372-41587372 on Build GRCh38
rs189252979 Chr.6: 41587649-41587649 on Build GRCh38
rs374970122 Chr.6: 41587554-41587554 on Build GRCh38
rs199766169 Chr.6: 41587451-41587451 on Build GRCh38
rs926873281 Chr.6: 41587634-41587634 on Build GRCh38
rs199781164 Chr.6: 41587348-41587348 on Build GRCh38
rs1015772056 Chr.6: 41587238-41587238 on Build GRCh38
rs758748324 Chr.6: 41587430-41587430 on Build GRCh38
rs377455960 Chr.6: 41587454-41587454 on Build GRCh38
rs781449571 Chr.6: 41587404-41587404 on Build GRCh38
rs1045338018 Chr.6: 41587275-41587275 on Build GRCh38
rs367628227 Chr.6: 41587309-41587309 on Build GRCh38
rs537591559 Chr.6: 41587573-41587573 on Build GRCh38
rs747986308 Chr.6: 41587524-41587524 on Build GRCh38
rs544247285 Chr.6: 41587262-41587262 on Build GRCh38
rs775254201 Chr.6: 41587549-41587549 on Build GRCh38
rs758240552 Chr.6: 41587447-41587448 on Build GRCh38
rs201392271 Chr.6: 41587352-41587352 on Build GRCh38
rs144456961 Chr.6: 41587437-41587437 on Build GRCh38
rs553327432 Chr.6: 41587657-41587657 on Build GRCh38
rs1051551781 Chr.6: 41587683-41587683 on Build GRCh38
rs750905734 Chr.6: 41587563-41587563 on Build GRCh38
rs562659251 Chr.6: 41587409-41587409 on Build GRCh38
rs200927605 Chr.6: 41587625-41587634 on Build GRCh38
rs781448348 Chr.6: 41587434-41587434 on Build GRCh38
rs1036772716 Chr.6: 41587620-41587620 on Build GRCh38
rs201340306 Chr.6: 41587546-41587546 on Build GRCh38
rs948648415 Chr.6: 41587600-41587600 on Build GRCh38
rs1027166668 Chr.6: 41587632-41587632 on Build GRCh38
rs761677679 Chr.6: 41587413-41587413 on Build GRCh38
rs374742106 Chr.6: 41587264-41587264 on Build GRCh38
rs141986171 Chr.6: 41587415-41587415 on Build GRCh38
rs768879722 Chr.6: 41587506-41587506 on Build GRCh38
rs759137856 Chr.6: 41587545-41587545 on Build GRCh38
rs199908081 Chr.6: 41587373-41587373 on Build GRCh38
rs776786302 Chr.6: 41587375-41587375 on Build GRCh38
rs202150520 Chr.6: 41587351-41587351 on Build GRCh38
rs749895513 Chr.6: 41587416-41587416 on Build GRCh38
rs780120879 Chr.6: 41587503-41587503 on Build GRCh38
rs963592430 Chr.6: 41587648-41587648 on Build GRCh38
rs761269429 Chr.6: 41587605-41587605 on Build GRCh38
rs777052553 Chr.6: 41587271-41587271 on Build GRCh38
rs762337588 Chr.6: 41587553-41587553 on Build GRCh38
rs752748756 Chr.6: 41587418-41587419 on Build GRCh38
rs550995575 Chr.6: 41587636-41587637 on Build GRCh38
rs1044000870 Chr.6: 41587603-41587603 on Build GRCh38
rs11267985 Chr.6: 41587626-41587636 on Build GRCh38
rs768002278 Chr.6: 41587493-41587493 on Build GRCh38
rs775110588 Chr.6: 41587330-41587330 on Build GRCh38
rs956063982 Chr.6: 41587578-41587578 on Build GRCh38
rs113771840 Chr.6: 41587674-41587674 on Build GRCh38
rs142575732 Chr.6: 41587502-41587502 on Build GRCh38
rs146299057 Chr.6: 41587429-41587429 on Build GRCh38
rs866835572 Chr.6: 41587407-41587407 on Build GRCh38
rs868669871 Chr.6: 41587614-41587614 on Build GRCh38
rs573858076 Chr.6: 41587664-41587664 on Build GRCh38
rs200305782 Chr.6: 41587377-41587377 on Build GRCh38
rs764585288 Chr.6: 41587466-41587466 on Build GRCh38
rs748410352 Chr.6: 41587364-41587364 on Build GRCh38
rs890373747 Chr.6: 41587398-41587398 on Build GRCh38
rs560002245 Chr.6: 41587353-41587353 on Build GRCh38
rs1044052878 Chr.6: 41587414-41587414 on Build GRCh38
rs961891630 Chr.6: 41587253-41587253 on Build GRCh38
rs1025465661 Chr.6: 41587585-41587585 on Build GRCh38
rs757678562 Chr.6: 41587483-41587483 on Build GRCh38
rs35584312 Chr.6: 41587474-41587474 on Build GRCh38
rs1054370621 Chr.6: 41587388-41587388 on Build GRCh38
rs755389584 Chr.6: 41587250-41587250 on Build GRCh38
rs547351834 Chr.6: 41587520-41587520 on Build GRCh38
rs769863758 Chr.6: 41587395-41587395 on Build GRCh38
rs780204198 Chr.6: 41587447-41587447 on Build GRCh38
rs1007471860 Chr.6: 41587411-41587411 on Build GRCh38
rs770352084 Chr.6: 41587541-41587541 on Build GRCh38
rs942191751 Chr.6: 41587488-41587488 on Build GRCh38
rs375722090 Chr.6: 41587423-41587423 on Build GRCh38
rs778211894 Chr.6: 41587369-41587369 on Build GRCh38
rs1027222657 Chr.6: 41587477-41587477 on Build GRCh38
rs771821096 Chr.6: 41587534-41587534 on Build GRCh38
rs920208250 Chr.6: 41587266-41587266 on Build GRCh38
rs755812624 Chr.6: 41587422-41587422 on Build GRCh38
rs372509837 Chr.6: 41587638-41587648 on Build GRCh38
rs999993883 Chr.6: 41587567-41587567 on Build GRCh38
rs1032859110 Chr.6: 41587436-41587436 on Build GRCh38
rs781775268 Chr.6: 41587484-41587484 on Build GRCh38
rs775762406 Chr.6: 41587405-41587405 on Build GRCh38
rs750938751 Chr.6: 41587490-41587490 on Build GRCh38
rs965362671 Chr.6: 41587673-41587673 on Build GRCh38
rs762973948 Chr.6: 41587339-41587339 on Build GRCh38
rs555684381 Chr.6: 41587542-41587542 on Build GRCh38
rs144343674 Chr.6: 41587453-41587453 on Build GRCh38
rs11758591 Chr.6: 41587255-41587255 on Build GRCh38
rs759774383 Chr.6: 41587297-41587297 on Build GRCh38
rs536117974 Chr.6: 41587540-41587540 on Build GRCh38
rs779084604 Chr.6: 41587254-41587254 on Build GRCh38
rs762700013 Chr.6: 41587408-41587408 on Build GRCh38
rs753448087 Chr.6: 41587455-41587455 on Build GRCh38

View Full Product Details