Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151660656-151661153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX27
Entrez Gene ID
81609
Gene Name
sorting nexin family member 27
Gene Aliases
MRT1, MY014
UniGene
Hs.192326
Gene Chromosome Location
Chr.1: 151612027-151699083 on Build GRCh38
Transcripts
NM_030918.5 XM_005245509.2 XM_005245510.3 XM_005245511.4 XM_011510024.2 XM_011510025.2 XM_011510026.2 XM_017002417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6786530 c.469C>T p.R157* Chr.1: 151660809-151660809 on Build GRCh38
3688989 c.458-1G>T p.? Chr.1: 151660797-151660797 on Build GRCh38
6274253 c.801+1G>T p.? Chr.1: 151660863-151660863 on Build GRCh38
6255998 c.801+5G>T p.? Chr.1: 151660867-151660867 on Build GRCh38
6786535 c.746T>C p.I249T Chr.1: 151660807-151660807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745510590 Chr.1: 151660847-151660847 on Build GRCh38
rs370465109 Chr.1: 151660735-151660735 on Build GRCh38
rs915905974 Chr.1: 151660740-151660740 on Build GRCh38
rs1030343321 Chr.1: 151660792-151660792 on Build GRCh38
rs949056408 Chr.1: 151660976-151660976 on Build GRCh38
rs1035871222 Chr.1: 151660679-151660679 on Build GRCh38
rs752497304 Chr.1: 151660785-151660785 on Build GRCh38
rs567304344 Chr.1: 151661072-151661072 on Build GRCh38
rs755975481 Chr.1: 151660793-151660793 on Build GRCh38
rs1043172592 Chr.1: 151661029-151661029 on Build GRCh38
rs917610630 Chr.1: 151660974-151660975 on Build GRCh38
rs772609394 Chr.1: 151660856-151660856 on Build GRCh38
rs999390991 Chr.1: 151661065-151661065 on Build GRCh38
rs552244633 Chr.1: 151660881-151660881 on Build GRCh38
rs764061917 Chr.1: 151660803-151660803 on Build GRCh38
rs990947240 Chr.1: 151660960-151660960 on Build GRCh38
rs777119235 Chr.1: 151660877-151660877 on Build GRCh38
rs933215056 Chr.1: 151660940-151660940 on Build GRCh38
rs187462607 Chr.1: 151660711-151660711 on Build GRCh38
rs908638420 Chr.1: 151660851-151660851 on Build GRCh38
rs374312634 Chr.1: 151660924-151660924 on Build GRCh38
rs769067271 Chr.1: 151660859-151660859 on Build GRCh38
rs770764167 Chr.1: 151661087-151661097 on Build GRCh38
rs942260205 Chr.1: 151661105-151661105 on Build GRCh38
rs779687444 Chr.1: 151660857-151660857 on Build GRCh38
rs190457089 Chr.1: 151660795-151660795 on Build GRCh38
rs779601993 Chr.1: 151660691-151660691 on Build GRCh38
rs766649395 Chr.1: 151660781-151660781 on Build GRCh38
rs902942823 Chr.1: 151661044-151661044 on Build GRCh38
rs146222009 Chr.1: 151660833-151660833 on Build GRCh38
rs763121024 Chr.1: 151660904-151660904 on Build GRCh38
rs771626737 Chr.1: 151660908-151660910 on Build GRCh38
rs1050360865 Chr.1: 151660997-151660997 on Build GRCh38
rs769728243 Chr.1: 151660989-151660989 on Build GRCh38
rs550317831 Chr.1: 151660734-151660734 on Build GRCh38
rs376971230 Chr.1: 151660908-151660908 on Build GRCh38
rs888767727 Chr.1: 151661011-151661011 on Build GRCh38
rs1011261817 Chr.1: 151661015-151661015 on Build GRCh38
rs923136110 Chr.1: 151660917-151660917 on Build GRCh38
rs115780664 Chr.1: 151660880-151660880 on Build GRCh38
rs773119919 Chr.1: 151660894-151660894 on Build GRCh38
rs370865331 Chr.1: 151660773-151660773 on Build GRCh38
rs796184993 Chr.1: 151661119-151661119 on Build GRCh38
rs566412454 Chr.1: 151660906-151660906 on Build GRCh38
rs960232006 Chr.1: 151661110-151661110 on Build GRCh38
rs778326282 Chr.1: 151660835-151660835 on Build GRCh38
rs753881769 Chr.1: 151660813-151660813 on Build GRCh38

View Full Product Details