Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152349490-152350033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Gene Symbol
FLG2
Entrez Gene ID
388698
Gene Name
filaggrin family member 2
Gene Aliases
IFPS
UniGene
Hs.156124
Gene Chromosome Location
Chr.1: 152348735-152360028 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

SNP Detail

SNP ID Chromosome Location
rs74456239 Chr.1: 152349889-152349889 on Build GRCh38
rs752175863 Chr.1: 152349538-152349538 on Build GRCh38
rs543319532 Chr.1: 152349858-152349858 on Build GRCh38
rs79507400 Chr.1: 152349774-152349774 on Build GRCh38
rs747396124 Chr.1: 152349684-152349684 on Build GRCh38
rs150033871 Chr.1: 152349852-152349852 on Build GRCh38
rs923521801 Chr.1: 152349931-152349931 on Build GRCh38
rs368758486 Chr.1: 152349536-152349536 on Build GRCh38
rs1039323452 Chr.1: 152349619-152349619 on Build GRCh38
rs922369109 Chr.1: 152349668-152349668 on Build GRCh38
rs960248577 Chr.1: 152349826-152349826 on Build GRCh38
rs574243070 Chr.1: 152349854-152349854 on Build GRCh38
rs531411182 Chr.1: 152349521-152349521 on Build GRCh38
rs563478563 Chr.1: 152349867-152349867 on Build GRCh38
rs1003593504 Chr.1: 152349848-152349848 on Build GRCh38
rs758039024 Chr.1: 152349553-152349553 on Build GRCh38
rs947134770 Chr.1: 152350001-152350001 on Build GRCh38
rs746397391 Chr.1: 152349857-152349857 on Build GRCh38
rs572513799 Chr.1: 152349859-152349859 on Build GRCh38
rs932349329 Chr.1: 152349695-152349695 on Build GRCh38
rs946943311 Chr.1: 152349809-152349809 on Build GRCh38
rs952587298 Chr.1: 152349537-152349537 on Build GRCh38
rs913425614 Chr.1: 152349983-152349983 on Build GRCh38
rs189315357 Chr.1: 152349561-152349561 on Build GRCh38
rs976610360 Chr.1: 152349974-152349974 on Build GRCh38
rs964708853 Chr.1: 152349923-152349923 on Build GRCh38
rs990825538 Chr.1: 152349532-152349532 on Build GRCh38
rs919319060 Chr.1: 152349525-152349525 on Build GRCh38
rs768827285 Chr.1: 152349988-152349988 on Build GRCh38
rs375646543 Chr.1: 152349569-152349569 on Build GRCh38
rs1035032873 Chr.1: 152349520-152349520 on Build GRCh38
rs984779592 Chr.1: 152349563-152349563 on Build GRCh38
rs185715055 Chr.1: 152349817-152349817 on Build GRCh38
rs961234612 Chr.1: 152349965-152349965 on Build GRCh38
rs993986694 Chr.1: 152349840-152349840 on Build GRCh38
rs367983405 Chr.1: 152349796-152349796 on Build GRCh38
rs923524685 Chr.1: 152349633-152349636 on Build GRCh38
rs1005447356 Chr.1: 152349906-152349906 on Build GRCh38
rs983983154 Chr.1: 152349539-152349539 on Build GRCh38
rs11204982 Chr.1: 152349566-152349566 on Build GRCh38
rs78056768 Chr.1: 152349816-152349816 on Build GRCh38
rs953808509 Chr.1: 152349991-152349991 on Build GRCh38
rs763111847 Chr.1: 152349954-152349954 on Build GRCh38
rs181894175 Chr.1: 152349587-152349587 on Build GRCh38
rs891515600 Chr.1: 152349688-152349688 on Build GRCh38
rs1027640765 Chr.1: 152349871-152349871 on Build GRCh38
rs933462911 Chr.1: 152349645-152349645 on Build GRCh38
rs538703427 Chr.1: 152349758-152349758 on Build GRCh38
rs781351873 Chr.1: 152349795-152349795 on Build GRCh38
rs975150731 Chr.1: 152349927-152349927 on Build GRCh38
rs115279484 Chr.1: 152349651-152349651 on Build GRCh38
rs939770394 Chr.1: 152349620-152349620 on Build GRCh38
rs1042685137 Chr.1: 152349717-152349717 on Build GRCh38
rs909206300 Chr.1: 152349913-152349913 on Build GRCh38

View Full Product Details