Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42104714-42105240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf132
Entrez Gene ID
647024
Gene Name
chromosome 6 open reading frame 132
Gene Aliases
bA7K24.2
UniGene
Hs.444277
Gene Chromosome Location
Chr.6: 42101119-42142977 on Build GRCh38
Transcripts
NM_001164446.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5583338 c.3183C>T p.L1061L Chr.6: 42104729-42104729 on Build GRCh38
4153390 c.3030C>T p.S1010S Chr.6: 42104882-42104882 on Build GRCh38
5786795 c.2976C>T p.F992F Chr.6: 42104936-42104936 on Build GRCh38
5746578 c.3014A>T p.Y1005F Chr.6: 42104898-42104898 on Build GRCh38
5726521 c.2865C>T p.P955P Chr.6: 42105047-42105047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767543354 Chr.6: 42105192-42105192 on Build GRCh38
rs764476138 Chr.6: 42105185-42105185 on Build GRCh38
rs745628273 Chr.6: 42104758-42104758 on Build GRCh38
rs946293342 Chr.6: 42104843-42104843 on Build GRCh38
rs763584177 Chr.6: 42105171-42105171 on Build GRCh38
rs777538356 Chr.6: 42104972-42104980 on Build GRCh38
rs554441543 Chr.6: 42105052-42105052 on Build GRCh38
rs745398490 Chr.6: 42105189-42105189 on Build GRCh38
rs556246123 Chr.6: 42104771-42104771 on Build GRCh38
rs773725456 Chr.6: 42104766-42104766 on Build GRCh38
rs914601209 Chr.6: 42105132-42105132 on Build GRCh38
rs904727575 Chr.6: 42104891-42104891 on Build GRCh38
rs372314325 Chr.6: 42105113-42105113 on Build GRCh38
rs201213632 Chr.6: 42104890-42104890 on Build GRCh38
rs921581995 Chr.6: 42104789-42104789 on Build GRCh38
rs921922989 Chr.6: 42105094-42105094 on Build GRCh38
rs1002579306 Chr.6: 42104939-42104939 on Build GRCh38
rs775701868 Chr.6: 42105166-42105166 on Build GRCh38
rs1034457540 Chr.6: 42104896-42104896 on Build GRCh38
rs775515263 Chr.6: 42104760-42104760 on Build GRCh38
rs921657682 Chr.6: 42105036-42105036 on Build GRCh38
rs750645269 Chr.6: 42105200-42105200 on Build GRCh38
rs527623068 Chr.6: 42104872-42104872 on Build GRCh38
rs911585892 Chr.6: 42105072-42105072 on Build GRCh38
rs941775644 Chr.6: 42104769-42104769 on Build GRCh38
rs757968574 Chr.6: 42104972-42104974 on Build GRCh38
rs890678218 Chr.6: 42104841-42104841 on Build GRCh38
rs193099422 Chr.6: 42104960-42104960 on Build GRCh38
rs911731444 Chr.6: 42104737-42104760 on Build GRCh38
rs751633338 Chr.6: 42104969-42104969 on Build GRCh38
rs748442086 Chr.6: 42104975-42104977 on Build GRCh38
rs987114583 Chr.6: 42105065-42105065 on Build GRCh38
rs976044308 Chr.6: 42104807-42104807 on Build GRCh38
rs747640125 Chr.6: 42105123-42105123 on Build GRCh38
rs760002910 Chr.6: 42104893-42104893 on Build GRCh38
rs765646309 Chr.6: 42105145-42105145 on Build GRCh38
rs973226969 Chr.6: 42105027-42105027 on Build GRCh38
rs368362618 Chr.6: 42105074-42105074 on Build GRCh38
rs983740965 Chr.6: 42104764-42104764 on Build GRCh38
rs74901308 Chr.6: 42104954-42104954 on Build GRCh38
rs931623940 Chr.6: 42104800-42104800 on Build GRCh38
rs372744791 Chr.6: 42105208-42105208 on Build GRCh38
rs756280141 Chr.6: 42105201-42105201 on Build GRCh38
rs574344855 Chr.6: 42105137-42105137 on Build GRCh38
rs1026713143 Chr.6: 42104973-42104973 on Build GRCh38
rs1037441117 Chr.6: 42104773-42104773 on Build GRCh38
rs868289022 Chr.6: 42105193-42105193 on Build GRCh38
rs369241153 Chr.6: 42105128-42105128 on Build GRCh38
rs767249931 Chr.6: 42104889-42104889 on Build GRCh38
rs1054245138 Chr.6: 42104836-42104836 on Build GRCh38
rs757347835 Chr.6: 42105191-42105191 on Build GRCh38
rs755728475 Chr.6: 42105141-42105141 on Build GRCh38
rs922057098 Chr.6: 42104809-42104809 on Build GRCh38
rs780791033 Chr.6: 42105110-42105110 on Build GRCh38
rs377009471 Chr.6: 42105122-42105122 on Build GRCh38
rs185573419 Chr.6: 42104971-42104971 on Build GRCh38
rs1000268462 Chr.6: 42104892-42104892 on Build GRCh38
rs1012444857 Chr.6: 42104755-42104755 on Build GRCh38
rs907108291 Chr.6: 42104920-42104920 on Build GRCh38
rs952251030 Chr.6: 42104983-42104983 on Build GRCh38
rs150599390 Chr.6: 42104972-42104977 on Build GRCh38
rs747791855 Chr.6: 42104990-42104990 on Build GRCh38
rs749308841 Chr.6: 42105105-42105105 on Build GRCh38
rs201999772 Chr.6: 42105081-42105081 on Build GRCh38
rs769769064 Chr.6: 42104759-42104759 on Build GRCh38
rs547708981 Chr.6: 42104978-42104978 on Build GRCh38
rs771126330 Chr.6: 42104747-42104747 on Build GRCh38
rs1015209209 Chr.6: 42105009-42105009 on Build GRCh38
rs113527767 Chr.6: 42104763-42104763 on Build GRCh38
rs781367520 Chr.6: 42105037-42105037 on Build GRCh38
rs932049334 Chr.6: 42104824-42104824 on Build GRCh38
rs747245892 Chr.6: 42104746-42104746 on Build GRCh38
rs764291728 Chr.6: 42104968-42104968 on Build GRCh38
rs1055987854 Chr.6: 42105158-42105158 on Build GRCh38
rs781030787 Chr.6: 42104754-42104754 on Build GRCh38
rs966077790 Chr.6: 42104784-42104784 on Build GRCh38
rs372924845 Chr.6: 42104945-42104945 on Build GRCh38
rs908078988 Chr.6: 42104777-42104777 on Build GRCh38

View Full Product Details