Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46248582-46249116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCAN2
Entrez Gene ID
10231
Gene Name
regulator of calcineurin 2
Gene Aliases
CSP2, DSCR1L1, MCIP2, RCN2, ZAKI-4, ZAKI4
UniGene
Hs.440168
Gene Chromosome Location
Chr.6: 46220730-46492067 on Build GRCh38
Transcripts
NM_001251973.1 NM_001251974.1 NM_005822.3 XM_011514226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1728379 c.106T>C p.F36L Chr.6: 46248878-46248878 on Build GRCh38
4490270 c.360C>T p.T120T Chr.6: 46248762-46248762 on Build GRCh38
1643093 c.132G>A p.T44T Chr.6: 46248852-46248852 on Build GRCh38
1187047 c.301C>A p.R101S Chr.6: 46248821-46248821 on Build GRCh38
5870813 c.239G>A p.G80E Chr.6: 46248883-46248883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778895039 Chr.6: 46248722-46248722 on Build GRCh38
rs575309512 Chr.6: 46248660-46248660 on Build GRCh38
rs375007430 Chr.6: 46249092-46249092 on Build GRCh38
rs745608061 Chr.6: 46248918-46248918 on Build GRCh38
rs758968874 Chr.6: 46248708-46248708 on Build GRCh38
rs369113192 Chr.6: 46248852-46248852 on Build GRCh38
rs764192474 Chr.6: 46248832-46248832 on Build GRCh38
rs964535584 Chr.6: 46248758-46248758 on Build GRCh38
rs559639342 Chr.6: 46248874-46248874 on Build GRCh38
rs879637482 Chr.6: 46249061-46249061 on Build GRCh38
rs989209804 Chr.6: 46248853-46248853 on Build GRCh38
rs557124120 Chr.6: 46248639-46248639 on Build GRCh38
rs546359032 Chr.6: 46248664-46248664 on Build GRCh38
rs577549173 Chr.6: 46248633-46248633 on Build GRCh38
rs937720205 Chr.6: 46248623-46248623 on Build GRCh38
rs201045399 Chr.6: 46248875-46248875 on Build GRCh38
rs887337959 Chr.6: 46248624-46248624 on Build GRCh38
rs1026020003 Chr.6: 46248670-46248670 on Build GRCh38
rs764247757 Chr.6: 46248684-46248684 on Build GRCh38
rs899617794 Chr.6: 46248682-46248682 on Build GRCh38
rs1006146142 Chr.6: 46248726-46248726 on Build GRCh38
rs200662612 Chr.6: 46248927-46248927 on Build GRCh38
rs752115055 Chr.6: 46248885-46248885 on Build GRCh38
rs776671267 Chr.6: 46248843-46248843 on Build GRCh38
rs949197775 Chr.6: 46249018-46249018 on Build GRCh38
rs760684422 Chr.6: 46248818-46248818 on Build GRCh38
rs376237384 Chr.6: 46248787-46248787 on Build GRCh38
rs368221717 Chr.6: 46248947-46248947 on Build GRCh38
rs927415392 Chr.6: 46248953-46248953 on Build GRCh38
rs779930446 Chr.6: 46248935-46248935 on Build GRCh38
rs201620447 Chr.6: 46248735-46248735 on Build GRCh38
rs554279752 Chr.6: 46248695-46248695 on Build GRCh38
rs771988647 Chr.6: 46248790-46248790 on Build GRCh38
rs540583582 Chr.6: 46248768-46248768 on Build GRCh38
rs376511659 Chr.6: 46248907-46248907 on Build GRCh38
rs780747078 Chr.6: 46248714-46248714 on Build GRCh38
rs750809841 Chr.6: 46248707-46248707 on Build GRCh38
rs1016014133 Chr.6: 46248728-46248728 on Build GRCh38
rs1009459234 Chr.6: 46248880-46248880 on Build GRCh38
rs1030361933 Chr.6: 46248799-46248799 on Build GRCh38
rs771291090 Chr.6: 46248673-46248673 on Build GRCh38
rs766934770 Chr.6: 46248879-46248879 on Build GRCh38
rs747640016 Chr.6: 46248718-46248718 on Build GRCh38
rs1006073180 Chr.6: 46249076-46249076 on Build GRCh38
rs1047814359 Chr.6: 46249047-46249047 on Build GRCh38
rs1054893642 Chr.6: 46248631-46248631 on Build GRCh38
rs967927340 Chr.6: 46248901-46248901 on Build GRCh38
rs1057344175 Chr.6: 46248989-46248989 on Build GRCh38
rs760788709 Chr.6: 46248683-46248683 on Build GRCh38
rs886348289 Chr.6: 46248697-46248697 on Build GRCh38
rs372526925 Chr.6: 46248899-46248899 on Build GRCh38
rs775422870 Chr.6: 46248798-46248798 on Build GRCh38
rs770580714 Chr.6: 46248756-46248756 on Build GRCh38
rs996959996 Chr.6: 46248703-46248703 on Build GRCh38
rs777363221 Chr.6: 46248889-46248889 on Build GRCh38
rs917657273 Chr.6: 46248992-46248992 on Build GRCh38
rs778491920 Chr.6: 46248913-46248913 on Build GRCh38
rs991949523 Chr.6: 46249031-46249032 on Build GRCh38
rs974772001 Chr.6: 46248762-46248762 on Build GRCh38
rs754118907 Chr.6: 46248685-46248685 on Build GRCh38
rs755643862 Chr.6: 46248886-46248886 on Build GRCh38
rs937478068 Chr.6: 46248956-46248956 on Build GRCh38
rs748999642 Chr.6: 46248737-46248737 on Build GRCh38
rs767460997 Chr.6: 46249019-46249019 on Build GRCh38
rs750795216 Chr.6: 46248858-46248858 on Build GRCh38
rs544295398 Chr.6: 46248946-46248946 on Build GRCh38
rs746974035 Chr.6: 46248942-46248942 on Build GRCh38
rs913238064 Chr.6: 46248866-46248866 on Build GRCh38
rs775547367 Chr.6: 46248677-46248677 on Build GRCh38

View Full Product Details