Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49431255-49431765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
UniGene
Hs.485527
Gene Chromosome Location
Chr.6: 49430360-49463328 on Build GRCh38
Transcripts
NM_000255.3 XM_005249143.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3353875 c.2244A>C p.Q748H Chr.6: 49431737-49431737 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948025531 Chr.6: 49431359-49431359 on Build GRCh38
rs548852414 Chr.6: 49431655-49431655 on Build GRCh38
rs764166500 Chr.6: 49431512-49431512 on Build GRCh38
rs560764129 Chr.6: 49431659-49431659 on Build GRCh38
rs766839518 Chr.6: 49431725-49431725 on Build GRCh38
rs76470114 Chr.6: 49431535-49431535 on Build GRCh38
rs774736275 Chr.6: 49431731-49431731 on Build GRCh38
rs935012218 Chr.6: 49431442-49431442 on Build GRCh38
rs530424880 Chr.6: 49431638-49431638 on Build GRCh38
rs576032707 Chr.6: 49431587-49431587 on Build GRCh38
rs1141335 Chr.6: 49431299-49431299 on Build GRCh38
rs192552805 Chr.6: 49431449-49431449 on Build GRCh38
rs886061558 Chr.6: 49431625-49431625 on Build GRCh38
rs761107295 Chr.6: 49431448-49431448 on Build GRCh38
rs944869479 Chr.6: 49431477-49431477 on Build GRCh38
rs199521080 Chr.6: 49431677-49431677 on Build GRCh38
rs892702467 Chr.6: 49431511-49431511 on Build GRCh38
rs916475972 Chr.6: 49431350-49431350 on Build GRCh38
rs1141334 Chr.6: 49431354-49431354 on Build GRCh38
rs113025987 Chr.6: 49431627-49431627 on Build GRCh38
rs924927027 Chr.6: 49431440-49431440 on Build GRCh38
rs776011450 Chr.6: 49431318-49431318 on Build GRCh38
rs1025441665 Chr.6: 49431283-49431283 on Build GRCh38
rs10713340 Chr.6: 49431527-49431527 on Build GRCh38
rs144168425 Chr.6: 49431447-49431447 on Build GRCh38
rs187747098 Chr.6: 49431471-49431471 on Build GRCh38
rs576059123 Chr.6: 49431365-49431368 on Build GRCh38
rs14341 Chr.6: 49431698-49431698 on Build GRCh38
rs1044264539 Chr.6: 49431528-49431528 on Build GRCh38
rs886061556 Chr.6: 49431528-49431528 on Build GRCh38
rs763361965 Chr.6: 49431724-49431724 on Build GRCh38
rs762165830 Chr.6: 49431679-49431679 on Build GRCh38
rs951281693 Chr.6: 49431330-49431330 on Build GRCh38
rs1141333 Chr.6: 49431413-49431413 on Build GRCh38
rs909993475 Chr.6: 49431338-49431338 on Build GRCh38
rs960855643 Chr.6: 49431735-49431735 on Build GRCh38
rs760186185 Chr.6: 49431739-49431739 on Build GRCh38
rs113362290 Chr.6: 49431425-49431425 on Build GRCh38
rs764175806 Chr.6: 49431553-49431554 on Build GRCh38
rs868507940 Chr.6: 49431402-49431402 on Build GRCh38
rs904284870 Chr.6: 49431595-49431595 on Build GRCh38
rs946944037 Chr.6: 49431519-49431519 on Build GRCh38
rs199933641 Chr.6: 49431694-49431694 on Build GRCh38
rs959637047 Chr.6: 49431661-49431661 on Build GRCh38
rs886061557 Chr.6: 49431566-49431566 on Build GRCh38
rs1001669081 Chr.6: 49431597-49431597 on Build GRCh38
rs1803945 Chr.6: 49431620-49431620 on Build GRCh38
rs541575016 Chr.6: 49431422-49431422 on Build GRCh38
rs942763588 Chr.6: 49431376-49431376 on Build GRCh38
rs984360853 Chr.6: 49431332-49431332 on Build GRCh38
rs868855862 Chr.6: 49431685-49431685 on Build GRCh38
rs182726681 Chr.6: 49431450-49431450 on Build GRCh38
rs773680972 Chr.6: 49431717-49431717 on Build GRCh38
rs192605776 Chr.6: 49431602-49431602 on Build GRCh38
rs934232023 Chr.6: 49431459-49431459 on Build GRCh38

View Full Product Details