Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52189328-52189823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17A
Entrez Gene ID
3605
Gene Name
interleukin 17A
Gene Aliases
CTLA-8, CTLA8, IL-17, IL-17A, IL17
UniGene
Hs.41724
Gene Chromosome Location
Chr.6: 52186387-52190638 on Build GRCh38
Transcripts
NM_002190.2

SNP Detail

SNP ID Chromosome Location
rs1029057638 Chr.6: 52189689-52189689 on Build GRCh38
rs991121455 Chr.6: 52189599-52189599 on Build GRCh38
rs563055165 Chr.6: 52189772-52189772 on Build GRCh38
rs1050974284 Chr.6: 52189388-52189388 on Build GRCh38
rs985060575 Chr.6: 52189415-52189415 on Build GRCh38
rs1010137001 Chr.6: 52189605-52189605 on Build GRCh38
rs557766068 Chr.6: 52189509-52189509 on Build GRCh38
rs1043834159 Chr.6: 52189647-52189647 on Build GRCh38
rs937153637 Chr.6: 52189566-52189566 on Build GRCh38
rs902141429 Chr.6: 52189373-52189373 on Build GRCh38
rs535655543 Chr.6: 52189389-52189389 on Build GRCh38
rs753620625 Chr.6: 52189677-52189677 on Build GRCh38
rs960930534 Chr.6: 52189589-52189589 on Build GRCh38
rs1027700263 Chr.6: 52189476-52189476 on Build GRCh38
rs139300148 Chr.6: 52189484-52189484 on Build GRCh38
rs775540570 Chr.6: 52189352-52189352 on Build GRCh38
rs1054314888 Chr.6: 52189571-52189571 on Build GRCh38
rs542981146 Chr.6: 52189648-52189648 on Build GRCh38
rs890826327 Chr.6: 52189690-52189690 on Build GRCh38
rs898286120 Chr.6: 52189759-52189759 on Build GRCh38
rs995364356 Chr.6: 52189766-52189766 on Build GRCh38
rs1006421208 Chr.6: 52189757-52189757 on Build GRCh38
rs1007214031 Chr.6: 52189706-52189706 on Build GRCh38
rs549257684 Chr.6: 52189380-52189380 on Build GRCh38
rs953486792 Chr.6: 52189391-52189391 on Build GRCh38
rs1018513922 Chr.6: 52189459-52189459 on Build GRCh38
rs997607463 Chr.6: 52189360-52189360 on Build GRCh38
rs866828728 Chr.6: 52189542-52189542 on Build GRCh38
rs927134248 Chr.6: 52189543-52189543 on Build GRCh38
rs998011804 Chr.6: 52189659-52189659 on Build GRCh38
rs115651024 Chr.6: 52189463-52189463 on Build GRCh38
rs1045421266 Chr.6: 52189508-52189508 on Build GRCh38
rs371003208 Chr.6: 52189516-52189516 on Build GRCh38
rs949870024 Chr.6: 52189494-52189494 on Build GRCh38
rs7747909 Chr.6: 52189451-52189451 on Build GRCh38
rs893084428 Chr.6: 52189600-52189600 on Build GRCh38
rs901809741 Chr.6: 52189631-52189631 on Build GRCh38
rs909436774 Chr.6: 52189433-52189433 on Build GRCh38
rs567728004 Chr.6: 52189365-52189365 on Build GRCh38
rs953537834 Chr.6: 52189718-52189718 on Build GRCh38
rs1042150722 Chr.6: 52189614-52189614 on Build GRCh38
rs1041217448 Chr.6: 52189562-52189577 on Build GRCh38
rs927991260 Chr.6: 52189565-52189565 on Build GRCh38

View Full Product Details