Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 54121306-54121851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLIP
Entrez Gene ID
90523
Gene Name
muscular LMNA-interacting protein
Gene Aliases
C6orf142
UniGene
Hs.449276
Gene Chromosome Location
Chr.6: 54018916-54266325 on Build GRCh38
Transcripts
NM_001281746.1 NM_001281747.1 NM_138569.2 XM_005249476.4 XM_005249478.4 XM_005249480.4 XM_006715245.3 XM_017011465.1 XM_017011466.1 XM_017011467.1 XM_017011468.1 XM_017011469.1 XM_017011470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3628586 c.94C>T p.L32L Chr.6: 54121477-54121477 on Build GRCh38
6287297 c.171T>G p.V57V Chr.6: 54121554-54121554 on Build GRCh38
4398955 c.218G>A p.R73K Chr.6: 54121601-54121601 on Build GRCh38
4396686 c.164T>A p.F55Y Chr.6: 54121547-54121547 on Build GRCh38
3411202 c.106T>A p.F36I Chr.6: 54121489-54121489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956723952 Chr.6: 54121768-54121768 on Build GRCh38
rs1035477427 Chr.6: 54121699-54121699 on Build GRCh38
rs1003357404 Chr.6: 54121727-54121727 on Build GRCh38
rs756641503 Chr.6: 54121378-54121378 on Build GRCh38
rs946349198 Chr.6: 54121478-54121478 on Build GRCh38
rs548052923 Chr.6: 54121591-54121591 on Build GRCh38
rs749621534 Chr.6: 54121519-54121519 on Build GRCh38
rs1023022250 Chr.6: 54121372-54121372 on Build GRCh38
rs371899212 Chr.6: 54121649-54121649 on Build GRCh38
rs889581333 Chr.6: 54121496-54121496 on Build GRCh38
rs770251749 Chr.6: 54121506-54121506 on Build GRCh38
rs778697864 Chr.6: 54121371-54121373 on Build GRCh38
rs971631299 Chr.6: 54121659-54121659 on Build GRCh38
rs748656231 Chr.6: 54121500-54121500 on Build GRCh38
rs762746398 Chr.6: 54121585-54121585 on Build GRCh38
rs756579880 Chr.6: 54121620-54121620 on Build GRCh38
rs146730657 Chr.6: 54121558-54121558 on Build GRCh38
rs566668859 Chr.6: 54121640-54121640 on Build GRCh38
rs532143199 Chr.6: 54121370-54121370 on Build GRCh38
rs753813583 Chr.6: 54121443-54121443 on Build GRCh38
rs143453914 Chr.6: 54121488-54121488 on Build GRCh38
rs200836848 Chr.6: 54121594-54121594 on Build GRCh38
rs781316348 Chr.6: 54121492-54121492 on Build GRCh38
rs758576910 Chr.6: 54121469-54121469 on Build GRCh38
rs778431444 Chr.6: 54121442-54121442 on Build GRCh38
rs761699152 Chr.6: 54121446-54121446 on Build GRCh38
rs766218656 Chr.6: 54121590-54121590 on Build GRCh38
rs887243822 Chr.6: 54121582-54121582 on Build GRCh38
rs967933864 Chr.6: 54121825-54121825 on Build GRCh38
rs778161074 Chr.6: 54121507-54121507 on Build GRCh38
rs546664177 Chr.6: 54121626-54121626 on Build GRCh38
rs771792128 Chr.6: 54121542-54121542 on Build GRCh38
rs759722425 Chr.6: 54121636-54121636 on Build GRCh38
rs2145754 Chr.6: 54121705-54121705 on Build GRCh38
rs752753056 Chr.6: 54121617-54121617 on Build GRCh38
rs924248808 Chr.6: 54121785-54121785 on Build GRCh38
rs946982804 Chr.6: 54121445-54121445 on Build GRCh38
rs1012925336 Chr.6: 54121371-54121371 on Build GRCh38
rs144363072 Chr.6: 54121735-54121735 on Build GRCh38
rs1025589609 Chr.6: 54121643-54121643 on Build GRCh38
rs775074227 Chr.6: 54121434-54121434 on Build GRCh38
rs990751362 Chr.6: 54121733-54121733 on Build GRCh38
rs987940990 Chr.6: 54121770-54121770 on Build GRCh38
rs773486671 Chr.6: 54121398-54121398 on Build GRCh38
rs760276534 Chr.6: 54121436-54121436 on Build GRCh38
rs770944621 Chr.6: 54121579-54121579 on Build GRCh38
rs1022116267 Chr.6: 54121772-54121772 on Build GRCh38
rs1002749825 Chr.6: 54121679-54121679 on Build GRCh38
rs370171631 Chr.6: 54121593-54121593 on Build GRCh38
rs545824776 Chr.6: 54121403-54121403 on Build GRCh38
rs778552215 Chr.6: 54121414-54121414 on Build GRCh38
rs768253884 Chr.6: 54121568-54121568 on Build GRCh38
rs767611774 Chr.6: 54121606-54121606 on Build GRCh38
rs934149017 Chr.6: 54121790-54121790 on Build GRCh38
rs933715505 Chr.6: 54121503-54121503 on Build GRCh38
rs928106337 Chr.6: 54121389-54121389 on Build GRCh38
rs978297517 Chr.6: 54121781-54121781 on Build GRCh38
rs962679513 Chr.6: 54121357-54121357 on Build GRCh38
rs968043937 Chr.6: 54121766-54121766 on Build GRCh38
rs570030626 Chr.6: 54121343-54121343 on Build GRCh38
rs776055476 Chr.6: 54121575-54121575 on Build GRCh38
rs971881727 Chr.6: 54121376-54121376 on Build GRCh38
rs34491532 Chr.6: 54121458-54121458 on Build GRCh38
rs771883821 Chr.6: 54121404-54121404 on Build GRCh38
rs764075337 Chr.6: 54121482-54121482 on Build GRCh38
rs775268876 Chr.6: 54121555-54121555 on Build GRCh38
rs750204318 Chr.6: 54121459-54121459 on Build GRCh38
rs986988827 Chr.6: 54121802-54121802 on Build GRCh38
rs750162948 Chr.6: 54121581-54121581 on Build GRCh38
rs761764090 Chr.6: 54121576-54121576 on Build GRCh38

View Full Product Details