Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55773962-55774466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP5
Entrez Gene ID
653
Gene Name
bone morphogenetic protein 5
Gene Aliases
-
UniGene
Hs.296648
Gene Chromosome Location
Chr.6: 55753653-55875625 on Build GRCh38
Transcripts
NM_021073.2 XM_005249304.3 XM_011514816.2 XM_011514817.2 XM_017011198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
174738 c.977G>A p.R326H Chr.6: 55774099-55774099 on Build GRCh38
7192807 c.926A>G p.E309G Chr.6: 55774150-55774150 on Build GRCh38
192221 c.920C>T p.A307V Chr.6: 55774156-55774156 on Build GRCh38
2156389 c.994C>T p.H332Y Chr.6: 55774082-55774082 on Build GRCh38
6107360 c.884A>G p.Q295R Chr.6: 55774192-55774192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199557041 Chr.6: 55774250-55774250 on Build GRCh38
rs971850770 Chr.6: 55773995-55773995 on Build GRCh38
rs754440232 Chr.6: 55774277-55774277 on Build GRCh38
rs376630543 Chr.6: 55774156-55774156 on Build GRCh38
rs367662677 Chr.6: 55774071-55774071 on Build GRCh38
rs557166594 Chr.6: 55774359-55774359 on Build GRCh38
rs983219554 Chr.6: 55774361-55774361 on Build GRCh38
rs750041762 Chr.6: 55774183-55774183 on Build GRCh38
rs751006049 Chr.6: 55774199-55774199 on Build GRCh38
rs1004361735 Chr.6: 55774154-55774154 on Build GRCh38
rs747912353 Chr.6: 55774134-55774134 on Build GRCh38
rs780974519 Chr.6: 55774221-55774221 on Build GRCh38
rs578227645 Chr.6: 55774300-55774300 on Build GRCh38
rs189315038 Chr.6: 55774368-55774368 on Build GRCh38
rs372083649 Chr.6: 55774240-55774240 on Build GRCh38
rs1026776985 Chr.6: 55774015-55774015 on Build GRCh38
rs751279868 Chr.6: 55774274-55774274 on Build GRCh38
rs930892360 Chr.6: 55774426-55774426 on Build GRCh38
rs200747178 Chr.6: 55774002-55774002 on Build GRCh38
rs988823878 Chr.6: 55774347-55774347 on Build GRCh38
rs752572133 Chr.6: 55774286-55774286 on Build GRCh38
rs759974481 Chr.6: 55774056-55774056 on Build GRCh38
rs139564850 Chr.6: 55774020-55774020 on Build GRCh38
rs767872170 Chr.6: 55774062-55774062 on Build GRCh38
rs201300716 Chr.6: 55774152-55774152 on Build GRCh38
rs756863265 Chr.6: 55774017-55774018 on Build GRCh38
rs1003391561 Chr.6: 55774181-55774181 on Build GRCh38
rs376050074 Chr.6: 55774254-55774254 on Build GRCh38
rs771089709 Chr.6: 55774236-55774236 on Build GRCh38
rs765928484 Chr.6: 55774265-55774265 on Build GRCh38
rs1012924212 Chr.6: 55774158-55774158 on Build GRCh38
rs537238573 Chr.6: 55774323-55774323 on Build GRCh38
rs959940519 Chr.6: 55774247-55774247 on Build GRCh38
rs757915843 Chr.6: 55774096-55774096 on Build GRCh38
rs542682614 Chr.6: 55774405-55774405 on Build GRCh38
rs752036358 Chr.6: 55774037-55774037 on Build GRCh38
rs41271330 Chr.6: 55774230-55774230 on Build GRCh38
rs940179861 Chr.6: 55774371-55774371 on Build GRCh38
rs568443463 Chr.6: 55774370-55774370 on Build GRCh38
rs758148894 Chr.6: 55774184-55774184 on Build GRCh38
rs575324695 Chr.6: 55774155-55774155 on Build GRCh38
rs746918830 Chr.6: 55774100-55774100 on Build GRCh38
rs908189585 Chr.6: 55774026-55774026 on Build GRCh38
rs934761877 Chr.6: 55774337-55774337 on Build GRCh38
rs773469282 Chr.6: 55774029-55774029 on Build GRCh38
rs538635494 Chr.6: 55774241-55774241 on Build GRCh38
rs774672647 Chr.6: 55774238-55774238 on Build GRCh38
rs900780090 Chr.6: 55774410-55774410 on Build GRCh38
rs367881870 Chr.6: 55774163-55774163 on Build GRCh38
rs776122373 Chr.6: 55774174-55774174 on Build GRCh38
rs943055484 Chr.6: 55774044-55774044 on Build GRCh38
rs866784947 Chr.6: 55774114-55774114 on Build GRCh38
rs750168609 Chr.6: 55774078-55774078 on Build GRCh38
rs371086729 Chr.6: 55774001-55774001 on Build GRCh38
rs530615736 Chr.6: 55774069-55774069 on Build GRCh38
rs978592036 Chr.6: 55774320-55774320 on Build GRCh38
rs369939910 Chr.6: 55774281-55774281 on Build GRCh38
rs765877188 Chr.6: 55774189-55774189 on Build GRCh38
rs551347386 Chr.6: 55774006-55774006 on Build GRCh38
rs764507596 Chr.6: 55774251-55774251 on Build GRCh38
rs752637340 Chr.6: 55774233-55774233 on Build GRCh38
rs990042383 Chr.6: 55774031-55774031 on Build GRCh38
rs777169320 Chr.6: 55774253-55774253 on Build GRCh38
rs771765601 Chr.6: 55773998-55773998 on Build GRCh38
rs80225326 Chr.6: 55774182-55774182 on Build GRCh38
rs767889125 Chr.6: 55774168-55774168 on Build GRCh38
rs1039070988 Chr.6: 55774064-55774064 on Build GRCh38
rs756066025 Chr.6: 55774289-55774289 on Build GRCh38
rs763002455 Chr.6: 55774034-55774034 on Build GRCh38
rs911902090 Chr.6: 55774367-55774367 on Build GRCh38
rs866515989 Chr.6: 55774106-55774106 on Build GRCh38
rs536905359 Chr.6: 55774420-55774420 on Build GRCh38
rs944745406 Chr.6: 55774383-55774383 on Build GRCh38
rs144630424 Chr.6: 55774120-55774120 on Build GRCh38
rs748147968 Chr.6: 55774229-55774229 on Build GRCh38
rs550837639 Chr.6: 55774115-55774115 on Build GRCh38
rs952433364 Chr.6: 55774017-55774017 on Build GRCh38
rs778695144 Chr.6: 55774075-55774075 on Build GRCh38
rs762058439 Chr.6: 55774018-55774018 on Build GRCh38
rs766509217 Chr.6: 55774035-55774035 on Build GRCh38
rs754562107 Chr.6: 55774204-55774204 on Build GRCh38
rs773415942 Chr.6: 55774153-55774153 on Build GRCh38
rs898664713 Chr.6: 55774423-55774423 on Build GRCh38
rs371743666 Chr.6: 55774099-55774099 on Build GRCh38
rs777556185 Chr.6: 55774231-55774231 on Build GRCh38
rs995450354 Chr.6: 55774205-55774205 on Build GRCh38

View Full Product Details