Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63278402-63278948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGSN
Entrez Gene ID
51557
Gene Name
lengsin, lens protein with glutamine synthetase domain
Gene Aliases
GLULD1, LGS
UniGene
Hs.149585
Gene Chromosome Location
Chr.6: 63275951-63567921 on Build GRCh38
Transcripts
NM_001143940.1 NM_016571.2 XM_011535889.2 XM_011535892.2 XM_017010929.1 XM_017010930.1 XM_017010931.1

SNP Detail

SNP ID Chromosome Location
rs929047262 Chr.6: 63278846-63278846 on Build GRCh38
rs1049787378 Chr.6: 63278443-63278443 on Build GRCh38
rs74406329 Chr.6: 63278615-63278615 on Build GRCh38
rs759760504 Chr.6: 63278583-63278583 on Build GRCh38
rs941298418 Chr.6: 63278460-63278460 on Build GRCh38
rs751833613 Chr.6: 63278484-63278484 on Build GRCh38
rs906988787 Chr.6: 63278577-63278577 on Build GRCh38
rs922783816 Chr.6: 63278823-63278823 on Build GRCh38
rs987126360 Chr.6: 63278874-63278874 on Build GRCh38
rs973586487 Chr.6: 63278787-63278787 on Build GRCh38
rs367898201 Chr.6: 63278476-63278477 on Build GRCh38
rs138623581 Chr.6: 63278885-63278885 on Build GRCh38
rs978638105 Chr.6: 63278804-63278804 on Build GRCh38
rs966924754 Chr.6: 63278792-63278792 on Build GRCh38
rs896995102 Chr.6: 63278493-63278493 on Build GRCh38
rs71551541 Chr.6: 63278716-63278717 on Build GRCh38
rs755041705 Chr.6: 63278773-63278773 on Build GRCh38
rs1011119193 Chr.6: 63278748-63278748 on Build GRCh38
rs868118635 Chr.6: 63278721-63278721 on Build GRCh38
rs933994580 Chr.6: 63278440-63278440 on Build GRCh38
rs1035730231 Chr.6: 63278611-63278611 on Build GRCh38
rs536140874 Chr.6: 63278669-63278669 on Build GRCh38
rs943340281 Chr.6: 63278498-63278498 on Build GRCh38
rs979392206 Chr.6: 63278632-63278632 on Build GRCh38
rs946588668 Chr.6: 63278602-63278602 on Build GRCh38
rs796719871 Chr.6: 63278480-63278480 on Build GRCh38
rs911259891 Chr.6: 63278876-63278876 on Build GRCh38
rs1010708384 Chr.6: 63278553-63278553 on Build GRCh38
rs1034014931 Chr.6: 63278609-63278609 on Build GRCh38
rs1023539648 Chr.6: 63278772-63278772 on Build GRCh38
rs370433619 Chr.6: 63278603-63278603 on Build GRCh38
rs765387068 Chr.6: 63278795-63278795 on Build GRCh38
rs9443684 Chr.6: 63278532-63278532 on Build GRCh38
rs1039261839 Chr.6: 63278521-63278521 on Build GRCh38
rs1711932 Chr.6: 63278457-63278457 on Build GRCh38
rs79254644 Chr.6: 63278901-63278901 on Build GRCh38
rs374288689 Chr.6: 63278576-63278576 on Build GRCh38
rs557407103 Chr.6: 63278881-63278881 on Build GRCh38
rs955466386 Chr.6: 63278865-63278865 on Build GRCh38
rs985540803 Chr.6: 63278867-63278867 on Build GRCh38
rs1001220868 Chr.6: 63278605-63278605 on Build GRCh38
rs574945444 Chr.6: 63278593-63278593 on Build GRCh38
rs769306526 Chr.6: 63278615-63278615 on Build GRCh38
rs76865677 Chr.6: 63278614-63278614 on Build GRCh38

View Full Product Details