Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 71301658-71302168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OGFRL1
Entrez Gene ID
79627
Gene Name
opioid growth factor receptor like 1
Gene Aliases
dJ331H24.1
UniGene
Hs.648434
Gene Chromosome Location
Chr.6: 71288296-71309059 on Build GRCh38
Transcripts
NM_001324266.1 NM_024576.4 XM_011536117.2 XM_011536118.2 XM_017011303.1 XM_017011304.1 XM_017011305.1 XM_017011306.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6012369 c.1109C>T p.P370L Chr.6: 71301802-71301802 on Build GRCh38
1081325 c.978G>A p.Q326Q Chr.6: 71301671-71301671 on Build GRCh38
1081326 c.1106C>T p.A369V Chr.6: 71301799-71301799 on Build GRCh38
1081327 c.1142G>A p.S381N Chr.6: 71301835-71301835 on Build GRCh38
6471377 c.1251A>T p.K417N Chr.6: 71301944-71301944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557063314 Chr.6: 71301731-71301731 on Build GRCh38
rs34358027 Chr.6: 71301879-71301881 on Build GRCh38
rs867933703 Chr.6: 71301749-71301749 on Build GRCh38
rs538989294 Chr.6: 71301721-71301721 on Build GRCh38
rs761750634 Chr.6: 71302041-71302041 on Build GRCh38
rs762505052 Chr.6: 71302068-71302068 on Build GRCh38
rs769801546 Chr.6: 71301854-71301854 on Build GRCh38
rs199528974 Chr.6: 71302053-71302053 on Build GRCh38
rs111886368 Chr.6: 71301691-71301691 on Build GRCh38
rs758024187 Chr.6: 71301941-71301941 on Build GRCh38
rs148929332 Chr.6: 71301744-71301744 on Build GRCh38
rs773278608 Chr.6: 71301874-71301874 on Build GRCh38
rs763894913 Chr.6: 71301776-71301776 on Build GRCh38
rs765325049 Chr.6: 71301936-71301936 on Build GRCh38
rs769889329 Chr.6: 71301752-71301752 on Build GRCh38
rs746104722 Chr.6: 71302033-71302033 on Build GRCh38
rs745527543 Chr.6: 71301715-71301715 on Build GRCh38
rs747304575 Chr.6: 71301827-71301827 on Build GRCh38
rs992771500 Chr.6: 71301878-71301878 on Build GRCh38
rs761836813 Chr.6: 71301928-71301928 on Build GRCh38
rs113493807 Chr.6: 71301971-71301971 on Build GRCh38
rs554816887 Chr.6: 71302025-71302025 on Build GRCh38
rs72265796 Chr.6: 71301877-71301879 on Build GRCh38
rs754658300 Chr.6: 71302086-71302086 on Build GRCh38
rs772248967 Chr.6: 71302038-71302038 on Build GRCh38
rs536739278 Chr.6: 71301947-71301947 on Build GRCh38
rs201290981 Chr.6: 71301860-71301860 on Build GRCh38
rs775714137 Chr.6: 71301720-71301720 on Build GRCh38
rs540548321 Chr.6: 71301682-71301682 on Build GRCh38
rs754629854 Chr.6: 71301831-71301831 on Build GRCh38
rs777304416 Chr.6: 71302074-71302075 on Build GRCh38
rs1001911124 Chr.6: 71301857-71301857 on Build GRCh38
rs774377620 Chr.6: 71302027-71302027 on Build GRCh38
rs770259674 Chr.6: 71301968-71301968 on Build GRCh38
rs77957107 Chr.6: 71302036-71302036 on Build GRCh38
rs182141917 Chr.6: 71301775-71301775 on Build GRCh38
rs996529829 Chr.6: 71301687-71301687 on Build GRCh38
rs752409796 Chr.6: 71301773-71301773 on Build GRCh38
rs778568465 Chr.6: 71301807-71301807 on Build GRCh38
rs373960221 Chr.6: 71301942-71301942 on Build GRCh38
rs200057637 Chr.6: 71301906-71301906 on Build GRCh38
rs765983077 Chr.6: 71301700-71301700 on Build GRCh38
rs763961445 Chr.6: 71301900-71301900 on Build GRCh38
rs376961605 Chr.6: 71301809-71301809 on Build GRCh38
rs747781719 Chr.6: 71301939-71301940 on Build GRCh38
rs749324254 Chr.6: 71301981-71301981 on Build GRCh38
rs779807137 Chr.6: 71301813-71301813 on Build GRCh38
rs940260323 Chr.6: 71302087-71302087 on Build GRCh38
rs754718119 Chr.6: 71301915-71301915 on Build GRCh38
rs868757013 Chr.6: 71301951-71301951 on Build GRCh38
rs773316142 Chr.6: 71302056-71302056 on Build GRCh38
rs771026271 Chr.6: 71301890-71301890 on Build GRCh38
rs772497490 Chr.6: 71301718-71301718 on Build GRCh38
rs773367085 Chr.6: 71301756-71301756 on Build GRCh38
rs766642252 Chr.6: 71301772-71301772 on Build GRCh38
rs1041301177 Chr.6: 71302111-71302111 on Build GRCh38
rs371639663 Chr.6: 71301789-71301789 on Build GRCh38
rs757908221 Chr.6: 71302034-71302035 on Build GRCh38
rs143633069 Chr.6: 71301943-71301943 on Build GRCh38
rs765930578 Chr.6: 71302075-71302075 on Build GRCh38
rs892275404 Chr.6: 71301704-71301704 on Build GRCh38
rs146800581 Chr.6: 71301737-71301737 on Build GRCh38
rs753835864 Chr.6: 71301783-71301783 on Build GRCh38
rs986891884 Chr.6: 71302046-71302046 on Build GRCh38
rs769096614 Chr.6: 71301836-71301836 on Build GRCh38
rs369232745 Chr.6: 71302003-71302003 on Build GRCh38
rs1034656476 Chr.6: 71301866-71301866 on Build GRCh38
rs763170227 Chr.6: 71301758-71301758 on Build GRCh38
rs751108686 Chr.6: 71302082-71302082 on Build GRCh38
rs749931429 Chr.6: 71301939-71301939 on Build GRCh38
rs200574638 Chr.6: 71302007-71302007 on Build GRCh38
rs748671488 Chr.6: 71301847-71301847 on Build GRCh38
rs34010066 Chr.6: 71302022-71302022 on Build GRCh38
rs901321494 Chr.6: 71302129-71302129 on Build GRCh38
rs781506948 Chr.6: 71301846-71301846 on Build GRCh38
rs760336617 Chr.6: 71301893-71301893 on Build GRCh38
rs748583194 Chr.6: 71301952-71301952 on Build GRCh38
rs778660135 Chr.6: 71301713-71301713 on Build GRCh38
rs774660399 Chr.6: 71301892-71301892 on Build GRCh38
rs755989224 Chr.6: 71301689-71301689 on Build GRCh38
rs777810665 Chr.6: 71301690-71301690 on Build GRCh38
rs758111965 Chr.6: 71301810-71301810 on Build GRCh38

View Full Product Details