Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83591096-83591600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP91
Entrez Gene ID
9892
Gene Name
synaptosome associated protein 91
Gene Aliases
AP180, CALM
UniGene
Hs.368046
Gene Chromosome Location
Chr.6: 83552885-83709408 on Build GRCh38
Transcripts
NM_001242792.1 NM_001242793.1 NM_001242794.1 NM_001256717.1 NM_001256718.1 NM_014841.2 XM_005248770.4 XM_006715615.1 XM_011536265.1 XM_011536266.1 XM_011536269.1 XM_011536271.1 XM_011536273.2 XM_011536275.2 XM_011536276.2 XM_011536277.1 XM_017011554.1 XM_017011555.1 XM_017011556.1 XM_017011557.1 XM_017011558.1 XM_017011559.1 XM_017011560.1 XM_017011561.1 XM_017011562.1 XM_017011563.1 XM_017011564.1 XM_017011565.1 XM_017011566.1 XM_017011567.1 XM_017011568.1 XM_017011569.1 XM_017011570.1 XM_017011571.1 XM_017011572.1 XM_017011573.1 XM_017011574.1 XM_017011575.1 XM_017011576.1 XM_017011577.1 XM_017011578.1 XM_017011579.1 XM_017011580.1 XM_017011581.1 XM_017011582.1 XM_017011583.1 XM_017011584.1 XM_017011585.1 XM_017011586.1 XM_017011587.1 XM_017011588.1 XM_017011589.1 XM_017011590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
228448 c.2012C>T p.A671V Chr.6: 83591213-83591213 on Build GRCh38
3949495 c.1985G>A p.S662N Chr.6: 83591240-83591240 on Build GRCh38
3949497 c.1979C>A p.A660D Chr.6: 83591246-83591246 on Build GRCh38
7317230 c.1991C>T p.S664L Chr.6: 83591234-83591234 on Build GRCh38
5600535 c.1957C>T p.P653S Chr.6: 83591268-83591268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780662923 Chr.6: 83591445-83591445 on Build GRCh38
rs80324933 Chr.6: 83591329-83591329 on Build GRCh38
rs775115842 Chr.6: 83591299-83591299 on Build GRCh38
rs375839747 Chr.6: 83591292-83591292 on Build GRCh38
rs912684309 Chr.6: 83591309-83591309 on Build GRCh38
rs764046983 Chr.6: 83591186-83591186 on Build GRCh38
rs755272450 Chr.6: 83591252-83591252 on Build GRCh38
rs890245614 Chr.6: 83591333-83591333 on Build GRCh38
rs931800338 Chr.6: 83591542-83591542 on Build GRCh38
rs572298497 Chr.6: 83591186-83591187 on Build GRCh38
rs899860086 Chr.6: 83591328-83591328 on Build GRCh38
rs1011191203 Chr.6: 83591480-83591480 on Build GRCh38
rs774424276 Chr.6: 83591170-83591170 on Build GRCh38
rs776273835 Chr.6: 83591308-83591308 on Build GRCh38
rs532942362 Chr.6: 83591444-83591444 on Build GRCh38
rs371512281 Chr.6: 83591509-83591509 on Build GRCh38
rs762162588 Chr.6: 83591230-83591230 on Build GRCh38
rs752034185 Chr.6: 83591411-83591411 on Build GRCh38
rs766514515 Chr.6: 83591246-83591246 on Build GRCh38
rs990385127 Chr.6: 83591281-83591281 on Build GRCh38
rs1013112192 Chr.6: 83591433-83591433 on Build GRCh38
rs771047718 Chr.6: 83591163-83591163 on Build GRCh38
rs777039637 Chr.6: 83591228-83591228 on Build GRCh38
rs1049046479 Chr.6: 83591357-83591357 on Build GRCh38
rs1033990495 Chr.6: 83591271-83591271 on Build GRCh38
rs746490602 Chr.6: 83591300-83591300 on Build GRCh38
rs778793061 Chr.6: 83591277-83591277 on Build GRCh38
rs765599711 Chr.6: 83591266-83591266 on Build GRCh38
rs780298228 Chr.6: 83591317-83591317 on Build GRCh38
rs956594106 Chr.6: 83591528-83591529 on Build GRCh38
rs975516117 Chr.6: 83591531-83591531 on Build GRCh38
rs765522350 Chr.6: 83591236-83591236 on Build GRCh38
rs766769545 Chr.6: 83591344-83591344 on Build GRCh38
rs549361005 Chr.6: 83591460-83591460 on Build GRCh38
rs199865724 Chr.6: 83591250-83591250 on Build GRCh38
rs550808759 Chr.6: 83591502-83591502 on Build GRCh38
rs755544011 Chr.6: 83591424-83591424 on Build GRCh38
rs147096228 Chr.6: 83591404-83591404 on Build GRCh38
rs764266611 Chr.6: 83591180-83591180 on Build GRCh38
rs907002233 Chr.6: 83591420-83591420 on Build GRCh38
rs1015177450 Chr.6: 83591128-83591128 on Build GRCh38
rs188735307 Chr.6: 83591301-83591301 on Build GRCh38
rs370831877 Chr.6: 83591204-83591204 on Build GRCh38
rs1004717034 Chr.6: 83591183-83591183 on Build GRCh38
rs1024801812 Chr.6: 83591472-83591473 on Build GRCh38
rs915191442 Chr.6: 83591534-83591534 on Build GRCh38
rs2273364 Chr.6: 83591431-83591431 on Build GRCh38
rs887685930 Chr.6: 83591399-83591399 on Build GRCh38
rs759322601 Chr.6: 83591192-83591192 on Build GRCh38
rs994967435 Chr.6: 83591149-83591149 on Build GRCh38
rs912742339 Chr.6: 83591510-83591510 on Build GRCh38
rs757160925 Chr.6: 83591335-83591336 on Build GRCh38
rs1021214218 Chr.6: 83591489-83591489 on Build GRCh38
rs750956338 Chr.6: 83591241-83591241 on Build GRCh38
rs760416976 Chr.6: 83591218-83591218 on Build GRCh38
rs540264016 Chr.6: 83591287-83591287 on Build GRCh38
rs200962227 Chr.6: 83591193-83591193 on Build GRCh38
rs372039519 Chr.6: 83591312-83591312 on Build GRCh38
rs774813804 Chr.6: 83591199-83591199 on Build GRCh38
rs2273363 Chr.6: 83591278-83591278 on Build GRCh38
rs989786044 Chr.6: 83591529-83591529 on Build GRCh38
rs556501720 Chr.6: 83591255-83591255 on Build GRCh38
rs965914793 Chr.6: 83591515-83591515 on Build GRCh38
rs749161642 Chr.6: 83591257-83591257 on Build GRCh38
rs200471899 Chr.6: 83591574-83591574 on Build GRCh38
rs1031640691 Chr.6: 83591523-83591523 on Build GRCh38
rs894615478 Chr.6: 83591432-83591432 on Build GRCh38
rs45610234 Chr.6: 83591227-83591227 on Build GRCh38
rs375343309 Chr.6: 83591318-83591318 on Build GRCh38
rs1027558356 Chr.6: 83591131-83591131 on Build GRCh38

View Full Product Details