Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88047697-88048197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPACA1
Entrez Gene ID
81833
Gene Name
sperm acrosome associated 1
Gene Aliases
SAMP32
UniGene
Hs.161241
Gene Chromosome Location
Chr.6: 88046907-88066832 on Build GRCh38
Transcripts
NM_030960.2 XM_011536160.2 XM_017011335.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4758574 c.28G>A p.A10T Chr.6: 88047933-88047933 on Build GRCh38
4758575 c.142G>A p.E48K Chr.6: 88048047-88048047 on Build GRCh38
4758576 c.185A>G p.Y62C Chr.6: 88048090-88048090 on Build GRCh38
5737687 c.83G>A p.R28H Chr.6: 88047988-88047988 on Build GRCh38
4650723 c.135C>T p.G45G Chr.6: 88048040-88048040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757434993 Chr.6: 88047985-88047985 on Build GRCh38
rs1041627570 Chr.6: 88048054-88048054 on Build GRCh38
rs370125470 Chr.6: 88048029-88048029 on Build GRCh38
rs61738528 Chr.6: 88047986-88047986 on Build GRCh38
rs764362411 Chr.6: 88048069-88048069 on Build GRCh38
rs774874210 Chr.6: 88047940-88047940 on Build GRCh38
rs1010464714 Chr.6: 88047844-88047844 on Build GRCh38
rs754914513 Chr.6: 88048097-88048097 on Build GRCh38
rs113084849 Chr.6: 88047741-88047741 on Build GRCh38
rs747296120 Chr.6: 88048160-88048160 on Build GRCh38
rs747814975 Chr.6: 88047890-88047890 on Build GRCh38
rs748761136 Chr.6: 88048021-88048021 on Build GRCh38
rs762827583 Chr.6: 88048008-88048014 on Build GRCh38
rs936433024 Chr.6: 88048048-88048048 on Build GRCh38
rs920981790 Chr.6: 88048020-88048020 on Build GRCh38
rs764233782 Chr.6: 88047982-88047982 on Build GRCh38
rs772750137 Chr.6: 88047894-88047894 on Build GRCh38
rs1015503176 Chr.6: 88047840-88047840 on Build GRCh38
rs912384960 Chr.6: 88047970-88047970 on Build GRCh38
rs996916620 Chr.6: 88047806-88047806 on Build GRCh38
rs111801093 Chr.6: 88047744-88047744 on Build GRCh38
rs747981584 Chr.6: 88048170-88048170 on Build GRCh38
rs752948005 Chr.6: 88047954-88047954 on Build GRCh38
rs763429718 Chr.6: 88047898-88047898 on Build GRCh38
rs777344108 Chr.6: 88048124-88048124 on Build GRCh38
rs769382887 Chr.6: 88047999-88047999 on Build GRCh38
rs200613021 Chr.6: 88047984-88047984 on Build GRCh38
rs974068233 Chr.6: 88047935-88047935 on Build GRCh38
rs780828165 Chr.6: 88047881-88047881 on Build GRCh38
rs760925142 Chr.6: 88048049-88048049 on Build GRCh38
rs202046887 Chr.6: 88047931-88047931 on Build GRCh38
rs746352729 Chr.6: 88048150-88048150 on Build GRCh38
rs755937359 Chr.6: 88048123-88048123 on Build GRCh38
rs776529556 Chr.6: 88048156-88048156 on Build GRCh38
rs912417617 Chr.6: 88047771-88047771 on Build GRCh38
rs539240585 Chr.6: 88047943-88047945 on Build GRCh38
rs61729414 Chr.6: 88047945-88047945 on Build GRCh38
rs746971408 Chr.6: 88047892-88047892 on Build GRCh38
rs372679476 Chr.6: 88048149-88048149 on Build GRCh38
rs772501010 Chr.6: 88048151-88048151 on Build GRCh38
rs899924024 Chr.6: 88047800-88047800 on Build GRCh38
rs765418925 Chr.6: 88048091-88048091 on Build GRCh38
rs767790083 Chr.6: 88047946-88047946 on Build GRCh38
rs1008441069 Chr.6: 88047780-88047780 on Build GRCh38
rs145474393 Chr.6: 88048080-88048080 on Build GRCh38
rs538428968 Chr.6: 88048087-88048087 on Build GRCh38
rs1020394351 Chr.6: 88047844-88047844 on Build GRCh38
rs774784623 Chr.6: 88048025-88048025 on Build GRCh38
rs78486869 Chr.6: 88047832-88047832 on Build GRCh38
rs943750011 Chr.6: 88047793-88047793 on Build GRCh38
rs772477775 Chr.6: 88048035-88048035 on Build GRCh38
rs368664648 Chr.6: 88047811-88047811 on Build GRCh38
rs1034478413 Chr.6: 88047897-88047897 on Build GRCh38
rs914982716 Chr.6: 88047913-88047913 on Build GRCh38
rs767321835 Chr.6: 88048107-88048107 on Build GRCh38
rs376457887 Chr.6: 88047992-88047992 on Build GRCh38
rs946290868 Chr.6: 88048171-88048171 on Build GRCh38
rs780033715 Chr.6: 88047856-88047856 on Build GRCh38
rs754144472 Chr.6: 88048073-88048073 on Build GRCh38
rs929885371 Chr.6: 88047748-88047748 on Build GRCh38
rs570117863 Chr.6: 88047922-88047922 on Build GRCh38
rs377337225 Chr.6: 88048095-88048095 on Build GRCh38
rs148871108 Chr.6: 88048022-88048022 on Build GRCh38
rs751546525 Chr.6: 88047988-88047988 on Build GRCh38
rs760870755 Chr.6: 88047976-88047976 on Build GRCh38
rs748955369 Chr.6: 88048138-88048138 on Build GRCh38
rs777430082 Chr.6: 88048003-88048003 on Build GRCh38
rs971568614 Chr.6: 88047885-88047885 on Build GRCh38
rs775754516 Chr.6: 88048046-88048046 on Build GRCh38
rs768589484 Chr.6: 88048118-88048118 on Build GRCh38
rs952742957 Chr.6: 88047828-88047828 on Build GRCh38
rs1041179233 Chr.6: 88047785-88047785 on Build GRCh38
rs760015701 Chr.6: 88048031-88048031 on Build GRCh38
rs747805683 Chr.6: 88047998-88047998 on Build GRCh38
rs535615759 Chr.6: 88048145-88048145 on Build GRCh38
rs754790909 Chr.6: 88047990-88047990 on Build GRCh38
rs1039483658 Chr.6: 88047797-88047797 on Build GRCh38
rs548709812 Chr.6: 88047863-88047863 on Build GRCh38
rs1046901763 Chr.6: 88047763-88047763 on Build GRCh38
rs570807749 Chr.6: 88047953-88047953 on Build GRCh38

View Full Product Details