Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96204153-96204698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT9
Entrez Gene ID
10690
Gene Name
fucosyltransferase 9
Gene Aliases
Fuc-TIX
UniGene
Hs.49117
Gene Chromosome Location
Chr.6: 96015969-96215612 on Build GRCh38
Transcripts
NM_006581.3 XM_011535383.2 XM_011535385.2 XM_017010188.1 XM_017010190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082697 c.1046C>T p.S349F Chr.6: 96204201-96204201 on Build GRCh38
128039 c.1014C>T p.C338C Chr.6: 96204169-96204169 on Build GRCh38
4519540 c.1015G>A p.D339N Chr.6: 96204170-96204170 on Build GRCh38
3630664 c.1028G>A p.R343K Chr.6: 96204183-96204183 on Build GRCh38
5606568 c.1062G>A p.E354E Chr.6: 96204217-96204217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192681787 Chr.6: 96204479-96204479 on Build GRCh38
rs772492669 Chr.6: 96204233-96204233 on Build GRCh38
rs949116706 Chr.6: 96204594-96204594 on Build GRCh38
rs868515812 Chr.6: 96204201-96204201 on Build GRCh38
rs374869820 Chr.6: 96204217-96204217 on Build GRCh38
rs138904451 Chr.6: 96204242-96204242 on Build GRCh38
rs902927451 Chr.6: 96204451-96204451 on Build GRCh38
rs899351392 Chr.6: 96204395-96204395 on Build GRCh38
rs978375081 Chr.6: 96204280-96204280 on Build GRCh38
rs554399990 Chr.6: 96204183-96204183 on Build GRCh38
rs952644345 Chr.6: 96204593-96204593 on Build GRCh38
rs377446515 Chr.6: 96204668-96204668 on Build GRCh38
rs559267775 Chr.6: 96204362-96204362 on Build GRCh38
rs759805468 Chr.6: 96204279-96204279 on Build GRCh38
rs774487481 Chr.6: 96204277-96204277 on Build GRCh38
rs548165417 Chr.6: 96204663-96204663 on Build GRCh38
rs745490987 Chr.6: 96204504-96204504 on Build GRCh38
rs375490773 Chr.6: 96204226-96204226 on Build GRCh38
rs949105596 Chr.6: 96204306-96204306 on Build GRCh38
rs1021309040 Chr.6: 96204182-96204182 on Build GRCh38
rs532146075 Chr.6: 96204514-96204514 on Build GRCh38
rs925213779 Chr.6: 96204502-96204502 on Build GRCh38
rs746363786 Chr.6: 96204232-96204232 on Build GRCh38
rs146087318 Chr.6: 96204178-96204178 on Build GRCh38
rs1008934389 Chr.6: 96204455-96204455 on Build GRCh38
rs954828366 Chr.6: 96204426-96204426 on Build GRCh38
rs765403829 Chr.6: 96204293-96204293 on Build GRCh38
rs1029105339 Chr.6: 96204417-96204417 on Build GRCh38
rs576668059 Chr.6: 96204316-96204316 on Build GRCh38
rs111391528 Chr.6: 96204444-96204444 on Build GRCh38
rs139604379 Chr.6: 96204359-96204359 on Build GRCh38
rs1042763019 Chr.6: 96204449-96204449 on Build GRCh38
rs543256719 Chr.6: 96204483-96204483 on Build GRCh38
rs41285452 Chr.6: 96204392-96204392 on Build GRCh38
rs756657667 Chr.6: 96204207-96204207 on Build GRCh38
rs969359206 Chr.6: 96204299-96204299 on Build GRCh38
rs149412113 Chr.6: 96204264-96204264 on Build GRCh38
rs747282714 Chr.6: 96204243-96204243 on Build GRCh38
rs769704584 Chr.6: 96204276-96204276 on Build GRCh38
rs548225071 Chr.6: 96204248-96204248 on Build GRCh38
rs758696937 Chr.6: 96204347-96204347 on Build GRCh38
rs552067796 Chr.6: 96204567-96204567 on Build GRCh38
rs147140326 Chr.6: 96204604-96204604 on Build GRCh38
rs909177658 Chr.6: 96204614-96204614 on Build GRCh38
rs867693522 Chr.6: 96204430-96204430 on Build GRCh38
rs1012780213 Chr.6: 96204532-96204532 on Build GRCh38
rs753066572 Chr.6: 96204313-96204313 on Build GRCh38
rs147507075 Chr.6: 96204302-96204302 on Build GRCh38
rs982233049 Chr.6: 96204599-96204599 on Build GRCh38
rs528099271 Chr.6: 96204638-96204638 on Build GRCh38
rs753422213 Chr.6: 96204198-96204198 on Build GRCh38
rs750318433 Chr.6: 96204273-96204273 on Build GRCh38
rs777195087 Chr.6: 96204190-96204190 on Build GRCh38
rs145125486 Chr.6: 96204363-96204363 on Build GRCh38
rs867019790 Chr.6: 96204228-96204229 on Build GRCh38
rs4078339 Chr.6: 96204495-96204495 on Build GRCh38
rs777003484 Chr.6: 96204466-96204466 on Build GRCh38
rs1036859174 Chr.6: 96204343-96204343 on Build GRCh38
rs984820674 Chr.6: 96204300-96204300 on Build GRCh38
rs755741141 Chr.6: 96204640-96204640 on Build GRCh38
rs768946973 Chr.6: 96204245-96204245 on Build GRCh38
rs907550244 Chr.6: 96204613-96204613 on Build GRCh38
rs761648409 Chr.6: 96204674-96204674 on Build GRCh38
rs561257134 Chr.6: 96204416-96204416 on Build GRCh38
rs1022875227 Chr.6: 96204589-96204589 on Build GRCh38
rs9986564 Chr.6: 96204227-96204227 on Build GRCh38
rs755759092 Chr.6: 96204188-96204188 on Build GRCh38
rs756702235 Chr.6: 96204382-96204382 on Build GRCh38

View Full Product Details