Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116616801-116617310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPH4A
Entrez Gene ID
345895
Gene Name
radial spoke head 4 homolog A
Gene Aliases
CILD11, RSHL3, RSPH6B, dJ412I7.1
UniGene
Hs.160380
Gene Chromosome Location
Chr.6: 116598184-116633051 on Build GRCh38
Transcripts
NM_001010892.2 NM_001161664.1 XM_017010826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318119 c.226G>T p.G76* Chr.6: 116616849-116616849 on Build GRCh38
7243354 c.314C>T p.A105V Chr.6: 116616937-116616937 on Build GRCh38
4825473 c.278C>G p.S93C Chr.6: 116616901-116616901 on Build GRCh38
3619692 c.373C>T p.P125S Chr.6: 116616996-116616996 on Build GRCh38
1439992 c.523G>T p.E175* Chr.6: 116617146-116617146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577977924 Chr.6: 116616965-116616965 on Build GRCh38
rs764663856 Chr.6: 116617145-116617146 on Build GRCh38
rs372902497 Chr.6: 116617067-116617067 on Build GRCh38
rs118204042 Chr.6: 116616948-116616948 on Build GRCh38
rs757678261 Chr.6: 116617168-116617168 on Build GRCh38
rs143264253 Chr.6: 116617219-116617219 on Build GRCh38
rs531594790 Chr.6: 116617242-116617242 on Build GRCh38
rs761742508 Chr.6: 116617101-116617101 on Build GRCh38
rs376372993 Chr.6: 116617186-116617186 on Build GRCh38
rs773878067 Chr.6: 116616870-116616870 on Build GRCh38
rs775559853 Chr.6: 116616982-116616982 on Build GRCh38
rs202006186 Chr.6: 116616883-116616883 on Build GRCh38
rs1008393343 Chr.6: 116616876-116616876 on Build GRCh38
rs545486771 Chr.6: 116617036-116617036 on Build GRCh38
rs372083168 Chr.6: 116617261-116617261 on Build GRCh38
rs766527231 Chr.6: 116616893-116616893 on Build GRCh38
rs1030816346 Chr.6: 116616936-116616936 on Build GRCh38
rs756868889 Chr.6: 116617053-116617053 on Build GRCh38
rs139745647 Chr.6: 116616930-116616930 on Build GRCh38
rs762142077 Chr.6: 116617048-116617049 on Build GRCh38
rs748926290 Chr.6: 116616940-116616940 on Build GRCh38
rs769325632 Chr.6: 116616843-116616843 on Build GRCh38
rs763466678 Chr.6: 116616967-116616968 on Build GRCh38
rs758078631 Chr.6: 116617183-116617183 on Build GRCh38
rs369824862 Chr.6: 116617010-116617010 on Build GRCh38
rs562856574 Chr.6: 116616964-116616964 on Build GRCh38
rs775574466 Chr.6: 116616859-116616861 on Build GRCh38
rs141226759 Chr.6: 116617207-116617207 on Build GRCh38
rs760588703 Chr.6: 116616885-116616885 on Build GRCh38
rs756610987 Chr.6: 116617048-116617048 on Build GRCh38
rs201187909 Chr.6: 116616910-116616910 on Build GRCh38
rs118204041 Chr.6: 116617083-116617083 on Build GRCh38
rs371869815 Chr.6: 116616865-116616865 on Build GRCh38
rs775607244 Chr.6: 116617233-116617233 on Build GRCh38
rs532331878 Chr.6: 116617115-116617115 on Build GRCh38
rs764547571 Chr.6: 116617154-116617154 on Build GRCh38
rs774765832 Chr.6: 116617042-116617043 on Build GRCh38
rs745371405 Chr.6: 116617064-116617064 on Build GRCh38
rs77016758 Chr.6: 116617119-116617119 on Build GRCh38
rs553472573 Chr.6: 116616847-116616847 on Build GRCh38
rs778605738 Chr.6: 116617060-116617060 on Build GRCh38
rs766758184 Chr.6: 116617019-116617019 on Build GRCh38
rs754588754 Chr.6: 116617119-116617120 on Build GRCh38
rs762313827 Chr.6: 116617273-116617273 on Build GRCh38
rs145831200 Chr.6: 116616860-116616860 on Build GRCh38
rs764651259 Chr.6: 116617248-116617248 on Build GRCh38
rs778146751 Chr.6: 116617051-116617051 on Build GRCh38
rs767717727 Chr.6: 116617040-116617040 on Build GRCh38
rs769799020 Chr.6: 116616826-116616827 on Build GRCh38
rs767490154 Chr.6: 116616882-116616882 on Build GRCh38
rs866390964 Chr.6: 116617046-116617046 on Build GRCh38
rs772232278 Chr.6: 116617229-116617229 on Build GRCh38
rs140811116 Chr.6: 116617274-116617274 on Build GRCh38
rs13213314 Chr.6: 116617069-116617069 on Build GRCh38
rs751712749 Chr.6: 116616895-116616895 on Build GRCh38
rs530756069 Chr.6: 116616891-116616891 on Build GRCh38
rs895120131 Chr.6: 116616867-116616867 on Build GRCh38
rs764572415 Chr.6: 116616995-116616996 on Build GRCh38
rs963891526 Chr.6: 116617137-116617137 on Build GRCh38
rs555565982 Chr.6: 116616888-116616888 on Build GRCh38
rs959870218 Chr.6: 116617059-116617059 on Build GRCh38
rs775450268 Chr.6: 116617121-116617121 on Build GRCh38
rs151279930 Chr.6: 116617262-116617262 on Build GRCh38
rs976414886 Chr.6: 116617020-116617020 on Build GRCh38
rs755522050 Chr.6: 116616902-116616902 on Build GRCh38
rs758027694 Chr.6: 116617286-116617286 on Build GRCh38
rs759417066 Chr.6: 116616878-116616878 on Build GRCh38
rs1018616962 Chr.6: 116616993-116616993 on Build GRCh38
rs200149193 Chr.6: 116616823-116616823 on Build GRCh38
rs750812905 Chr.6: 116617279-116617279 on Build GRCh38
rs753180816 Chr.6: 116617041-116617041 on Build GRCh38
rs778370377 Chr.6: 116616963-116616963 on Build GRCh38
rs769643510 Chr.6: 116617210-116617210 on Build GRCh38
rs549196993 Chr.6: 116617188-116617188 on Build GRCh38
rs753464266 Chr.6: 116617118-116617119 on Build GRCh38
rs748597085 Chr.6: 116616826-116616826 on Build GRCh38
rs746056069 Chr.6: 116616824-116616825 on Build GRCh38
rs768436105 Chr.6: 116617016-116617016 on Build GRCh38
rs767821337 Chr.6: 116617091-116617092 on Build GRCh38
rs754223456 Chr.6: 116617163-116617163 on Build GRCh38
rs750012036 Chr.6: 116617173-116617173 on Build GRCh38
rs992709696 Chr.6: 116617068-116617068 on Build GRCh38
rs751661438 Chr.6: 116617035-116617035 on Build GRCh38
rs767593671 Chr.6: 116617118-116617118 on Build GRCh38
rs754915761 Chr.6: 116617197-116617197 on Build GRCh38
rs756442852 Chr.6: 116616924-116616924 on Build GRCh38
rs761222226 Chr.6: 116617134-116617134 on Build GRCh38
rs374708076 Chr.6: 116617017-116617017 on Build GRCh38
rs774655338 Chr.6: 116617117-116617117 on Build GRCh38
rs781780861 Chr.6: 116616909-116616909 on Build GRCh38
rs776466480 Chr.6: 116617093-116617093 on Build GRCh38
rs765600281 Chr.6: 116617169-116617169 on Build GRCh38
rs368842442 Chr.6: 116617052-116617052 on Build GRCh38
rs777066141 Chr.6: 116617258-116617258 on Build GRCh38
rs763188022 Chr.6: 116617018-116617018 on Build GRCh38
rs773168644 Chr.6: 116617216-116617216 on Build GRCh38
rs750605299 Chr.6: 116616889-116616889 on Build GRCh38

View Full Product Details