Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118964588-118965093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184A
Entrez Gene ID
79632
Gene Name
family with sequence similarity 184 member A
Gene Aliases
C6orf60
UniGene
Hs.443789
Gene Chromosome Location
Chr.6: 118959829-119149193 on Build GRCh38
Transcripts
NM_001100411.2 NM_001288576.1 NM_024581.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754462 c.3072A>G p.R1024R Chr.6: 118964733-118964733 on Build GRCh38
3857895 c.3044A>C p.K1015T Chr.6: 118964761-118964761 on Build GRCh38
6300796 c.3082T>C p.F1028L Chr.6: 118964723-118964723 on Build GRCh38
6256786 c.3110C>A p.T1037N Chr.6: 118964695-118964695 on Build GRCh38
3857894 c.3122T>C p.I1041T Chr.6: 118964683-118964683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530694270 Chr.6: 118964847-118964847 on Build GRCh38
rs950845073 Chr.6: 118964938-118964938 on Build GRCh38
rs377693723 Chr.6: 118964712-118964712 on Build GRCh38
rs916626288 Chr.6: 118964877-118964877 on Build GRCh38
rs577013255 Chr.6: 118964859-118964859 on Build GRCh38
rs770479167 Chr.6: 118964709-118964709 on Build GRCh38
rs371174293 Chr.6: 118964657-118964657 on Build GRCh38
rs1012990154 Chr.6: 118964823-118964823 on Build GRCh38
rs370309300 Chr.6: 118964737-118964737 on Build GRCh38
rs777536474 Chr.6: 118964696-118964696 on Build GRCh38
rs1030992093 Chr.6: 118964764-118964764 on Build GRCh38
rs561843233 Chr.6: 118964843-118964843 on Build GRCh38
rs983544110 Chr.6: 118964836-118964836 on Build GRCh38
rs185184863 Chr.6: 118964977-118964977 on Build GRCh38
rs751596500 Chr.6: 118964782-118964782 on Build GRCh38
rs892843205 Chr.6: 118964792-118964792 on Build GRCh38
rs1026500398 Chr.6: 118964883-118964883 on Build GRCh38
rs766496153 Chr.6: 118964775-118964775 on Build GRCh38
rs369860223 Chr.6: 118964791-118964791 on Build GRCh38
rs759566937 Chr.6: 118964717-118964717 on Build GRCh38
rs773957483 Chr.6: 118964640-118964641 on Build GRCh38
rs984895297 Chr.6: 118964959-118964959 on Build GRCh38
rs768057121 Chr.6: 118964796-118964796 on Build GRCh38
rs572964519 Chr.6: 118964724-118964724 on Build GRCh38
rs2275761 Chr.6: 118964623-118964623 on Build GRCh38
rs138876814 Chr.6: 118964658-118964658 on Build GRCh38
rs374741756 Chr.6: 118964788-118964788 on Build GRCh38
rs373300678 Chr.6: 118964642-118964642 on Build GRCh38
rs1039189460 Chr.6: 118965063-118965063 on Build GRCh38
rs557050685 Chr.6: 118964823-118964823 on Build GRCh38
rs763022397 Chr.6: 118964773-118964773 on Build GRCh38
rs529334897 Chr.6: 118965057-118965057 on Build GRCh38
rs758567248 Chr.6: 118964652-118964652 on Build GRCh38
rs542078881 Chr.6: 118964802-118964802 on Build GRCh38
rs558140727 Chr.6: 118964613-118964613 on Build GRCh38
rs546677344 Chr.6: 118964976-118964976 on Build GRCh38
rs533362547 Chr.6: 118964968-118964968 on Build GRCh38
rs115858136 Chr.6: 118964637-118964637 on Build GRCh38
rs923256340 Chr.6: 118965058-118965058 on Build GRCh38
rs564181875 Chr.6: 118964912-118964912 on Build GRCh38
rs544509175 Chr.6: 118964864-118964864 on Build GRCh38
rs934440239 Chr.6: 118964753-118964753 on Build GRCh38
rs900198867 Chr.6: 118965070-118965070 on Build GRCh38
rs372665988 Chr.6: 118964659-118964659 on Build GRCh38
rs567861181 Chr.6: 118964889-118964889 on Build GRCh38
rs773554311 Chr.6: 118964772-118964772 on Build GRCh38
rs928506061 Chr.6: 118964962-118964962 on Build GRCh38
rs981391438 Chr.6: 118964956-118964956 on Build GRCh38
rs949408175 Chr.6: 118964891-118964891 on Build GRCh38
rs947142550 Chr.6: 118965052-118965052 on Build GRCh38
rs781565620 Chr.6: 118964667-118964667 on Build GRCh38
rs755422251 Chr.6: 118964664-118964664 on Build GRCh38
rs773674626 Chr.6: 118964714-118964714 on Build GRCh38
rs964773906 Chr.6: 118964991-118964991 on Build GRCh38
rs374725909 Chr.6: 118964675-118964675 on Build GRCh38
rs748760593 Chr.6: 118964646-118964654 on Build GRCh38
rs763784178 Chr.6: 118964751-118964751 on Build GRCh38
rs770118249 Chr.6: 118964693-118964693 on Build GRCh38
rs1017296451 Chr.6: 118964716-118964716 on Build GRCh38
rs756567719 Chr.6: 118964804-118964804 on Build GRCh38
rs748888206 Chr.6: 118964698-118964698 on Build GRCh38
rs376795227 Chr.6: 118964650-118964650 on Build GRCh38
rs892958890 Chr.6: 118965011-118965011 on Build GRCh38
rs775538811 Chr.6: 118964734-118964734 on Build GRCh38
rs765775033 Chr.6: 118964842-118964842 on Build GRCh38

View Full Product Details