Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 122717707-122718216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKIB
Entrez Gene ID
5570
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor beta
Gene Aliases
PRKACN2
UniGene
Hs.741340
Gene Chromosome Location
Chr.6: 122471917-122726373 on Build GRCh38
Transcripts
NM_001270393.1 NM_001270394.1 NM_001270395.1 NM_032471.5 NM_181794.2 NM_181795.2 XM_011535930.2 XM_011535931.2 XM_011535932.2 XM_011535933.2 XM_011535934.2 XM_011535935.2 XM_011535936.1 XM_011535937.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4913124 c.165A>G p.K55K Chr.6: 122717938-122717938 on Build GRCh38
4349840 c.148G>A p.G50R Chr.6: 122717915-122717915 on Build GRCh38
5727576 c.130G>A p.D44N Chr.6: 122717924-122717924 on Build GRCh38
5404271 c.66C>T p.V22V Chr.6: 122717839-122717839 on Build GRCh38
5869224 c.158C>T p.S53F Chr.6: 122717952-122717952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779165919 Chr.6: 122717789-122717789 on Build GRCh38
rs6900745 Chr.6: 122717743-122717743 on Build GRCh38
rs913670546 Chr.6: 122717861-122717861 on Build GRCh38
rs373640646 Chr.6: 122717839-122717839 on Build GRCh38
rs368144109 Chr.6: 122717840-122717840 on Build GRCh38
rs182893341 Chr.6: 122717925-122717925 on Build GRCh38
rs779218695 Chr.6: 122717930-122717930 on Build GRCh38
rs376807018 Chr.6: 122717996-122717996 on Build GRCh38
rs916107664 Chr.6: 122718119-122718119 on Build GRCh38
rs755530084 Chr.6: 122717916-122717916 on Build GRCh38
rs779001899 Chr.6: 122717985-122717985 on Build GRCh38
rs746002457 Chr.6: 122717795-122717795 on Build GRCh38
rs6920792 Chr.6: 122717742-122717742 on Build GRCh38
rs1018286031 Chr.6: 122718086-122718086 on Build GRCh38
rs187660514 Chr.6: 122717954-122717954 on Build GRCh38
rs113084392 Chr.6: 122717741-122717742 on Build GRCh38
rs548436062 Chr.6: 122717935-122717935 on Build GRCh38
rs780055354 Chr.6: 122717824-122717824 on Build GRCh38
rs552648535 Chr.6: 122717756-122717756 on Build GRCh38
rs955025747 Chr.6: 122718006-122718006 on Build GRCh38
rs192705881 Chr.6: 122717973-122717973 on Build GRCh38
rs897923399 Chr.6: 122717969-122717969 on Build GRCh38
rs766292816 Chr.6: 122717746-122717746 on Build GRCh38
rs762424679 Chr.6: 122717844-122717844 on Build GRCh38
rs370529874 Chr.6: 122717758-122717758 on Build GRCh38
rs989131299 Chr.6: 122717859-122717859 on Build GRCh38
rs200804998 Chr.6: 122717753-122717753 on Build GRCh38
rs948911427 Chr.6: 122718145-122718145 on Build GRCh38
rs143671322 Chr.6: 122717855-122717855 on Build GRCh38
rs972209701 Chr.6: 122717772-122717772 on Build GRCh38
rs759576783 Chr.6: 122717867-122717867 on Build GRCh38
rs371888186 Chr.6: 122717984-122717984 on Build GRCh38
rs772638920 Chr.6: 122717998-122717998 on Build GRCh38
rs1031919345 Chr.6: 122718071-122718071 on Build GRCh38
rs77803694 Chr.6: 122717892-122717892 on Build GRCh38
rs775556782 Chr.6: 122717972-122717972 on Build GRCh38
rs990705154 Chr.6: 122718112-122718112 on Build GRCh38
rs751676406 Chr.6: 122717991-122717991 on Build GRCh38
rs71543256 Chr.6: 122717742-122717743 on Build GRCh38
rs77659800 Chr.6: 122717942-122717942 on Build GRCh38
rs553055285 Chr.6: 122717987-122717987 on Build GRCh38
rs568397309 Chr.6: 122717941-122717941 on Build GRCh38
rs1030424007 Chr.6: 122717808-122717808 on Build GRCh38
rs928397295 Chr.6: 122718162-122718162 on Build GRCh38
rs78311405 Chr.6: 122717826-122717826 on Build GRCh38
rs989552960 Chr.6: 122718164-122718164 on Build GRCh38
rs927772654 Chr.6: 122717909-122717909 on Build GRCh38
rs749074833 Chr.6: 122717873-122717873 on Build GRCh38
rs754904917 Chr.6: 122717751-122717751 on Build GRCh38
rs34684181 Chr.6: 122718056-122718056 on Build GRCh38
rs757472170 Chr.6: 122717784-122717784 on Build GRCh38
rs767511320 Chr.6: 122717870-122717870 on Build GRCh38
rs928968908 Chr.6: 122718166-122718166 on Build GRCh38
rs751045973 Chr.6: 122717857-122717857 on Build GRCh38
rs748975089 Chr.6: 122717944-122717944 on Build GRCh38
rs780368191 Chr.6: 122717807-122717807 on Build GRCh38
rs767631692 Chr.6: 122717970-122717970 on Build GRCh38
rs764897033 Chr.6: 122717732-122717732 on Build GRCh38
rs746579503 Chr.6: 122718155-122718155 on Build GRCh38
rs1047605618 Chr.6: 122717931-122717931 on Build GRCh38
rs772179729 Chr.6: 122717797-122717797 on Build GRCh38
rs750672113 Chr.6: 122717885-122717885 on Build GRCh38
rs752581156 Chr.6: 122717871-122717871 on Build GRCh38
rs955320847 Chr.6: 122718017-122718017 on Build GRCh38
rs373072930 Chr.6: 122717960-122717960 on Build GRCh38
rs541831537 Chr.6: 122718158-122718158 on Build GRCh38
rs749486807 Chr.6: 122717825-122717825 on Build GRCh38
rs954992568 Chr.6: 122717817-122717817 on Build GRCh38
rs765435840 Chr.6: 122717882-122717882 on Build GRCh38
rs781640241 Chr.6: 122717923-122717923 on Build GRCh38
rs936320257 Chr.6: 122717828-122717828 on Build GRCh38
rs879483664 Chr.6: 122717742-122717743 on Build GRCh38
rs541322760 Chr.6: 122717974-122717974 on Build GRCh38
rs148112687 Chr.6: 122717914-122717914 on Build GRCh38
rs758487029 Chr.6: 122717888-122717888 on Build GRCh38
rs201327015 Chr.6: 122717915-122717915 on Build GRCh38
rs564352869 Chr.6: 122717749-122717749 on Build GRCh38
rs868175380 Chr.6: 122718105-122718105 on Build GRCh38
rs141912516 Chr.6: 122717948-122717948 on Build GRCh38

View Full Product Details