Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155264388-155264882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK2
Entrez Gene ID
1196
Gene Name
CDC like kinase 2
Gene Aliases
-
UniGene
Hs.73986
Gene Chromosome Location
Chr.1: 155262868-155273529 on Build GRCh38
Transcripts
NM_001294338.1 NM_001294339.1 NM_003993.3 XM_005244878.1 XM_011509143.1 XM_011509144.1 XM_011509147.2 XM_017000241.1 XM_017000242.1 XM_017000243.1 XM_017000244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987113 c.1093A>T p.S365C Chr.1: 155264518-155264518 on Build GRCh38
2210344 c.1046C>A p.P349Q Chr.1: 155264659-155264659 on Build GRCh38
5950005 c.1100G>A p.G367D Chr.1: 155264511-155264511 on Build GRCh38
6845651 c.947G>A p.S316N Chr.1: 155264758-155264758 on Build GRCh38
222569 c.1003G>A p.E335K Chr.1: 155264702-155264702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377657872 Chr.1: 155264810-155264810 on Build GRCh38
rs764237042 Chr.1: 155264736-155264736 on Build GRCh38
rs765257771 Chr.1: 155264679-155264679 on Build GRCh38
rs761763945 Chr.1: 155264490-155264490 on Build GRCh38
rs781556451 Chr.1: 155264559-155264559 on Build GRCh38
rs756312423 Chr.1: 155264733-155264733 on Build GRCh38
rs773800212 Chr.1: 155264435-155264435 on Build GRCh38
rs34045048 Chr.1: 155264496-155264496 on Build GRCh38
rs768285011 Chr.1: 155264794-155264794 on Build GRCh38
rs950949755 Chr.1: 155264422-155264422 on Build GRCh38
rs760675850 Chr.1: 155264814-155264814 on Build GRCh38
rs765282300 Chr.1: 155264507-155264507 on Build GRCh38
rs56127810 Chr.1: 155264462-155264462 on Build GRCh38
rs199568173 Chr.1: 155264465-155264465 on Build GRCh38
rs746513098 Chr.1: 155264787-155264787 on Build GRCh38
rs749638322 Chr.1: 155264584-155264584 on Build GRCh38
rs765234417 Chr.1: 155264445-155264448 on Build GRCh38
rs139698018 Chr.1: 155264741-155264741 on Build GRCh38
rs192160292 Chr.1: 155264798-155264798 on Build GRCh38
rs545434812 Chr.1: 155264452-155264452 on Build GRCh38
rs761515095 Chr.1: 155264667-155264667 on Build GRCh38
rs779398967 Chr.1: 155264591-155264591 on Build GRCh38
rs773709847 Chr.1: 155264606-155264608 on Build GRCh38
rs771413195 Chr.1: 155264589-155264589 on Build GRCh38
rs146326736 Chr.1: 155264426-155264426 on Build GRCh38
rs370101901 Chr.1: 155264581-155264581 on Build GRCh38
rs374353302 Chr.1: 155264658-155264658 on Build GRCh38
rs751618842 Chr.1: 155264556-155264556 on Build GRCh38
rs752753326 Chr.1: 155264704-155264704 on Build GRCh38
rs565267723 Chr.1: 155264859-155264859 on Build GRCh38
rs540715476 Chr.1: 155264858-155264858 on Build GRCh38
rs775091170 Chr.1: 155264616-155264616 on Build GRCh38
rs760333362 Chr.1: 155264638-155264638 on Build GRCh38
rs775181022 Chr.1: 155264453-155264453 on Build GRCh38
rs779179029 Chr.1: 155264770-155264770 on Build GRCh38
rs779370556 Chr.1: 155264761-155264761 on Build GRCh38
rs748592980 Chr.1: 155264793-155264793 on Build GRCh38
rs538876543 Chr.1: 155264442-155264442 on Build GRCh38
rs759670888 Chr.1: 155264687-155264687 on Build GRCh38
rs771618837 Chr.1: 155264602-155264602 on Build GRCh38
rs899964776 Chr.1: 155264795-155264795 on Build GRCh38
rs758934483 Chr.1: 155264781-155264781 on Build GRCh38
rs767748618 Chr.1: 155264694-155264694 on Build GRCh38
rs755171669 Chr.1: 155264558-155264558 on Build GRCh38
rs773336636 Chr.1: 155264685-155264685 on Build GRCh38
rs559089129 Chr.1: 155264784-155264784 on Build GRCh38
rs557205500 Chr.1: 155264456-155264456 on Build GRCh38
rs745377544 Chr.1: 155264595-155264595 on Build GRCh38
rs979260595 Chr.1: 155264529-155264529 on Build GRCh38
rs776210842 Chr.1: 155264795-155264795 on Build GRCh38
rs975133183 Chr.1: 155264543-155264543 on Build GRCh38
rs554549747 Chr.1: 155264800-155264800 on Build GRCh38
rs773256594 Chr.1: 155264808-155264808 on Build GRCh38
rs915037388 Chr.1: 155264691-155264691 on Build GRCh38
rs753020535 Chr.1: 155264568-155264568 on Build GRCh38
rs144312459 Chr.1: 155264826-155264826 on Build GRCh38
rs749488583 Chr.1: 155264737-155264737 on Build GRCh38
rs1038581662 Chr.1: 155264763-155264763 on Build GRCh38
rs780217124 Chr.1: 155264477-155264477 on Build GRCh38
rs374795363 Chr.1: 155264577-155264577 on Build GRCh38
rs936059878 Chr.1: 155264609-155264609 on Build GRCh38
rs367831473 Chr.1: 155264464-155264464 on Build GRCh38

View Full Product Details