Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134964747-134965278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBS1L
Entrez Gene ID
10767
Gene Name
HBS1 like translational GTPase
Gene Aliases
EF-1a, ERFS, HBS1, HSPC276, eRF3c
UniGene
Hs.378532
Gene Chromosome Location
Chr.6: 134960378-135054898 on Build GRCh38
Transcripts
NM_001145158.1 NM_006620.3

SNP Detail

SNP ID Chromosome Location
rs990937384 Chr.6: 134965152-134965156 on Build GRCh38
rs938712926 Chr.6: 134965163-134965163 on Build GRCh38
rs766251088 Chr.6: 134965232-134965232 on Build GRCh38
rs894970427 Chr.6: 134964862-134964862 on Build GRCh38
rs949235161 Chr.6: 134965203-134965203 on Build GRCh38
rs538672813 Chr.6: 134965159-134965159 on Build GRCh38
rs916479417 Chr.6: 134965180-134965180 on Build GRCh38
rs142376385 Chr.6: 134964981-134964981 on Build GRCh38
rs1022157094 Chr.6: 134964908-134964908 on Build GRCh38
rs538337722 Chr.6: 134964904-134964904 on Build GRCh38
rs767435148 Chr.6: 134965061-134965061 on Build GRCh38
rs952376845 Chr.6: 134965143-134965143 on Build GRCh38
rs903577255 Chr.6: 134964964-134964964 on Build GRCh38
rs966141896 Chr.6: 134964996-134964996 on Build GRCh38
rs397719100 Chr.6: 134964841-134964842 on Build GRCh38
rs571390376 Chr.6: 134965130-134965132 on Build GRCh38
rs186774858 Chr.6: 134964888-134964888 on Build GRCh38
rs554599145 Chr.6: 134964930-134964930 on Build GRCh38
rs546589091 Chr.6: 134965213-134965213 on Build GRCh38
rs748146499 Chr.6: 134965117-134965117 on Build GRCh38
rs1028408785 Chr.6: 134964954-134964954 on Build GRCh38
rs572779523 Chr.6: 134965020-134965020 on Build GRCh38
rs558056680 Chr.6: 134964823-134964824 on Build GRCh38
rs553773467 Chr.6: 134964994-134964994 on Build GRCh38
rs11549504 Chr.6: 134965092-134965092 on Build GRCh38
rs11969408 Chr.6: 134964823-134964823 on Build GRCh38
rs113902835 Chr.6: 134965043-134965043 on Build GRCh38
rs893211864 Chr.6: 134964854-134964854 on Build GRCh38
rs998812200 Chr.6: 134964774-134964774 on Build GRCh38
rs962117102 Chr.6: 134965072-134965072 on Build GRCh38
rs75286696 Chr.6: 134965023-134965023 on Build GRCh38
rs1056821153 Chr.6: 134964845-134964845 on Build GRCh38
rs1029228263 Chr.6: 134965018-134965018 on Build GRCh38
rs984900524 Chr.6: 134965047-134965047 on Build GRCh38
rs987019619 Chr.6: 134965038-134965038 on Build GRCh38
rs909396775 Chr.6: 134965062-134965062 on Build GRCh38
rs57393645 Chr.6: 134964822-134964823 on Build GRCh38
rs187304186 Chr.6: 134965076-134965076 on Build GRCh38
rs940795669 Chr.6: 134964777-134964777 on Build GRCh38
rs191703562 Chr.6: 134965155-134965155 on Build GRCh38
rs76925158 Chr.6: 134964779-134964779 on Build GRCh38
rs201863277 Chr.6: 134964823-134964823 on Build GRCh38
rs907203322 Chr.6: 134964782-134964782 on Build GRCh38
rs879015613 Chr.6: 134964823-134964824 on Build GRCh38
rs140190917 Chr.6: 134964784-134964784 on Build GRCh38
rs529827859 Chr.6: 134965186-134965186 on Build GRCh38
rs112748112 Chr.6: 134965179-134965179 on Build GRCh38
rs191735861 Chr.6: 134964977-134964977 on Build GRCh38
rs976353436 Chr.6: 134965007-134965007 on Build GRCh38
rs1030627952 Chr.6: 134964985-134964985 on Build GRCh38

View Full Product Details