Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136356614-136357130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP7
Entrez Gene ID
9053
Gene Name
microtubule associated protein 7
Gene Aliases
E-MAP-115, EMAP115
UniGene
Hs.486548
Gene Chromosome Location
Chr.6: 136342281-136551015 on Build GRCh38
Transcripts
NM_001198608.1 NM_001198609.1 NM_001198611.1 NM_001198614.1 NM_001198615.1 NM_001198616.1 NM_001198617.1 NM_001198618.1 NM_001198619.1 NM_003980.4 XM_006715598.2 XM_006715599.2 XM_006715600.2 XM_006715601.3 XM_011536243.2 XM_011536244.2 XM_011536245.1 XM_011536246.2 XM_017011471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6478006 c.2012C>T p.P671L Chr.6: 136356761-136356761 on Build GRCh38
6478007 c.1946C>T p.P649L Chr.6: 136356761-136356761 on Build GRCh38
3697376 c.1945C>T p.P649S Chr.6: 136356762-136356762 on Build GRCh38
6406980 c.1987G>A p.A663T Chr.6: 136356786-136356786 on Build GRCh38
3733680 c.1919C>G p.S640C Chr.6: 136356788-136356788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751795272 Chr.6: 136356671-136356671 on Build GRCh38
rs920235449 Chr.6: 136356716-136356716 on Build GRCh38
rs397885336 Chr.6: 136356793-136356794 on Build GRCh38
rs763429507 Chr.6: 136356766-136356766 on Build GRCh38
rs1022497706 Chr.6: 136356897-136356897 on Build GRCh38
rs149970196 Chr.6: 136356760-136356760 on Build GRCh38
rs142810521 Chr.6: 136356997-136357002 on Build GRCh38
rs746534400 Chr.6: 136356795-136356796 on Build GRCh38
rs771653978 Chr.6: 136356755-136356755 on Build GRCh38
rs144889369 Chr.6: 136356761-136356761 on Build GRCh38
rs559114726 Chr.6: 136357005-136357005 on Build GRCh38
rs773853765 Chr.6: 136356765-136356765 on Build GRCh38
rs987317651 Chr.6: 136356652-136356652 on Build GRCh38
rs777652225 Chr.6: 136356721-136356721 on Build GRCh38
rs775090511 Chr.6: 136356757-136356757 on Build GRCh38
rs527451128 Chr.6: 136356777-136356777 on Build GRCh38
rs759769001 Chr.6: 136356801-136356801 on Build GRCh38
rs772041234 Chr.6: 136356837-136356837 on Build GRCh38
rs751017655 Chr.6: 136356643-136356643 on Build GRCh38
rs113911877 Chr.6: 136356924-136356924 on Build GRCh38
rs374285627 Chr.6: 136356649-136356649 on Build GRCh38
rs964615486 Chr.6: 136357031-136357031 on Build GRCh38
rs966810139 Chr.6: 136356907-136356907 on Build GRCh38
rs774028309 Chr.6: 136356737-136356737 on Build GRCh38
rs570702137 Chr.6: 136356839-136356839 on Build GRCh38
rs368916469 Chr.6: 136356804-136356804 on Build GRCh38
rs966069255 Chr.6: 136356983-136356983 on Build GRCh38
rs1008692023 Chr.6: 136356927-136356927 on Build GRCh38
rs781184636 Chr.6: 136356691-136356691 on Build GRCh38
rs777886442 Chr.6: 136356816-136356816 on Build GRCh38
rs911962730 Chr.6: 136356925-136356925 on Build GRCh38
rs1018691950 Chr.6: 136356951-136356951 on Build GRCh38
rs756178200 Chr.6: 136356815-136356815 on Build GRCh38
rs753748851 Chr.6: 136356818-136356818 on Build GRCh38
rs974662571 Chr.6: 136357055-136357055 on Build GRCh38
rs780761202 Chr.6: 136357040-136357040 on Build GRCh38
rs569238352 Chr.6: 136357050-136357050 on Build GRCh38
rs147827400 Chr.6: 136356699-136356699 on Build GRCh38
rs114822509 Chr.6: 136356770-136356770 on Build GRCh38
rs770553006 Chr.6: 136356798-136356803 on Build GRCh38
rs368416191 Chr.6: 136356675-136356675 on Build GRCh38
rs953547183 Chr.6: 136356908-136356908 on Build GRCh38
rs755870251 Chr.6: 136357047-136357047 on Build GRCh38
rs986200745 Chr.6: 136356916-136356916 on Build GRCh38
rs371808838 Chr.6: 136356805-136356805 on Build GRCh38
rs745456930 Chr.6: 136356739-136356739 on Build GRCh38
rs770743019 Chr.6: 136356735-136356735 on Build GRCh38
rs371588242 Chr.6: 136356694-136356694 on Build GRCh38
rs757144312 Chr.6: 136356819-136356819 on Build GRCh38
rs745671784 Chr.6: 136356828-136356828 on Build GRCh38
rs1033693132 Chr.6: 136356821-136356821 on Build GRCh38
rs142693512 Chr.6: 136356825-136356825 on Build GRCh38
rs113554781 Chr.6: 136357016-136357016 on Build GRCh38
rs780706798 Chr.6: 136356819-136356820 on Build GRCh38
rs930532126 Chr.6: 136356725-136356725 on Build GRCh38
rs777694089 Chr.6: 136357093-136357093 on Build GRCh38
rs751878044 Chr.6: 136356800-136356800 on Build GRCh38
rs539121253 Chr.6: 136356853-136356853 on Build GRCh38
rs943254525 Chr.6: 136356679-136356679 on Build GRCh38
rs758758122 Chr.6: 136356645-136356645 on Build GRCh38
rs1047961829 Chr.6: 136356744-136356744 on Build GRCh38
rs1021495950 Chr.6: 136356887-136356887 on Build GRCh38
rs956981212 Chr.6: 136356849-136356849 on Build GRCh38
rs752952554 Chr.6: 136356814-136356814 on Build GRCh38
rs758129133 Chr.6: 136356650-136356650 on Build GRCh38
rs140358034 Chr.6: 136356726-136356726 on Build GRCh38

View Full Product Details