Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136557364-136557875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K5
Entrez Gene ID
4217
Gene Name
mitogen-activated protein kinase kinase kinase 5
Gene Aliases
ASK1, MAPKKK5, MEKK5
UniGene
Hs.186486
Gene Chromosome Location
Chr.6: 136557046-136793098 on Build GRCh38
Transcripts
NM_005923.3 XM_017010872.1 XM_017010873.1 XM_017010875.1 XM_017010877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1073695 c.4109G>A p.R1370Q Chr.6: 136557774-136557774 on Build GRCh38
3620984 c.4066G>A p.G1356R Chr.6: 136557817-136557817 on Build GRCh38
1568059 c.4107T>G p.F1369L Chr.6: 136557776-136557776 on Build GRCh38
1487257 c.4071G>C p.G1357G Chr.6: 136557812-136557812 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs73554162 Chr.6: 136557581-136557581 on Build GRCh38
rs972536796 Chr.6: 136557600-136557600 on Build GRCh38
rs150970264 Chr.6: 136557675-136557675 on Build GRCh38
rs751129150 Chr.6: 136557775-136557775 on Build GRCh38
rs34705939 Chr.6: 136557614-136557614 on Build GRCh38
rs779923697 Chr.6: 136557764-136557764 on Build GRCh38
rs200961701 Chr.6: 136557613-136557613 on Build GRCh38
rs75456458 Chr.6: 136557442-136557442 on Build GRCh38
rs969641868 Chr.6: 136557560-136557560 on Build GRCh38
rs933524994 Chr.6: 136557693-136557693 on Build GRCh38
rs768924733 Chr.6: 136557783-136557783 on Build GRCh38
rs942621949 Chr.6: 136557667-136557667 on Build GRCh38
rs750368502 Chr.6: 136557757-136557757 on Build GRCh38
rs533774529 Chr.6: 136557848-136557848 on Build GRCh38
rs903905768 Chr.6: 136557455-136557455 on Build GRCh38
rs777835597 Chr.6: 136557462-136557462 on Build GRCh38
rs200153510 Chr.6: 136557842-136557842 on Build GRCh38
rs79903899 Chr.6: 136557614-136557614 on Build GRCh38
rs1031582111 Chr.6: 136557480-136557480 on Build GRCh38
rs1006814231 Chr.6: 136557420-136557420 on Build GRCh38
rs546655377 Chr.6: 136557415-136557415 on Build GRCh38
rs761939372 Chr.6: 136557718-136557718 on Build GRCh38
rs765892456 Chr.6: 136557572-136557572 on Build GRCh38
rs960936419 Chr.6: 136557482-136557482 on Build GRCh38
rs377633516 Chr.6: 136557736-136557736 on Build GRCh38
rs529730213 Chr.6: 136557844-136557844 on Build GRCh38
rs902008907 Chr.6: 136557690-136557690 on Build GRCh38
rs769494129 Chr.6: 136557821-136557821 on Build GRCh38
rs999656670 Chr.6: 136557460-136557460 on Build GRCh38
rs747725941 Chr.6: 136557419-136557419 on Build GRCh38
rs915692263 Chr.6: 136557583-136557583 on Build GRCh38
rs1006612270 Chr.6: 136557737-136557737 on Build GRCh38
rs911101649 Chr.6: 136557647-136557647 on Build GRCh38
rs979731938 Chr.6: 136557563-136557563 on Build GRCh38
rs780751982 Chr.6: 136557800-136557800 on Build GRCh38
rs1050925500 Chr.6: 136557704-136557704 on Build GRCh38
rs889094485 Chr.6: 136557411-136557411 on Build GRCh38
rs760748312 Chr.6: 136557759-136557759 on Build GRCh38
rs369638455 Chr.6: 136557806-136557806 on Build GRCh38
rs1035481136 Chr.6: 136557530-136557530 on Build GRCh38
rs538134839 Chr.6: 136557683-136557683 on Build GRCh38
rs971831887 Chr.6: 136557470-136557470 on Build GRCh38
rs140092793 Chr.6: 136557488-136557488 on Build GRCh38
rs933816200 Chr.6: 136557616-136557616 on Build GRCh38
rs779495325 Chr.6: 136557833-136557833 on Build GRCh38
rs1043704233 Chr.6: 136557697-136557697 on Build GRCh38
rs758073603 Chr.6: 136557758-136557758 on Build GRCh38
rs925487768 Chr.6: 136557663-136557663 on Build GRCh38
rs753894742 Chr.6: 136557708-136557708 on Build GRCh38
rs978471337 Chr.6: 136557621-136557621 on Build GRCh38
rs397885833 Chr.6: 136557627-136557627 on Build GRCh38
rs528274047 Chr.6: 136557397-136557397 on Build GRCh38
rs1035595829 Chr.6: 136557797-136557797 on Build GRCh38
rs987048850 Chr.6: 136557633-136557634 on Build GRCh38
rs903899090 Chr.6: 136557711-136557711 on Build GRCh38
rs1023869567 Chr.6: 136557517-136557517 on Build GRCh38

View Full Product Details