Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138775987-138776492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC28A
Entrez Gene ID
25901
Gene Name
coiled-coil domain containing 28A
Gene Aliases
C6orf80, CCRL1AP
UniGene
Hs.412019
Gene Chromosome Location
Chr.6: 138773517-138793319 on Build GRCh38
Transcripts
NM_015439.2 XM_011535728.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4761910 c.278A>T p.E93V Chr.6: 138776128-138776128 on Build GRCh38
252418 c.281G>A p.R94Q Chr.6: 138776131-138776131 on Build GRCh38
6566190 c.363G>C p.V121V Chr.6: 138776213-138776213 on Build GRCh38
1073857 c.231C>A p.V77V Chr.6: 138776081-138776081 on Build GRCh38
290994 c.343delA p.N117fs*5 Chr.6: 138776193-138776193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766471236 Chr.6: 138776267-138776267 on Build GRCh38
rs144069729 Chr.6: 138776385-138776385 on Build GRCh38
rs776898848 Chr.6: 138776226-138776226 on Build GRCh38
rs113967966 Chr.6: 138776403-138776403 on Build GRCh38
rs756347761 Chr.6: 138776193-138776193 on Build GRCh38
rs1051409771 Chr.6: 138776420-138776420 on Build GRCh38
rs751461177 Chr.6: 138776268-138776268 on Build GRCh38
rs919331894 Chr.6: 138776436-138776436 on Build GRCh38
rs954116457 Chr.6: 138776290-138776290 on Build GRCh38
rs941018304 Chr.6: 138776358-138776358 on Build GRCh38
rs984024730 Chr.6: 138776316-138776316 on Build GRCh38
rs369021139 Chr.6: 138776263-138776263 on Build GRCh38
rs909598745 Chr.6: 138776332-138776332 on Build GRCh38
rs2273510 Chr.6: 138776095-138776095 on Build GRCh38
rs367760529 Chr.6: 138776047-138776047 on Build GRCh38
rs1006645685 Chr.6: 138776447-138776447 on Build GRCh38
rs1036683151 Chr.6: 138776368-138776368 on Build GRCh38
rs761106319 Chr.6: 138776204-138776204 on Build GRCh38
rs948490175 Chr.6: 138776276-138776276 on Build GRCh38
rs761780999 Chr.6: 138776149-138776149 on Build GRCh38
rs769348828 Chr.6: 138776261-138776261 on Build GRCh38
rs371434427 Chr.6: 138776096-138776096 on Build GRCh38
rs752495790 Chr.6: 138776330-138776330 on Build GRCh38
rs754039548 Chr.6: 138776119-138776119 on Build GRCh38
rs207467466 Chr.6: 138776131-138776131 on Build GRCh38
rs199985786 Chr.6: 138776221-138776221 on Build GRCh38
rs769309242 Chr.6: 138776069-138776069 on Build GRCh38
rs1032355479 Chr.6: 138776187-138776187 on Build GRCh38
rs371958731 Chr.6: 138776149-138776149 on Build GRCh38
rs963307445 Chr.6: 138776391-138776391 on Build GRCh38
rs748870360 Chr.6: 138776181-138776181 on Build GRCh38
rs904157120 Chr.6: 138776099-138776099 on Build GRCh38
rs765327016 Chr.6: 138776130-138776130 on Build GRCh38
rs746872751 Chr.6: 138776045-138776045 on Build GRCh38
rs780882746 Chr.6: 138776162-138776162 on Build GRCh38
rs373859329 Chr.6: 138776286-138776286 on Build GRCh38
rs761049854 Chr.6: 138776098-138776098 on Build GRCh38
rs1040773 Chr.6: 138776037-138776037 on Build GRCh38
rs781022129 Chr.6: 138776298-138776298 on Build GRCh38
rs145073270 Chr.6: 138776129-138776129 on Build GRCh38
rs968701342 Chr.6: 138776161-138776161 on Build GRCh38
rs188039280 Chr.6: 138776175-138776175 on Build GRCh38
rs765500176 Chr.6: 138776253-138776253 on Build GRCh38
rs1044116171 Chr.6: 138776020-138776020 on Build GRCh38
rs779900224 Chr.6: 138776135-138776135 on Build GRCh38
rs979849915 Chr.6: 138776318-138776318 on Build GRCh38
rs146480742 Chr.6: 138776154-138776154 on Build GRCh38
rs776078449 Chr.6: 138776200-138776200 on Build GRCh38
rs1057347013 Chr.6: 138776014-138776014 on Build GRCh38
rs940965807 Chr.6: 138776039-138776039 on Build GRCh38
rs752169119 Chr.6: 138776089-138776089 on Build GRCh38
rs758371172 Chr.6: 138776036-138776036 on Build GRCh38
rs767679500 Chr.6: 138776156-138776156 on Build GRCh38
rs1017659408 Chr.6: 138776462-138776462 on Build GRCh38
rs762072633 Chr.6: 138776246-138776246 on Build GRCh38
rs201272407 Chr.6: 138776170-138776170 on Build GRCh38
rs758318037 Chr.6: 138776132-138776132 on Build GRCh38
rs772892091 Chr.6: 138776077-138776077 on Build GRCh38
rs750649147 Chr.6: 138776262-138776262 on Build GRCh38
rs760100984 Chr.6: 138776084-138776084 on Build GRCh38
rs748712875 Chr.6: 138776184-138776184 on Build GRCh38
rs770565680 Chr.6: 138776188-138776188 on Build GRCh38
rs557374783 Chr.6: 138776439-138776439 on Build GRCh38
rs776290021 Chr.6: 138776051-138776051 on Build GRCh38
rs542421367 Chr.6: 138776386-138776386 on Build GRCh38
rs183306815 Chr.6: 138776102-138776102 on Build GRCh38
rs747780063 Chr.6: 138776053-138776053 on Build GRCh38
rs568750889 Chr.6: 138776402-138776402 on Build GRCh38
rs917006843 Chr.6: 138776012-138776012 on Build GRCh38
rs769385125 Chr.6: 138776172-138776172 on Build GRCh38
rs750567585 Chr.6: 138776159-138776159 on Build GRCh38
rs764697552 Chr.6: 138776084-138776086 on Build GRCh38

View Full Product Details