Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 142751768-142752268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP2
Entrez Gene ID
3097
Gene Name
human immunodeficiency virus type I enhancer binding protein 2
Gene Aliases
HIV-EP2, MBP-2, MIBP1, MRD43, SHN2, ZAS2, ZNF40B
UniGene
Hs.510172
Gene Chromosome Location
Chr.6: 142751467-142945201 on Build GRCh38
Transcripts
NM_006734.3 XM_017010805.1

SNP Detail

SNP ID Chromosome Location
rs949435354 Chr.6: 142752232-142752232 on Build GRCh38
rs778794464 Chr.6: 142752110-142752111 on Build GRCh38
rs3383 Chr.6: 142751904-142751904 on Build GRCh38
rs966034522 Chr.6: 142751925-142751925 on Build GRCh38
rs890098620 Chr.6: 142752059-142752059 on Build GRCh38
rs995083339 Chr.6: 142752148-142752148 on Build GRCh38
rs900423839 Chr.6: 142751947-142751947 on Build GRCh38
rs963116965 Chr.6: 142752086-142752086 on Build GRCh38
rs902586139 Chr.6: 142751918-142751918 on Build GRCh38
rs553887384 Chr.6: 142752034-142752034 on Build GRCh38
rs936545521 Chr.6: 142751934-142751934 on Build GRCh38
rs572451175 Chr.6: 142752050-142752050 on Build GRCh38
rs920210387 Chr.6: 142751797-142751797 on Build GRCh38
rs1014721639 Chr.6: 142752079-142752079 on Build GRCh38
rs749801301 Chr.6: 142751937-142751937 on Build GRCh38
rs533214311 Chr.6: 142752235-142752235 on Build GRCh38
rs906923180 Chr.6: 142752063-142752063 on Build GRCh38
rs895408505 Chr.6: 142752138-142752138 on Build GRCh38
rs995429294 Chr.6: 142751879-142751879 on Build GRCh38
rs768235227 Chr.6: 142751816-142751816 on Build GRCh38
rs1001165213 Chr.6: 142752067-142752067 on Build GRCh38
rs547926191 Chr.6: 142751830-142751830 on Build GRCh38
rs935909209 Chr.6: 142751965-142751965 on Build GRCh38
rs751480374 Chr.6: 142752117-142752117 on Build GRCh38
rs555136416 Chr.6: 142752089-142752089 on Build GRCh38
rs991061036 Chr.6: 142752208-142752208 on Build GRCh38
rs1028268571 Chr.6: 142752150-142752150 on Build GRCh38
rs536878792 Chr.6: 142751868-142751868 on Build GRCh38
rs564239245 Chr.6: 142752097-142752097 on Build GRCh38
rs767374536 Chr.6: 142752179-142752179 on Build GRCh38
rs569388736 Chr.6: 142751840-142751840 on Build GRCh38
rs1027800815 Chr.6: 142751880-142751880 on Build GRCh38
rs991232180 Chr.6: 142751875-142751875 on Build GRCh38
rs917903506 Chr.6: 142751881-142751881 on Build GRCh38
rs575515522 Chr.6: 142751966-142751966 on Build GRCh38
rs1019045383 Chr.6: 142752168-142752168 on Build GRCh38
rs959616634 Chr.6: 142751869-142751869 on Build GRCh38
rs761893826 Chr.6: 142752166-142752166 on Build GRCh38
rs80298677 Chr.6: 142752223-142752223 on Build GRCh38
rs576573923 Chr.6: 142752167-142752167 on Build GRCh38
rs945832616 Chr.6: 142752033-142752033 on Build GRCh38
rs1054719726 Chr.6: 142752010-142752010 on Build GRCh38
rs938244300 Chr.6: 142751850-142751850 on Build GRCh38
rs760850258 Chr.6: 142752068-142752068 on Build GRCh38
rs543023218 Chr.6: 142752064-142752064 on Build GRCh38
rs188006358 Chr.6: 142752025-142752025 on Build GRCh38
rs143530387 Chr.6: 142751812-142751812 on Build GRCh38
rs974705787 Chr.6: 142751943-142751943 on Build GRCh38
rs763576508 Chr.6: 142752098-142752098 on Build GRCh38
rs769441219 Chr.6: 142751799-142751799 on Build GRCh38
rs949515369 Chr.6: 142751891-142751891 on Build GRCh38
rs550523593 Chr.6: 142751946-142751946 on Build GRCh38
rs950802545 Chr.6: 142751882-142751882 on Build GRCh38
rs371675321 Chr.6: 142752122-142752122 on Build GRCh38
rs911933810 Chr.6: 142751911-142751911 on Build GRCh38

View Full Product Details