Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143471287-143471818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX3
Entrez Gene ID
8504
Gene Name
peroxisomal biogenesis factor 3
Gene Aliases
PBD10A, TRG18
UniGene
Hs.592832
Gene Chromosome Location
Chr.6: 143450781-143490616 on Build GRCh38
Transcripts
NM_003630.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237955 c.457-1G>T p.? Chr.6: 143471382-143471382 on Build GRCh38
6105488 c.505C>T p.Q169* Chr.6: 143471431-143471431 on Build GRCh38
3016985 c.463C>A p.L155I Chr.6: 143471389-143471389 on Build GRCh38
6441561 c.470C>T p.P157L Chr.6: 143471396-143471396 on Build GRCh38
139675 c.472C>T p.P158S Chr.6: 143471398-143471398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748456607 Chr.6: 143471529-143471529 on Build GRCh38
rs62641230 Chr.6: 143471576-143471577 on Build GRCh38
rs17072661 Chr.6: 143471469-143471469 on Build GRCh38
rs969067771 Chr.6: 143471710-143471710 on Build GRCh38
rs750498713 Chr.6: 143471334-143471334 on Build GRCh38
rs780823197 Chr.6: 143471362-143471362 on Build GRCh38
rs763947386 Chr.6: 143471577-143471577 on Build GRCh38
rs145315420 Chr.6: 143471560-143471560 on Build GRCh38
rs947383195 Chr.6: 143471329-143471329 on Build GRCh38
rs767486635 Chr.6: 143471715-143471715 on Build GRCh38
rs769938240 Chr.6: 143471439-143471439 on Build GRCh38
rs888708548 Chr.6: 143471485-143471485 on Build GRCh38
rs748544408 Chr.6: 143471635-143471635 on Build GRCh38
rs914496796 Chr.6: 143471317-143471317 on Build GRCh38
rs749834871 Chr.6: 143471625-143471625 on Build GRCh38
rs911295447 Chr.6: 143471359-143471359 on Build GRCh38
rs754766387 Chr.6: 143471360-143471360 on Build GRCh38
rs776843928 Chr.6: 143471402-143471402 on Build GRCh38
rs1049583661 Chr.6: 143471779-143471779 on Build GRCh38
rs397840972 Chr.6: 143471789-143471790 on Build GRCh38
rs755817677 Chr.6: 143471364-143471364 on Build GRCh38
rs762088940 Chr.6: 143471417-143471417 on Build GRCh38
rs371345383 Chr.6: 143471644-143471644 on Build GRCh38
rs751560045 Chr.6: 143471463-143471463 on Build GRCh38
rs113728509 Chr.6: 143471734-143471734 on Build GRCh38
rs767297580 Chr.6: 143471562-143471562 on Build GRCh38
rs1021511497 Chr.6: 143471704-143471704 on Build GRCh38
rs753426564 Chr.6: 143471501-143471501 on Build GRCh38
rs199781957 Chr.6: 143471399-143471399 on Build GRCh38
rs767407454 Chr.6: 143471466-143471466 on Build GRCh38
rs766261961 Chr.6: 143471447-143471447 on Build GRCh38
rs747441378 Chr.6: 143471384-143471384 on Build GRCh38
rs756845913 Chr.6: 143471614-143471614 on Build GRCh38
rs762883796 Chr.6: 143471339-143471339 on Build GRCh38
rs374172558 Chr.6: 143471517-143471517 on Build GRCh38
rs759587912 Chr.6: 143471551-143471551 on Build GRCh38
rs752833595 Chr.6: 143471341-143471341 on Build GRCh38
rs780566467 Chr.6: 143471375-143471375 on Build GRCh38
rs911252313 Chr.6: 143471782-143471782 on Build GRCh38
rs371647828 Chr.6: 143471366-143471366 on Build GRCh38
rs778264620 Chr.6: 143471518-143471518 on Build GRCh38
rs775444802 Chr.6: 143471568-143471568 on Build GRCh38
rs752448283 Chr.6: 143471363-143471363 on Build GRCh38
rs769037641 Chr.6: 143471522-143471522 on Build GRCh38
rs751333722 Chr.6: 143471352-143471352 on Build GRCh38
rs554784039 Chr.6: 143471546-143471546 on Build GRCh38
rs770025999 Chr.6: 143471531-143471531 on Build GRCh38
rs778092667 Chr.6: 143471657-143471657 on Build GRCh38
rs781628193 Chr.6: 143471633-143471633 on Build GRCh38
rs753593976 Chr.6: 143471595-143471595 on Build GRCh38
rs1010089481 Chr.6: 143471666-143471666 on Build GRCh38
rs1005873123 Chr.6: 143471486-143471486 on Build GRCh38
rs761854963 Chr.6: 143471632-143471632 on Build GRCh38
rs866136282 Chr.6: 143471436-143471436 on Build GRCh38
rs762973381 Chr.6: 143471442-143471442 on Build GRCh38
rs13203340 Chr.6: 143471474-143471474 on Build GRCh38
rs763058857 Chr.6: 143471539-143471539 on Build GRCh38
rs1035449443 Chr.6: 143471557-143471557 on Build GRCh38
rs75545467 Chr.6: 143471357-143471357 on Build GRCh38
rs774381371 Chr.6: 143471610-143471611 on Build GRCh38
rs565641022 Chr.6: 143471476-143471476 on Build GRCh38
rs774283808 Chr.6: 143471548-143471548 on Build GRCh38
rs22908 Chr.6: 143471538-143471538 on Build GRCh38
rs374599874 Chr.6: 143471570-143471570 on Build GRCh38
rs748708593 Chr.6: 143471524-143471524 on Build GRCh38
rs772474699 Chr.6: 143471369-143471369 on Build GRCh38
rs759371755 Chr.6: 143471465-143471465 on Build GRCh38
rs200807211 Chr.6: 143471619-143471619 on Build GRCh38
rs747607754 Chr.6: 143471520-143471520 on Build GRCh38
rs376294031 Chr.6: 143471368-143471368 on Build GRCh38
rs768600934 Chr.6: 143471490-143471493 on Build GRCh38
rs3834741 Chr.6: 143471782-143471783 on Build GRCh38

View Full Product Details