Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 145798282-145798820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO30
Entrez Gene ID
84085
Gene Name
F-box protein 30
Gene Aliases
Fbx30
UniGene
Hs.421095
Gene Chromosome Location
Chr.6: 145798135-145864409 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

Gene Symbol
LOC100507557
Entrez Gene ID
100507557
Gene Name
uncharacterized LOC100507557
Gene Aliases
-
UniGene
Hs.124537
Gene Chromosome Location
Chr.6: 145734869-145886585 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

SNP Detail

SNP ID Chromosome Location
rs554992090 Chr.6: 145798530-145798530 on Build GRCh38
rs1039871675 Chr.6: 145798500-145798500 on Build GRCh38
rs748600367 Chr.6: 145798514-145798515 on Build GRCh38
rs1057088103 Chr.6: 145798316-145798316 on Build GRCh38
rs896286091 Chr.6: 145798777-145798777 on Build GRCh38
rs753765332 Chr.6: 145798586-145798586 on Build GRCh38
rs1042421303 Chr.6: 145798366-145798366 on Build GRCh38
rs529872261 Chr.6: 145798592-145798592 on Build GRCh38
rs991869837 Chr.6: 145798309-145798309 on Build GRCh38
rs934170932 Chr.6: 145798524-145798524 on Build GRCh38
rs997373654 Chr.6: 145798447-145798447 on Build GRCh38
rs1009505048 Chr.6: 145798618-145798618 on Build GRCh38
rs552128909 Chr.6: 145798778-145798778 on Build GRCh38
rs918130458 Chr.6: 145798655-145798655 on Build GRCh38
rs1013437814 Chr.6: 145798785-145798785 on Build GRCh38
rs572151402 Chr.6: 145798433-145798433 on Build GRCh38
rs964890678 Chr.6: 145798445-145798445 on Build GRCh38
rs77674334 Chr.6: 145798553-145798553 on Build GRCh38
rs1009943411 Chr.6: 145798365-145798365 on Build GRCh38
rs1033603424 Chr.6: 145798759-145798759 on Build GRCh38
rs370026681 Chr.6: 145798490-145798490 on Build GRCh38
rs938221444 Chr.6: 145798311-145798311 on Build GRCh38
rs540804998 Chr.6: 145798520-145798520 on Build GRCh38
rs563229477 Chr.6: 145798742-145798742 on Build GRCh38
rs950669475 Chr.6: 145798514-145798514 on Build GRCh38
rs1042702429 Chr.6: 145798722-145798722 on Build GRCh38
rs571793724 Chr.6: 145798688-145798688 on Build GRCh38
rs950332144 Chr.6: 145798363-145798365 on Build GRCh38
rs550006927 Chr.6: 145798723-145798723 on Build GRCh38
rs561105181 Chr.6: 145798526-145798526 on Build GRCh38
rs767724020 Chr.6: 145798564-145798564 on Build GRCh38
rs76957387 Chr.6: 145798401-145798401 on Build GRCh38
rs919017297 Chr.6: 145798355-145798355 on Build GRCh38
rs938154059 Chr.6: 145798635-145798635 on Build GRCh38
rs1017727246 Chr.6: 145798407-145798407 on Build GRCh38
rs1056637613 Chr.6: 145798645-145798645 on Build GRCh38
rs10917 Chr.6: 145798360-145798360 on Build GRCh38
rs896752356 Chr.6: 145798324-145798324 on Build GRCh38
rs191844060 Chr.6: 145798421-145798421 on Build GRCh38
rs774752959 Chr.6: 145798489-145798489 on Build GRCh38
rs778986812 Chr.6: 145798662-145798662 on Build GRCh38
rs754871975 Chr.6: 145798600-145798600 on Build GRCh38
rs532317892 Chr.6: 145798767-145798767 on Build GRCh38
rs903838748 Chr.6: 145798735-145798735 on Build GRCh38
rs945640907 Chr.6: 145798711-145798711 on Build GRCh38
rs910116087 Chr.6: 145798446-145798446 on Build GRCh38
rs903889684 Chr.6: 145798369-145798369 on Build GRCh38

View Full Product Details