Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 150065318-150065828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ULBP3
Entrez Gene ID
79465
Gene Name
UL16 binding protein 3
Gene Aliases
N2DL-3, NKG2DL3, RAET1N
UniGene
Hs.15422
Gene Chromosome Location
Chr.6: 150061046-150069147 on Build GRCh38
Transcripts
NM_024518.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1698130 c.556G>A p.V186I Chr.6: 150065470-150065470 on Build GRCh38
5564928 c.606G>A p.R202R Chr.6: 150065420-150065420 on Build GRCh38
1634567 c.470G>A p.R157K Chr.6: 150065556-150065556 on Build GRCh38
5498944 c.440G>A p.R147Q Chr.6: 150065586-150065586 on Build GRCh38
1568013 c.436G>A p.G146R Chr.6: 150065590-150065590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372155960 Chr.6: 150065587-150065587 on Build GRCh38
rs765630678 Chr.6: 150065444-150065444 on Build GRCh38
rs778446638 Chr.6: 150065550-150065550 on Build GRCh38
rs781112199 Chr.6: 150065718-150065734 on Build GRCh38
rs1047859864 Chr.6: 150065799-150065799 on Build GRCh38
rs781110610 Chr.6: 150065650-150065650 on Build GRCh38
rs375719409 Chr.6: 150065631-150065631 on Build GRCh38
rs776802014 Chr.6: 150065519-150065519 on Build GRCh38
rs1009574127 Chr.6: 150065596-150065596 on Build GRCh38
rs949303010 Chr.6: 150065683-150065683 on Build GRCh38
rs755287168 Chr.6: 150065442-150065442 on Build GRCh38
rs200807610 Chr.6: 150065598-150065598 on Build GRCh38
rs118067943 Chr.6: 150065380-150065380 on Build GRCh38
rs371828950 Chr.6: 150065617-150065617 on Build GRCh38
rs1030539497 Chr.6: 150065743-150065743 on Build GRCh38
rs777495265 Chr.6: 150065643-150065643 on Build GRCh38
rs764618699 Chr.6: 150065531-150065531 on Build GRCh38
rs373258012 Chr.6: 150065523-150065523 on Build GRCh38
rs748024043 Chr.6: 150065501-150065501 on Build GRCh38
rs906768155 Chr.6: 150065466-150065466 on Build GRCh38
rs35696295 Chr.6: 150065524-150065524 on Build GRCh38
rs775283081 Chr.6: 150065371-150065371 on Build GRCh38
rs367950741 Chr.6: 150065539-150065539 on Build GRCh38
rs773401265 Chr.6: 150065436-150065436 on Build GRCh38
rs567313341 Chr.6: 150065387-150065388 on Build GRCh38
rs747663456 Chr.6: 150065590-150065590 on Build GRCh38
rs371776868 Chr.6: 150065494-150065494 on Build GRCh38
rs572135818 Chr.6: 150065777-150065777 on Build GRCh38
rs746935840 Chr.6: 150065644-150065644 on Build GRCh38
rs758190832 Chr.6: 150065641-150065641 on Build GRCh38
rs1037746387 Chr.6: 150065711-150065711 on Build GRCh38
rs144986963 Chr.6: 150065593-150065593 on Build GRCh38
rs929102324 Chr.6: 150065658-150065658 on Build GRCh38
rs752315575 Chr.6: 150065640-150065640 on Build GRCh38
rs762592665 Chr.6: 150065710-150065710 on Build GRCh38
rs200023001 Chr.6: 150065647-150065647 on Build GRCh38
rs771989736 Chr.6: 150065510-150065510 on Build GRCh38
rs764410633 Chr.6: 150065624-150065624 on Build GRCh38
rs375838318 Chr.6: 150065537-150065537 on Build GRCh38
rs780740853 Chr.6: 150065771-150065771 on Build GRCh38
rs769063868 Chr.6: 150065671-150065671 on Build GRCh38
rs370222712 Chr.6: 150065682-150065682 on Build GRCh38
rs540354193 Chr.6: 150065355-150065355 on Build GRCh38
rs752079116 Chr.6: 150065627-150065627 on Build GRCh38
rs201334987 Chr.6: 150065704-150065704 on Build GRCh38
rs368701934 Chr.6: 150065522-150065522 on Build GRCh38
rs778590282 Chr.6: 150065639-150065639 on Build GRCh38
rs899277133 Chr.6: 150065725-150065725 on Build GRCh38
rs762480458 Chr.6: 150065629-150065629 on Build GRCh38
rs929464325 Chr.6: 150065456-150065456 on Build GRCh38
rs747912077 Chr.6: 150065568-150065568 on Build GRCh38
rs936495698 Chr.6: 150065435-150065435 on Build GRCh38
rs778154204 Chr.6: 150065454-150065454 on Build GRCh38
rs756557653 Chr.6: 150065389-150065389 on Build GRCh38
rs35374376 Chr.6: 150065586-150065586 on Build GRCh38
rs774412051 Chr.6: 150065390-150065390 on Build GRCh38
rs34547018 Chr.6: 150065616-150065616 on Build GRCh38
rs993610750 Chr.6: 150065741-150065741 on Build GRCh38
rs556206558 Chr.6: 150065679-150065679 on Build GRCh38
rs755794444 Chr.6: 150065472-150065472 on Build GRCh38
rs759069253 Chr.6: 150065690-150065690 on Build GRCh38
rs769427545 Chr.6: 150065368-150065368 on Build GRCh38
rs760685893 Chr.6: 150065437-150065437 on Build GRCh38
rs768073819 Chr.6: 150065686-150065686 on Build GRCh38
rs761853967 Chr.6: 150065410-150065410 on Build GRCh38
rs773028300 Chr.6: 150065511-150065511 on Build GRCh38
rs187565702 Chr.6: 150065695-150065695 on Build GRCh38
rs376892807 Chr.6: 150065499-150065499 on Build GRCh38
rs1007433768 Chr.6: 150065760-150065760 on Build GRCh38
rs759000459 Chr.6: 150065453-150065453 on Build GRCh38
rs754091822 Chr.6: 150065440-150065440 on Build GRCh38
rs202219794 Chr.6: 150065447-150065447 on Build GRCh38
rs758043059 Chr.6: 150065698-150065698 on Build GRCh38
rs751312870 Chr.6: 150065562-150065562 on Build GRCh38
rs531898596 Chr.6: 150065515-150065515 on Build GRCh38
rs763820344 Chr.6: 150065700-150065700 on Build GRCh38
rs1046424090 Chr.6: 150065513-150065513 on Build GRCh38
rs374852630 Chr.6: 150065594-150065594 on Build GRCh38
rs779800228 Chr.6: 150065470-150065470 on Build GRCh38
rs367595267 Chr.6: 150065604-150065604 on Build GRCh38
rs1042392933 Chr.6: 150065717-150065717 on Build GRCh38
rs745905985 Chr.6: 150065659-150065659 on Build GRCh38
rs892424181 Chr.6: 150065485-150065485 on Build GRCh38
rs148156422 Chr.6: 150065664-150065664 on Build GRCh38
rs771796839 Chr.6: 150065577-150065577 on Build GRCh38
rs753139438 Chr.6: 150065449-150065449 on Build GRCh38
rs767493824 Chr.6: 150065411-150065411 on Build GRCh38
rs145396250 Chr.6: 150065468-150065468 on Build GRCh38
rs763229930 Chr.6: 150065527-150065527 on Build GRCh38
rs773888499 Chr.6: 150065688-150065688 on Build GRCh38
rs766523449 Chr.6: 150065438-150065438 on Build GRCh38
rs1046585526 Chr.6: 150065458-150065458 on Build GRCh38
rs754541250 Chr.6: 150065636-150065636 on Build GRCh38
rs779114566 Chr.6: 150065490-150065499 on Build GRCh38
rs753344720 Chr.6: 150065630-150065630 on Build GRCh38
rs571728197 Chr.6: 150065536-150065536 on Build GRCh38
rs745491892 Chr.6: 150065359-150065359 on Build GRCh38
rs954577035 Chr.6: 150065751-150065751 on Build GRCh38
rs998294088 Chr.6: 150065740-150065740 on Build GRCh38
rs36063202 Chr.6: 150065495-150065495 on Build GRCh38
rs769782946 Chr.6: 150065663-150065663 on Build GRCh38
rs897613442 Chr.6: 150065407-150065407 on Build GRCh38
rs200021594 Chr.6: 150065446-150065446 on Build GRCh38
rs975176039 Chr.6: 150065375-150065375 on Build GRCh38
rs558845393 Chr.6: 150065732-150065732 on Build GRCh38
rs369938957 Chr.6: 150065373-150065373 on Build GRCh38
rs764702466 Chr.6: 150065691-150065691 on Build GRCh38
rs977639148 Chr.6: 150065581-150065581 on Build GRCh38
rs763120446 Chr.6: 150065670-150065670 on Build GRCh38
rs367947532 Chr.6: 150065463-150065463 on Build GRCh38
rs926607248 Chr.6: 150065699-150065699 on Build GRCh38
rs752025572 Chr.6: 150065535-150065535 on Build GRCh38

View Full Product Details