Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 152133142-152133638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
UniGene
Hs.12967
Gene Chromosome Location
Chr.6: 152121684-152637399 on Build GRCh38
Transcripts
NM_033071.3 NM_182961.3 XM_006715407.1 XM_006715408.2 XM_006715409.1 XM_006715410.2 XM_006715411.1 XM_006715412.2 XM_006715413.2 XM_006715414.1 XM_006715415.2 XM_006715416.2 XM_006715417.2 XM_006715420.2 XM_006715421.2 XM_006715422.1 XM_006715423.2 XM_006715424.2 XM_006715425.2 XM_011535641.2 XM_011535642.2 XM_011535643.1 XM_011535644.1 XM_011535645.2 XM_017010608.1 XM_017010609.1 XM_017010610.1 XM_017010611.1 XM_017010612.1 XM_017010613.1 XM_017010614.1 XM_017010615.1 XM_017010616.1 XM_017010617.1 XM_017010618.1 XM_017010619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869531 c.25777C>T p.R8593W Chr.6: 152133356-152133356 on Build GRCh38
5566966 c.25952G>A p.R8651H Chr.6: 152133325-152133325 on Build GRCh38
6490566 c.24699G>A p.V8233V Chr.6: 152133414-152133414 on Build GRCh38
394534 c.25894A>G p.K8632E Chr.6: 152133383-152133383 on Build GRCh38
5034536 c.25810C>T p.H8604Y Chr.6: 152133323-152133323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762570151 Chr.6: 152133509-152133509 on Build GRCh38
rs950849097 Chr.6: 152133582-152133582 on Build GRCh38
rs1020860180 Chr.6: 152133470-152133470 on Build GRCh38
rs546610573 Chr.6: 152133492-152133492 on Build GRCh38
rs751355623 Chr.6: 152133417-152133417 on Build GRCh38
rs201748466 Chr.6: 152133517-152133517 on Build GRCh38
rs893785327 Chr.6: 152133460-152133460 on Build GRCh38
rs763735663 Chr.6: 152133520-152133520 on Build GRCh38
rs1018898565 Chr.6: 152133605-152133605 on Build GRCh38
rs748474553 Chr.6: 152133477-152133477 on Build GRCh38
rs1055624956 Chr.6: 152133432-152133432 on Build GRCh38
rs938109329 Chr.6: 152133427-152133427 on Build GRCh38
rs1034606579 Chr.6: 152133471-152133471 on Build GRCh38
rs764889457 Chr.6: 152133387-152133387 on Build GRCh38
rs762289784 Chr.6: 152133356-152133356 on Build GRCh38
rs745969505 Chr.6: 152133434-152133434 on Build GRCh38
rs775060646 Chr.6: 152133500-152133500 on Build GRCh38
rs748509032 Chr.6: 152133311-152133311 on Build GRCh38
rs1001447509 Chr.6: 152133455-152133455 on Build GRCh38
rs768945531 Chr.6: 152133332-152133332 on Build GRCh38
rs752394397 Chr.6: 152133395-152133395 on Build GRCh38
rs767400686 Chr.6: 152133534-152133534 on Build GRCh38
rs909516634 Chr.6: 152133317-152133317 on Build GRCh38
rs139834542 Chr.6: 152133421-152133421 on Build GRCh38
rs751343627 Chr.6: 152133521-152133521 on Build GRCh38
rs753328975 Chr.6: 152133232-152133232 on Build GRCh38
rs886043195 Chr.6: 152133310-152133310 on Build GRCh38
rs1021311035 Chr.6: 152133202-152133202 on Build GRCh38
rs745877128 Chr.6: 152133303-152133303 on Build GRCh38
rs768914305 Chr.6: 152133467-152133467 on Build GRCh38
rs763862063 Chr.6: 152133412-152133412 on Build GRCh38
rs372823516 Chr.6: 152133308-152133308 on Build GRCh38
rs749543411 Chr.6: 152133461-152133461 on Build GRCh38
rs760367204 Chr.6: 152133261-152133261 on Build GRCh38
rs374954362 Chr.6: 152133245-152133245 on Build GRCh38
rs113393784 Chr.6: 152133239-152133239 on Build GRCh38
rs756313747 Chr.6: 152133437-152133437 on Build GRCh38
rs966274128 Chr.6: 152133614-152133614 on Build GRCh38
rs145939118 Chr.6: 152133268-152133268 on Build GRCh38
rs767524154 Chr.6: 152133324-152133325 on Build GRCh38
rs781435849 Chr.6: 152133294-152133294 on Build GRCh38
rs746958913 Chr.6: 152133272-152133272 on Build GRCh38
rs1011339433 Chr.6: 152133463-152133463 on Build GRCh38
rs376215168 Chr.6: 152133269-152133269 on Build GRCh38
rs191807870 Chr.6: 152133361-152133361 on Build GRCh38
rs983681646 Chr.6: 152133176-152133176 on Build GRCh38
rs17082176 Chr.6: 152133529-152133529 on Build GRCh38
rs780252743 Chr.6: 152133449-152133449 on Build GRCh38
rs370827602 Chr.6: 152133293-152133293 on Build GRCh38
rs1027711302 Chr.6: 152133568-152133568 on Build GRCh38
rs781236317 Chr.6: 152133426-152133426 on Build GRCh38
rs144888773 Chr.6: 152133325-152133325 on Build GRCh38
rs953889160 Chr.6: 152133602-152133602 on Build GRCh38
rs1046624539 Chr.6: 152133386-152133386 on Build GRCh38
rs758037265 Chr.6: 152133399-152133399 on Build GRCh38
rs369718676 Chr.6: 152133252-152133252 on Build GRCh38
rs57299193 Chr.6: 152133604-152133604 on Build GRCh38
rs77839944 Chr.6: 152133254-152133254 on Build GRCh38
rs377119157 Chr.6: 152133476-152133476 on Build GRCh38
rs776119707 Chr.6: 152133365-152133365 on Build GRCh38
rs759004104 Chr.6: 152133370-152133370 on Build GRCh38
rs761146246 Chr.6: 152133483-152133483 on Build GRCh38
rs774776856 Chr.6: 152133355-152133355 on Build GRCh38
rs149045775 Chr.6: 152133326-152133326 on Build GRCh38

View Full Product Details