Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 154086377-154086920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPRM1
Entrez Gene ID
4988
Gene Name
opioid receptor mu 1
Gene Aliases
LMOR, M-OR-1, MOP, MOR, MOR1, OPRM
UniGene
Hs.2353
Gene Chromosome Location
Chr.6: 154010496-154246867 on Build GRCh38
Transcripts
NM_000914.4 NM_001008503.2 NM_001008504.3 NM_001008505.2 NM_001145279.3 NM_001145280.3 NM_001145281.2 NM_001145282.2 NM_001145283.2 NM_001145284.3 NM_001145285.2 NM_001145286.2 NM_001145287.2 NM_001285522.1 NM_001285523.1 NM_001285524.1 NM_001285526.1 NR_104348.1 NR_104349.1 NR_104350.1 NR_104351.1 XM_011535851.2 XM_011535853.2 XM_011535856.2 XM_011535862.2 XM_017010903.1 XM_017010904.1 XM_017010905.1 XM_017010906.1

SNP Detail

SNP ID Chromosome Location
rs971974052 Chr.6: 154086732-154086732 on Build GRCh38
rs1011840062 Chr.6: 154086570-154086571 on Build GRCh38
rs776133638 Chr.6: 154086655-154086655 on Build GRCh38
rs953333139 Chr.6: 154086683-154086683 on Build GRCh38
rs967049148 Chr.6: 154086612-154086612 on Build GRCh38
rs184521684 Chr.6: 154086874-154086874 on Build GRCh38
rs543305101 Chr.6: 154086782-154086782 on Build GRCh38
rs1011808971 Chr.6: 154086822-154086822 on Build GRCh38
rs760133254 Chr.6: 154086567-154086567 on Build GRCh38
rs893356887 Chr.6: 154086552-154086552 on Build GRCh38
rs77402678 Chr.6: 154086539-154086539 on Build GRCh38
rs143211901 Chr.6: 154086490-154086490 on Build GRCh38
rs929324108 Chr.6: 154086517-154086517 on Build GRCh38
rs879889955 Chr.6: 154086687-154086687 on Build GRCh38
rs190580238 Chr.6: 154086875-154086875 on Build GRCh38
rs535029421 Chr.6: 154086515-154086515 on Build GRCh38
rs116467637 Chr.6: 154086524-154086524 on Build GRCh38
rs771358109 Chr.6: 154086538-154086538 on Build GRCh38
rs992374731 Chr.6: 154086452-154086452 on Build GRCh38
rs573267895 Chr.6: 154086833-154086833 on Build GRCh38
rs930037941 Chr.6: 154086468-154086468 on Build GRCh38
rs937431474 Chr.6: 154086774-154086774 on Build GRCh38
rs909171246 Chr.6: 154086424-154086427 on Build GRCh38
rs909046599 Chr.6: 154086686-154086686 on Build GRCh38
rs899636810 Chr.6: 154086565-154086565 on Build GRCh38
rs1027003999 Chr.6: 154086764-154086764 on Build GRCh38
rs772638457 Chr.6: 154086582-154086582 on Build GRCh38
rs148337066 Chr.6: 154086684-154086684 on Build GRCh38
rs567505857 Chr.6: 154086502-154086502 on Build GRCh38
rs1038298587 Chr.6: 154086462-154086462 on Build GRCh38
rs1033852015 Chr.6: 154086736-154086736 on Build GRCh38
rs1001921727 Chr.6: 154086689-154086689 on Build GRCh38
rs141462963 Chr.6: 154086745-154086745 on Build GRCh38
rs761440939 Chr.6: 154086814-154086814 on Build GRCh38
rs35303787 Chr.6: 154086437-154086437 on Build GRCh38
rs1009672121 Chr.6: 154086677-154086677 on Build GRCh38
rs563649 Chr.6: 154086832-154086832 on Build GRCh38
rs115429619 Chr.6: 154086866-154086866 on Build GRCh38
rs200072913 Chr.6: 154086424-154086424 on Build GRCh38
rs1046211840 Chr.6: 154086759-154086759 on Build GRCh38
rs192516541 Chr.6: 154086637-154086637 on Build GRCh38
rs6557336 Chr.6: 154086409-154086409 on Build GRCh38
rs1056443502 Chr.6: 154086549-154086549 on Build GRCh38
rs750123023 Chr.6: 154086465-154086465 on Build GRCh38
rs188150554 Chr.6: 154086403-154086403 on Build GRCh38
rs892370166 Chr.6: 154086670-154086670 on Build GRCh38
rs918589189 Chr.6: 154086464-154086464 on Build GRCh38
rs963226002 Chr.6: 154086702-154086702 on Build GRCh38
rs1049038083 Chr.6: 154086519-154086519 on Build GRCh38
rs950971309 Chr.6: 154086772-154086772 on Build GRCh38
rs755090496 Chr.6: 154086645-154086648 on Build GRCh38
rs992312468 Chr.6: 154086755-154086755 on Build GRCh38

View Full Product Details