Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160442671-160443184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A3
Entrez Gene ID
6581
Gene Name
solute carrier family 22 member 3
Gene Aliases
EMT, EMTH, OCT3
UniGene
Hs.567337
Gene Chromosome Location
Chr.6: 160348373-160454982 on Build GRCh38
Transcripts
NM_021977.3 XM_005267106.4 XM_011536075.2 XM_011536076.2 XM_017011203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7390236 c.1305G>C p.R435S Chr.6: 160442777-160442777 on Build GRCh38
5061601 c.1334G>T p.G445V Chr.6: 160442806-160442806 on Build GRCh38
3622531 c.1289G>A p.G430E Chr.6: 160442761-160442761 on Build GRCh38
285043 c.1294G>T p.A432S Chr.6: 160442766-160442766 on Build GRCh38
1226058 c.1353A>C p.E451D Chr.6: 160442825-160442825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564220215 Chr.6: 160442855-160442855 on Build GRCh38
rs549975194 Chr.6: 160442953-160442953 on Build GRCh38
rs773205025 Chr.6: 160442745-160442745 on Build GRCh38
rs756038430 Chr.6: 160442814-160442814 on Build GRCh38
rs76556187 Chr.6: 160443086-160443086 on Build GRCh38
rs953880219 Chr.6: 160443122-160443122 on Build GRCh38
rs748144815 Chr.6: 160442796-160442796 on Build GRCh38
rs748370844 Chr.6: 160442884-160442884 on Build GRCh38
rs536089083 Chr.6: 160443116-160443116 on Build GRCh38
rs374221836 Chr.6: 160442740-160442740 on Build GRCh38
rs748284851 Chr.6: 160442737-160442737 on Build GRCh38
rs8187721 Chr.6: 160442886-160442886 on Build GRCh38
rs149424049 Chr.6: 160442764-160442764 on Build GRCh38
rs749901413 Chr.6: 160442806-160442806 on Build GRCh38
rs539124761 Chr.6: 160443009-160443009 on Build GRCh38
rs768635320 Chr.6: 160442723-160442723 on Build GRCh38
rs777734604 Chr.6: 160442889-160442889 on Build GRCh38
rs180882957 Chr.6: 160442701-160442701 on Build GRCh38
rs11962480 Chr.6: 160442972-160442972 on Build GRCh38
rs767149577 Chr.6: 160442804-160442804 on Build GRCh38
rs1029358571 Chr.6: 160443101-160443101 on Build GRCh38
rs932582921 Chr.6: 160442827-160442827 on Build GRCh38
rs777186082 Chr.6: 160442907-160442907 on Build GRCh38
rs747482147 Chr.6: 160442748-160442748 on Build GRCh38
rs146494202 Chr.6: 160442967-160442967 on Build GRCh38
rs772134683 Chr.6: 160443100-160443100 on Build GRCh38
rs776535631 Chr.6: 160442777-160442777 on Build GRCh38
rs570481482 Chr.6: 160442994-160442994 on Build GRCh38
rs368718347 Chr.6: 160442768-160442768 on Build GRCh38
rs912462004 Chr.6: 160442869-160442869 on Build GRCh38
rs946636510 Chr.6: 160443050-160443050 on Build GRCh38
rs939912211 Chr.6: 160443149-160443149 on Build GRCh38
rs771417375 Chr.6: 160442752-160442752 on Build GRCh38
rs12212246 Chr.6: 160442788-160442788 on Build GRCh38
rs370786751 Chr.6: 160442850-160442850 on Build GRCh38
rs190324252 Chr.6: 160443107-160443107 on Build GRCh38
rs972896661 Chr.6: 160442909-160442909 on Build GRCh38
rs1042187625 Chr.6: 160443075-160443075 on Build GRCh38
rs373997883 Chr.6: 160442868-160442868 on Build GRCh38
rs144088841 Chr.6: 160442891-160442891 on Build GRCh38
rs17522235 Chr.6: 160443158-160443158 on Build GRCh38
rs746290271 Chr.6: 160442913-160442913 on Build GRCh38
rs771394311 Chr.6: 160442894-160442894 on Build GRCh38
rs986594357 Chr.6: 160443139-160443139 on Build GRCh38
rs761491332 Chr.6: 160442791-160442791 on Build GRCh38
rs377528900 Chr.6: 160442769-160442769 on Build GRCh38
rs549854721 Chr.6: 160443083-160443083 on Build GRCh38
rs775939483 Chr.6: 160442920-160442920 on Build GRCh38
rs762581168 Chr.6: 160442793-160442793 on Build GRCh38
rs199817918 Chr.6: 160442863-160442863 on Build GRCh38
rs912493002 Chr.6: 160443144-160443144 on Build GRCh38
rs893698684 Chr.6: 160443008-160443008 on Build GRCh38
rs779170058 Chr.6: 160442882-160442882 on Build GRCh38
rs1054886238 Chr.6: 160442984-160442984 on Build GRCh38
rs944693795 Chr.6: 160442870-160442870 on Build GRCh38
rs531535674 Chr.6: 160442919-160442919 on Build GRCh38
rs937314176 Chr.6: 160442974-160442974 on Build GRCh38
rs779084136 Chr.6: 160442735-160442735 on Build GRCh38
rs555627036 Chr.6: 160443148-160443148 on Build GRCh38
rs527260652 Chr.6: 160442968-160442968 on Build GRCh38

View Full Product Details