Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169706063-169706589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf120
Entrez Gene ID
387263
Gene Name
chromosome 6 open reading frame 120
Gene Aliases
-
UniGene
Hs.591375
Gene Chromosome Location
Chr.6: 169702112-169706358 on Build GRCh38
Transcripts
NM_001029863.2 NM_001317342.1 NM_018288.3 NM_133325.2 XM_017010998.1

Gene Symbol
PHF10
Entrez Gene ID
55274
Gene Name
PHD finger protein 10
Gene Aliases
BAF45A, XAP135
UniGene
Hs.435933
Gene Chromosome Location
Chr.6: 169703902-169724049 on Build GRCh38
Transcripts
NM_001029863.2 NM_001317342.1 NM_018288.3 NM_133325.2 XM_017010998.1

SNP Detail

SNP ID Chromosome Location
rs751808393 Chr.6: 169706279-169706279 on Build GRCh38
rs930722638 Chr.6: 169706264-169706264 on Build GRCh38
rs985749894 Chr.6: 169706375-169706375 on Build GRCh38
rs75557160 Chr.6: 169706177-169706177 on Build GRCh38
rs143848302 Chr.6: 169706281-169706281 on Build GRCh38
rs1018731633 Chr.6: 169706173-169706173 on Build GRCh38
rs200942930 Chr.6: 169706412-169706413 on Build GRCh38
rs192531475 Chr.6: 169706374-169706374 on Build GRCh38
rs551445835 Chr.6: 169706460-169706460 on Build GRCh38
rs944961057 Chr.6: 169706399-169706399 on Build GRCh38
rs1056460462 Chr.6: 169706528-169706528 on Build GRCh38
rs566976879 Chr.6: 169706478-169706478 on Build GRCh38
rs868143188 Chr.6: 169706390-169706390 on Build GRCh38
rs748685155 Chr.6: 169706454-169706454 on Build GRCh38
rs986305521 Chr.6: 169706161-169706161 on Build GRCh38
rs1039695544 Chr.6: 169706408-169706411 on Build GRCh38
rs1020091586 Chr.6: 169706327-169706327 on Build GRCh38
rs897930826 Chr.6: 169706260-169706260 on Build GRCh38
rs185238646 Chr.6: 169706274-169706274 on Build GRCh38
rs1032641539 Chr.6: 169706342-169706342 on Build GRCh38
rs550968783 Chr.6: 169706354-169706357 on Build GRCh38
rs977590754 Chr.6: 169706222-169706223 on Build GRCh38
rs560128917 Chr.6: 169706338-169706338 on Build GRCh38
rs905275750 Chr.6: 169706518-169706518 on Build GRCh38
rs967545094 Chr.6: 169706329-169706329 on Build GRCh38
rs35892429 Chr.6: 169706089-169706092 on Build GRCh38
rs541874856 Chr.6: 169706266-169706266 on Build GRCh38
rs762252163 Chr.6: 169706297-169706297 on Build GRCh38
rs1048888075 Chr.6: 169706304-169706307 on Build GRCh38
rs545197034 Chr.6: 169706299-169706299 on Build GRCh38
rs991149649 Chr.6: 169706255-169706255 on Build GRCh38
rs978446064 Chr.6: 169706337-169706337 on Build GRCh38
rs538396885 Chr.6: 169706521-169706521 on Build GRCh38
rs34033798 Chr.6: 169706324-169706325 on Build GRCh38
rs967929263 Chr.6: 169706222-169706222 on Build GRCh38
rs1029928257 Chr.6: 169706217-169706217 on Build GRCh38
rs1051747877 Chr.6: 169706450-169706451 on Build GRCh38
rs1014527758 Chr.6: 169706312-169706312 on Build GRCh38
rs981132602 Chr.6: 169706552-169706553 on Build GRCh38
rs190482358 Chr.6: 169706291-169706291 on Build GRCh38
rs895740187 Chr.6: 169706532-169706532 on Build GRCh38
rs531356056 Chr.6: 169706398-169706398 on Build GRCh38
rs1022723808 Chr.6: 169706530-169706536 on Build GRCh38
rs563675261 Chr.6: 169706290-169706290 on Build GRCh38
rs80278708 Chr.6: 169706191-169706191 on Build GRCh38
rs779644835 Chr.6: 169706140-169706140 on Build GRCh38
rs919406643 Chr.6: 169706453-169706453 on Build GRCh38
rs530914580 Chr.6: 169706412-169706412 on Build GRCh38
rs540620382 Chr.6: 169706206-169706206 on Build GRCh38
rs915584431 Chr.6: 169706259-169706259 on Build GRCh38
rs540028017 Chr.6: 169706175-169706175 on Build GRCh38
rs768194983 Chr.6: 169706199-169706199 on Build GRCh38
rs562662178 Chr.6: 169706285-169706285 on Build GRCh38
rs999017755 Chr.6: 169706488-169706488 on Build GRCh38
rs527478461 Chr.6: 169706352-169706352 on Build GRCh38
rs571330532 Chr.6: 169706445-169706445 on Build GRCh38
rs373331512 Chr.6: 169706304-169706305 on Build GRCh38
rs1049105960 Chr.6: 169706278-169706278 on Build GRCh38
rs3924590 Chr.6: 169706223-169706223 on Build GRCh38
rs1032730637 Chr.6: 169706223-169706223 on Build GRCh38
rs774347919 Chr.6: 169706434-169706436 on Build GRCh38
rs373607986 Chr.6: 169706461-169706461 on Build GRCh38
rs796897767 Chr.6: 169706325-169706326 on Build GRCh38

View Full Product Details