Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23256602-23257108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817178 c.519C>A p.F173L Chr.7: 23257043-23257043 on Build GRCh38
7223797 c.525C>T p.Y175Y Chr.7: 23257049-23257049 on Build GRCh38
3880266 c.463G>A p.V155I Chr.7: 23256987-23256987 on Build GRCh38
3880267 c.532C>A p.H178N Chr.7: 23257056-23257056 on Build GRCh38
6177314 c.382G>T p.A128S Chr.7: 23256906-23256906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015607354 Chr.7: 23256994-23256994 on Build GRCh38
rs767049394 Chr.7: 23256896-23256897 on Build GRCh38
rs775142701 Chr.7: 23257066-23257066 on Build GRCh38
rs965874755 Chr.7: 23256946-23256946 on Build GRCh38
rs749437746 Chr.7: 23256938-23256938 on Build GRCh38
rs146184286 Chr.7: 23256945-23256945 on Build GRCh38
rs1027821703 Chr.7: 23256760-23256760 on Build GRCh38
rs757763556 Chr.7: 23256870-23256870 on Build GRCh38
rs745336323 Chr.7: 23257057-23257057 on Build GRCh38
rs1003608501 Chr.7: 23256680-23256680 on Build GRCh38
rs1013757663 Chr.7: 23256823-23256823 on Build GRCh38
rs759624126 Chr.7: 23256954-23256954 on Build GRCh38
rs368466809 Chr.7: 23257021-23257021 on Build GRCh38
rs774172804 Chr.7: 23256894-23256894 on Build GRCh38
rs753928951 Chr.7: 23257023-23257023 on Build GRCh38
rs759090251 Chr.7: 23256979-23256979 on Build GRCh38
rs959338858 Chr.7: 23256691-23256691 on Build GRCh38
rs1009691008 Chr.7: 23256943-23256943 on Build GRCh38
rs1036938772 Chr.7: 23256779-23256779 on Build GRCh38
rs769203451 Chr.7: 23257062-23257062 on Build GRCh38
rs752919998 Chr.7: 23257057-23257057 on Build GRCh38
rs750909778 Chr.7: 23256925-23256925 on Build GRCh38
rs750886164 Chr.7: 23257005-23257005 on Build GRCh38
rs913103299 Chr.7: 23256652-23256652 on Build GRCh38
rs1042259382 Chr.7: 23256663-23256663 on Build GRCh38
rs746852051 Chr.7: 23257046-23257046 on Build GRCh38
rs375038413 Chr.7: 23256993-23256993 on Build GRCh38
rs758693391 Chr.7: 23256858-23256858 on Build GRCh38
rs367885589 Chr.7: 23256906-23256906 on Build GRCh38
rs1035086205 Chr.7: 23256681-23256681 on Build GRCh38
rs966207289 Chr.7: 23257006-23257006 on Build GRCh38
rs753498835 Chr.7: 23256841-23256841 on Build GRCh38
rs765420850 Chr.7: 23257031-23257031 on Build GRCh38
rs373796409 Chr.7: 23256886-23256886 on Build GRCh38
rs908064552 Chr.7: 23256807-23256807 on Build GRCh38
rs777412652 Chr.7: 23257044-23257044 on Build GRCh38
rs770827735 Chr.7: 23257049-23257049 on Build GRCh38
rs74758879 Chr.7: 23256812-23256812 on Build GRCh38
rs191779242 Chr.7: 23256986-23256986 on Build GRCh38
rs199355 Chr.7: 23256914-23256914 on Build GRCh38
rs976473026 Chr.7: 23256836-23256836 on Build GRCh38
rs745923611 Chr.7: 23256875-23256875 on Build GRCh38
rs371662186 Chr.7: 23256968-23256968 on Build GRCh38
rs529106597 Chr.7: 23256917-23256917 on Build GRCh38
rs956997820 Chr.7: 23256915-23256915 on Build GRCh38
rs1032857611 Chr.7: 23256633-23256633 on Build GRCh38
rs746942137 Chr.7: 23256961-23256961 on Build GRCh38
rs747926273 Chr.7: 23256947-23256947 on Build GRCh38
rs200717222 Chr.7: 23256987-23256987 on Build GRCh38
rs770018869 Chr.7: 23256877-23256877 on Build GRCh38
rs908945150 Chr.7: 23256767-23256767 on Build GRCh38
rs779137072 Chr.7: 23257029-23257029 on Build GRCh38
rs201016099 Chr.7: 23256954-23256954 on Build GRCh38
rs34247813 Chr.7: 23256808-23256808 on Build GRCh38
rs571818260 Chr.7: 23257001-23257001 on Build GRCh38
rs1027688361 Chr.7: 23256985-23256985 on Build GRCh38
rs767032879 Chr.7: 23257022-23257022 on Build GRCh38
rs772004069 Chr.7: 23256899-23256899 on Build GRCh38
rs902585243 Chr.7: 23256672-23256672 on Build GRCh38
rs142237543 Chr.7: 23256752-23256752 on Build GRCh38
rs949573834 Chr.7: 23257065-23257065 on Build GRCh38
rs915136487 Chr.7: 23256882-23256882 on Build GRCh38
rs372073674 Chr.7: 23257035-23257035 on Build GRCh38
rs780367013 Chr.7: 23256878-23256878 on Build GRCh38
rs765876485 Chr.7: 23256904-23256904 on Build GRCh38
rs776093177 Chr.7: 23256969-23256969 on Build GRCh38
rs989037046 Chr.7: 23256645-23256645 on Build GRCh38
rs753964158 Chr.7: 23256911-23256921 on Build GRCh38
rs902320388 Chr.7: 23256936-23256936 on Build GRCh38
rs756199110 Chr.7: 23256929-23256929 on Build GRCh38
rs939201212 Chr.7: 23256937-23256937 on Build GRCh38
rs781593180 Chr.7: 23256874-23256874 on Build GRCh38
rs765260216 Chr.7: 23256806-23256806 on Build GRCh38
rs760117940 Chr.7: 23256902-23256902 on Build GRCh38
rs758710963 Chr.7: 23257040-23257040 on Build GRCh38
rs952368091 Chr.7: 23256775-23256775 on Build GRCh38
rs763865368 Chr.7: 23257070-23257070 on Build GRCh38
rs764851389 Chr.7: 23256913-23256913 on Build GRCh38
rs755032435 Chr.7: 23257026-23257026 on Build GRCh38
rs201025235 Chr.7: 23256883-23256883 on Build GRCh38
rs199354 Chr.7: 23256953-23256953 on Build GRCh38
rs759640856 Chr.7: 23256702-23256702 on Build GRCh38
rs756675751 Chr.7: 23257011-23257011 on Build GRCh38
rs533281446 Chr.7: 23256880-23256880 on Build GRCh38
rs773044433 Chr.7: 23256901-23256901 on Build GRCh38
rs773690328 Chr.7: 23257081-23257081 on Build GRCh38
rs558951249 Chr.7: 23256644-23256644 on Build GRCh38
rs138282862 Chr.7: 23257050-23257050 on Build GRCh38
rs774135885 Chr.7: 23257075-23257076 on Build GRCh38
rs922357947 Chr.7: 23256837-23256837 on Build GRCh38

View Full Product Details