Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27094513-27095018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA1
Entrez Gene ID
3198
Gene Name
homeobox A1
Gene Aliases
BSAS, HOX1, HOX1F
UniGene
Hs.67397
Gene Chromosome Location
Chr.7: 27092993-27096006 on Build GRCh38
Transcripts
NM_005522.4 NM_153620.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5572447 c.669C>T p.Y223Y Chr.7: 27094779-27094779 on Build GRCh38
6339881 c.706T>A p.F236I Chr.7: 27094742-27094742 on Build GRCh38
3923432 c.654G>A p.G218G Chr.7: 27094794-27094794 on Build GRCh38
6447468 c.764C>T p.T255M Chr.7: 27094684-27094684 on Build GRCh38
1698450 c.671G>A p.G224D Chr.7: 27094777-27094777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767117973 Chr.7: 27094738-27094738 on Build GRCh38
rs576079236 Chr.7: 27094814-27094814 on Build GRCh38
rs765041517 Chr.7: 27094704-27094704 on Build GRCh38
rs138651027 Chr.7: 27094683-27094683 on Build GRCh38
rs780457782 Chr.7: 27094676-27094676 on Build GRCh38
rs888833693 Chr.7: 27094964-27094964 on Build GRCh38
rs747572885 Chr.7: 27094845-27094845 on Build GRCh38
rs572556009 Chr.7: 27094846-27094846 on Build GRCh38
rs896210855 Chr.7: 27094966-27094966 on Build GRCh38
rs756302694 Chr.7: 27094549-27094549 on Build GRCh38
rs1034749835 Chr.7: 27094662-27094662 on Build GRCh38
rs770156271 Chr.7: 27094566-27094566 on Build GRCh38
rs753632709 Chr.7: 27094646-27094646 on Build GRCh38
rs762883582 Chr.7: 27094710-27094710 on Build GRCh38
rs375418482 Chr.7: 27094569-27094569 on Build GRCh38
rs373337108 Chr.7: 27094626-27094626 on Build GRCh38
rs111259912 Chr.7: 27094856-27094856 on Build GRCh38
rs779419910 Chr.7: 27094757-27094757 on Build GRCh38
rs746134151 Chr.7: 27094773-27094773 on Build GRCh38
rs779609688 Chr.7: 27094865-27094865 on Build GRCh38
rs780133703 Chr.7: 27094710-27094712 on Build GRCh38
rs776669653 Chr.7: 27094772-27094772 on Build GRCh38
rs750655418 Chr.7: 27094581-27094583 on Build GRCh38
rs777296934 Chr.7: 27094669-27094669 on Build GRCh38
rs777453698 Chr.7: 27094558-27094558 on Build GRCh38
rs113439849 Chr.7: 27094573-27094573 on Build GRCh38
rs41311763 Chr.7: 27094651-27094651 on Build GRCh38
rs746342260 Chr.7: 27094927-27094927 on Build GRCh38
rs772208835 Chr.7: 27094688-27094688 on Build GRCh38
rs762869205 Chr.7: 27094797-27094797 on Build GRCh38
rs755351481 Chr.7: 27094744-27094744 on Build GRCh38
rs372137447 Chr.7: 27094567-27094567 on Build GRCh38
rs753177962 Chr.7: 27094823-27094823 on Build GRCh38
rs34462126 Chr.7: 27094764-27094764 on Build GRCh38
rs778380747 Chr.7: 27094539-27094539 on Build GRCh38
rs923347367 Chr.7: 27094781-27094781 on Build GRCh38
rs867291429 Chr.7: 27094787-27094787 on Build GRCh38
rs773883585 Chr.7: 27094802-27094802 on Build GRCh38
rs143597165 Chr.7: 27094707-27094707 on Build GRCh38
rs552669601 Chr.7: 27094790-27094790 on Build GRCh38
rs1012004915 Chr.7: 27094984-27094984 on Build GRCh38
rs765197009 Chr.7: 27094658-27094658 on Build GRCh38
rs183033372 Chr.7: 27094671-27094671 on Build GRCh38
rs1016469881 Chr.7: 27094974-27094974 on Build GRCh38
rs760937523 Chr.7: 27094689-27094689 on Build GRCh38
rs764089470 Chr.7: 27094801-27094801 on Build GRCh38
rs778191046 Chr.7: 27094837-27094837 on Build GRCh38
rs370033913 Chr.7: 27094769-27094769 on Build GRCh38
rs141900601 Chr.7: 27094696-27094696 on Build GRCh38
rs771124603 Chr.7: 27094693-27094693 on Build GRCh38
rs761281990 Chr.7: 27094807-27094807 on Build GRCh38
rs768343894 Chr.7: 27094949-27094949 on Build GRCh38
rs752182067 Chr.7: 27094664-27094664 on Build GRCh38
rs902455204 Chr.7: 27094884-27094884 on Build GRCh38
rs17465302 Chr.7: 27094830-27094830 on Build GRCh38
rs999346191 Chr.7: 27094893-27094893 on Build GRCh38
rs772297076 Chr.7: 27094607-27094607 on Build GRCh38
rs765680274 Chr.7: 27094816-27094816 on Build GRCh38
rs749684801 Chr.7: 27094679-27094679 on Build GRCh38
rs746688404 Chr.7: 27094565-27094565 on Build GRCh38
rs747097748 Chr.7: 27094767-27094767 on Build GRCh38
rs749973941 Chr.7: 27094810-27094810 on Build GRCh38
rs758942349 Chr.7: 27094836-27094836 on Build GRCh38
rs41303378 Chr.7: 27094553-27094553 on Build GRCh38
rs976248101 Chr.7: 27094778-27094778 on Build GRCh38
rs755681153 Chr.7: 27094546-27094546 on Build GRCh38
rs960473055 Chr.7: 27094665-27094665 on Build GRCh38
rs200421049 Chr.7: 27094670-27094670 on Build GRCh38
rs752269739 Chr.7: 27094548-27094548 on Build GRCh38
rs769512832 Chr.7: 27094779-27094779 on Build GRCh38
rs762030849 Chr.7: 27094586-27094586 on Build GRCh38
rs763538049 Chr.7: 27094722-27094722 on Build GRCh38
rs556902625 Chr.7: 27094697-27094697 on Build GRCh38
rs144812662 Chr.7: 27094574-27094574 on Build GRCh38
rs759512026 Chr.7: 27094654-27094654 on Build GRCh38
rs756293597 Chr.7: 27094658-27094671 on Build GRCh38
rs760807570 Chr.7: 27094642-27094642 on Build GRCh38
rs886062257 Chr.7: 27094623-27094623 on Build GRCh38
rs759032142 Chr.7: 27094762-27094762 on Build GRCh38
rs766139746 Chr.7: 27094644-27094644 on Build GRCh38
rs377716509 Chr.7: 27094734-27094734 on Build GRCh38
rs774571221 Chr.7: 27094578-27094578 on Build GRCh38
rs757856016 Chr.7: 27094667-27094667 on Build GRCh38
rs947408134 Chr.7: 27094859-27094859 on Build GRCh38
rs757644056 Chr.7: 27094864-27094864 on Build GRCh38
rs561337539 Chr.7: 27094857-27094857 on Build GRCh38
rs537351255 Chr.7: 27094684-27094684 on Build GRCh38

View Full Product Details