Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 32976107-32976600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP9
Entrez Gene ID
11328
Gene Name
FK506 binding protein 9
Gene Aliases
FKBP60, FKBP63, PPIase
UniGene
Hs.103934
Gene Chromosome Location
Chr.7: 32957404-33006931 on Build GRCh38
Transcripts
NM_001284341.1 NM_007270.4 XM_011515115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925347 c.611C>T p.P204L Chr.7: 32976407-32976407 on Build GRCh38
1252477 c.679C>T p.L227L Chr.7: 32976475-32976475 on Build GRCh38
3638179 c.611C>A p.P204H Chr.7: 32976407-32976407 on Build GRCh38
5654777 c.718C>T p.H240Y Chr.7: 32976355-32976355 on Build GRCh38
5654776 c.559C>T p.H187Y Chr.7: 32976355-32976355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199654330 Chr.7: 32976480-32976480 on Build GRCh38
rs771132804 Chr.7: 32976347-32976347 on Build GRCh38
rs757883393 Chr.7: 32976539-32976539 on Build GRCh38
rs757765676 Chr.7: 32976451-32976451 on Build GRCh38
rs1015290668 Chr.7: 32976555-32976555 on Build GRCh38
rs141686148 Chr.7: 32976374-32976374 on Build GRCh38
rs748786580 Chr.7: 32976329-32976329 on Build GRCh38
rs369192955 Chr.7: 32976324-32976324 on Build GRCh38
rs750481309 Chr.7: 32976396-32976396 on Build GRCh38
rs776765141 Chr.7: 32976138-32976138 on Build GRCh38
rs747725534 Chr.7: 32976447-32976447 on Build GRCh38
rs747969037 Chr.7: 32976536-32976537 on Build GRCh38
rs955248084 Chr.7: 32976281-32976281 on Build GRCh38
rs540278899 Chr.7: 32976315-32976315 on Build GRCh38
rs761858642 Chr.7: 32976509-32976509 on Build GRCh38
rs1011674698 Chr.7: 32976482-32976482 on Build GRCh38
rs766290864 Chr.7: 32976516-32976516 on Build GRCh38
rs962403820 Chr.7: 32976556-32976556 on Build GRCh38
rs1039014025 Chr.7: 32976503-32976503 on Build GRCh38
rs376303998 Chr.7: 32976361-32976361 on Build GRCh38
rs189401487 Chr.7: 32976287-32976287 on Build GRCh38
rs531164128 Chr.7: 32976412-32976412 on Build GRCh38
rs118076591 Chr.7: 32976530-32976530 on Build GRCh38
rs749978068 Chr.7: 32976323-32976323 on Build GRCh38
rs140953912 Chr.7: 32976362-32976362 on Build GRCh38
rs754479709 Chr.7: 32976432-32976432 on Build GRCh38
rs749530079 Chr.7: 32976485-32976485 on Build GRCh38
rs765037157 Chr.7: 32976529-32976529 on Build GRCh38
rs150199976 Chr.7: 32976247-32976247 on Build GRCh38
rs781693372 Chr.7: 32976545-32976545 on Build GRCh38
rs761817386 Chr.7: 32976380-32976380 on Build GRCh38
rs3750115 Chr.7: 32976290-32976290 on Build GRCh38
rs766099820 Chr.7: 32976397-32976397 on Build GRCh38
rs778376046 Chr.7: 32976341-32976341 on Build GRCh38
rs3750116 Chr.7: 32976508-32976508 on Build GRCh38
rs375434810 Chr.7: 32976145-32976145 on Build GRCh38
rs747529830 Chr.7: 32976342-32976342 on Build GRCh38
rs750920007 Chr.7: 32976550-32976550 on Build GRCh38
rs781179427 Chr.7: 32976327-32976328 on Build GRCh38
rs779631252 Chr.7: 32976325-32976325 on Build GRCh38
rs982926159 Chr.7: 32976286-32976286 on Build GRCh38
rs915547661 Chr.7: 32976575-32976575 on Build GRCh38
rs780358625 Chr.7: 32976507-32976509 on Build GRCh38
rs180715994 Chr.7: 32976558-32976558 on Build GRCh38
rs532106103 Chr.7: 32976364-32976364 on Build GRCh38
rs963921893 Chr.7: 32976141-32976141 on Build GRCh38
rs770082248 Chr.7: 32976476-32976476 on Build GRCh38
rs575411934 Chr.7: 32976232-32976232 on Build GRCh38
rs138478200 Chr.7: 32976381-32976381 on Build GRCh38
rs778568390 Chr.7: 32976436-32976436 on Build GRCh38
rs768312933 Chr.7: 32976330-32976330 on Build GRCh38
rs781596802 Chr.7: 32976452-32976452 on Build GRCh38
rs150187948 Chr.7: 32976369-32976369 on Build GRCh38
rs767467101 Chr.7: 32976511-32976511 on Build GRCh38
rs148386030 Chr.7: 32976551-32976551 on Build GRCh38
rs750220340 Chr.7: 32976383-32976383 on Build GRCh38
rs746207939 Chr.7: 32976454-32976454 on Build GRCh38
rs754671956 Chr.7: 32976522-32976522 on Build GRCh38
rs374892155 Chr.7: 32976351-32976351 on Build GRCh38
rs372550709 Chr.7: 32976355-32976355 on Build GRCh38
rs753473400 Chr.7: 32976518-32976518 on Build GRCh38
rs1029887874 Chr.7: 32976242-32976242 on Build GRCh38
rs773064696 Chr.7: 32976512-32976512 on Build GRCh38
rs755911645 Chr.7: 32976386-32976386 on Build GRCh38
rs756438298 Chr.7: 32976471-32976471 on Build GRCh38

View Full Product Details