Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 33097193-33097620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RP9
Entrez Gene ID
6100
Gene Name
retinitis pigmentosa 9 (autosomal dominant)
Gene Aliases
PAP-1, PAP1
UniGene
Hs.326805
Gene Chromosome Location
Chr.7: 33094797-33109390 on Build GRCh38
Transcripts
NM_203288.1 XM_011515468.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3394719 c.386A>T p.K129M Chr.7: 33097290-33097290 on Build GRCh38
1089262 c.328C>T p.R110C Chr.7: 33097348-33097348 on Build GRCh38
4830145 c.319C>A p.R107S Chr.7: 33097357-33097357 on Build GRCh38
3832683 c.401G>C p.R134T Chr.7: 33097275-33097275 on Build GRCh38
4903262 c.361C>T p.P121S Chr.7: 33097315-33097315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951630905 Chr.7: 33097283-33097283 on Build GRCh38
rs764261724 Chr.7: 33097325-33097325 on Build GRCh38
rs920753873 Chr.7: 33097311-33097311 on Build GRCh38
rs757707033 Chr.7: 33097303-33097303 on Build GRCh38
rs758778831 Chr.7: 33097263-33097263 on Build GRCh38
rs908952819 Chr.7: 33097329-33097329 on Build GRCh38
rs750225004 Chr.7: 33097341-33097341 on Build GRCh38
rs778451715 Chr.7: 33097282-33097282 on Build GRCh38
rs1001623936 Chr.7: 33097582-33097582 on Build GRCh38
rs760221836 Chr.7: 33097348-33097348 on Build GRCh38
rs753243542 Chr.7: 33097359-33097359 on Build GRCh38
rs1010058296 Chr.7: 33097552-33097552 on Build GRCh38
rs747429937 Chr.7: 33097428-33097428 on Build GRCh38
rs367979669 Chr.7: 33097357-33097357 on Build GRCh38
rs1042921866 Chr.7: 33097554-33097554 on Build GRCh38
rs781676257 Chr.7: 33097383-33097383 on Build GRCh38
rs765775300 Chr.7: 33097236-33097236 on Build GRCh38
rs867722694 Chr.7: 33097304-33097304 on Build GRCh38
rs1014528369 Chr.7: 33097560-33097560 on Build GRCh38
rs74690167 Chr.7: 33097398-33097398 on Build GRCh38
rs996335948 Chr.7: 33097506-33097506 on Build GRCh38
rs1038654176 Chr.7: 33097336-33097336 on Build GRCh38
rs764634519 Chr.7: 33097364-33097364 on Build GRCh38
rs575309519 Chr.7: 33097317-33097317 on Build GRCh38
rs6462460 Chr.7: 33097371-33097371 on Build GRCh38
rs767294005 Chr.7: 33097342-33097342 on Build GRCh38
rs770107330 Chr.7: 33097323-33097323 on Build GRCh38
rs775386910 Chr.7: 33097325-33097325 on Build GRCh38
rs762171551 Chr.7: 33097571-33097571 on Build GRCh38
rs781733714 Chr.7: 33097313-33097313 on Build GRCh38
rs773031086 Chr.7: 33097344-33097344 on Build GRCh38
rs150971445 Chr.7: 33097290-33097290 on Build GRCh38
rs948177667 Chr.7: 33097362-33097362 on Build GRCh38
rs1021506260 Chr.7: 33097482-33097482 on Build GRCh38
rs533613991 Chr.7: 33097533-33097533 on Build GRCh38
rs371503287 Chr.7: 33097519-33097519 on Build GRCh38
rs143749608 Chr.7: 33097404-33097404 on Build GRCh38
rs754518979 Chr.7: 33097361-33097361 on Build GRCh38
rs1010145062 Chr.7: 33097446-33097446 on Build GRCh38
rs753436304 Chr.7: 33097247-33097247 on Build GRCh38
rs760406324 Chr.7: 33097366-33097371 on Build GRCh38
rs200418776 Chr.7: 33097300-33097300 on Build GRCh38
rs148957070 Chr.7: 33097331-33097331 on Build GRCh38
rs1028713022 Chr.7: 33097518-33097518 on Build GRCh38
rs891742316 Chr.7: 33097425-33097425 on Build GRCh38
rs755860459 Chr.7: 33097229-33097229 on Build GRCh38
rs768515547 Chr.7: 33097332-33097332 on Build GRCh38
rs752110808 Chr.7: 33097366-33097366 on Build GRCh38
rs904315652 Chr.7: 33097496-33097496 on Build GRCh38
rs761796080 Chr.7: 33097339-33097339 on Build GRCh38
rs774286319 Chr.7: 33097335-33097335 on Build GRCh38

View Full Product Details