Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45075644-45076134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2
Entrez Gene ID
83605
Gene Name
CCM2 scaffolding protein
Gene Aliases
C7orf22, OSM, PP10187
UniGene
Hs.148272
Gene Chromosome Location
Chr.7: 44999746-45076470 on Build GRCh38
Transcripts
NM_001029835.2 NM_001167934.1 NM_001167935.1 NM_031443.3 NR_030770.1 XM_006715785.3 XM_006715786.3 XM_011515561.2 XM_011515562.1 XM_011515563.2 XM_011515564.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6530114 c.1217T>C p.F406S Chr.7: 45075876-45075876 on Build GRCh38
1313117 c.1185G>A p.V395V Chr.7: 45075844-45075844 on Build GRCh38
140580 c.1219G>T p.G407C Chr.7: 45075878-45075878 on Build GRCh38
3639078 c.1176C>T p.T392T Chr.7: 45075835-45075835 on Build GRCh38
6586048 c.1177A>T p.I393F Chr.7: 45075836-45075836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367588346 Chr.7: 45075755-45075755 on Build GRCh38
rs3214691 Chr.7: 45076076-45076077 on Build GRCh38
rs138150576 Chr.7: 45075785-45075785 on Build GRCh38
rs147293659 Chr.7: 45075961-45075961 on Build GRCh38
rs760734435 Chr.7: 45075811-45075811 on Build GRCh38
rs145734072 Chr.7: 45076035-45076035 on Build GRCh38
rs1050782162 Chr.7: 45075979-45075979 on Build GRCh38
rs572659898 Chr.7: 45075681-45075681 on Build GRCh38
rs768545818 Chr.7: 45076028-45076028 on Build GRCh38
rs370697198 Chr.7: 45075791-45075791 on Build GRCh38
rs370695543 Chr.7: 45075738-45075738 on Build GRCh38
rs763227672 Chr.7: 45075976-45075976 on Build GRCh38
rs779863927 Chr.7: 45075950-45075950 on Build GRCh38
rs368380083 Chr.7: 45075902-45075902 on Build GRCh38
rs746677393 Chr.7: 45076019-45076019 on Build GRCh38
rs766730001 Chr.7: 45075918-45075918 on Build GRCh38
rs549735292 Chr.7: 45076077-45076077 on Build GRCh38
rs904473908 Chr.7: 45075805-45075805 on Build GRCh38
rs767862839 Chr.7: 45076070-45076070 on Build GRCh38
rs751785013 Chr.7: 45075989-45075989 on Build GRCh38
rs765250660 Chr.7: 45075908-45075908 on Build GRCh38
rs764064114 Chr.7: 45075890-45075890 on Build GRCh38
rs745530586 Chr.7: 45075863-45075863 on Build GRCh38
rs773523983 Chr.7: 45076069-45076069 on Build GRCh38
rs575724801 Chr.7: 45076048-45076048 on Build GRCh38
rs375545788 Chr.7: 45075748-45075748 on Build GRCh38
rs371498958 Chr.7: 45076000-45076000 on Build GRCh38
rs956786423 Chr.7: 45076051-45076051 on Build GRCh38
rs374626897 Chr.7: 45075855-45075855 on Build GRCh38
rs774455845 Chr.7: 45076091-45076091 on Build GRCh38
rs370366469 Chr.7: 45075766-45075766 on Build GRCh38
rs764262513 Chr.7: 45075812-45075812 on Build GRCh38
rs781030372 Chr.7: 45076080-45076080 on Build GRCh38
rs143962510 Chr.7: 45075960-45075960 on Build GRCh38
rs201559289 Chr.7: 45075813-45075813 on Build GRCh38
rs200969851 Chr.7: 45076072-45076072 on Build GRCh38
rs753227642 Chr.7: 45075999-45075999 on Build GRCh38
rs148496534 Chr.7: 45076036-45076036 on Build GRCh38
rs140689222 Chr.7: 45076015-45076015 on Build GRCh38
rs920839648 Chr.7: 45075923-45075923 on Build GRCh38
rs767893429 Chr.7: 45075992-45075992 on Build GRCh38
rs142647512 Chr.7: 45075837-45075837 on Build GRCh38
rs771869053 Chr.7: 45075871-45075871 on Build GRCh38
rs760675025 Chr.7: 45075880-45075880 on Build GRCh38
rs200124379 Chr.7: 45075745-45075745 on Build GRCh38
rs892453100 Chr.7: 45075752-45075752 on Build GRCh38
rs200669376 Chr.7: 45075842-45075842 on Build GRCh38
rs772753973 Chr.7: 45075769-45075769 on Build GRCh38
rs149630877 Chr.7: 45075680-45075680 on Build GRCh38
rs989827020 Chr.7: 45075888-45075888 on Build GRCh38
rs763128519 Chr.7: 45075917-45075917 on Build GRCh38
rs761701382 Chr.7: 45075964-45075964 on Build GRCh38
rs371810028 Chr.7: 45076092-45076092 on Build GRCh38
rs371640199 Chr.7: 45075990-45075990 on Build GRCh38
rs775544897 Chr.7: 45075802-45075802 on Build GRCh38
rs971745260 Chr.7: 45075916-45075916 on Build GRCh38
rs934954209 Chr.7: 45075701-45075701 on Build GRCh38
rs753234415 Chr.7: 45075931-45075931 on Build GRCh38
rs773743887 Chr.7: 45076069-45076070 on Build GRCh38
rs144401535 Chr.7: 45075739-45075739 on Build GRCh38
rs930808528 Chr.7: 45075696-45075696 on Build GRCh38
rs187505526 Chr.7: 45075771-45075771 on Build GRCh38
rs141205369 Chr.7: 45075919-45075919 on Build GRCh38
rs777532251 Chr.7: 45076085-45076085 on Build GRCh38
rs144650652 Chr.7: 45076063-45076063 on Build GRCh38
rs903856880 Chr.7: 45075972-45075972 on Build GRCh38
rs749246649 Chr.7: 45076089-45076089 on Build GRCh38
rs747843619 Chr.7: 45076081-45076081 on Build GRCh38
rs544276945 Chr.7: 45075956-45075956 on Build GRCh38
rs368035399 Chr.7: 45076079-45076079 on Build GRCh38
rs762878478 Chr.7: 45076050-45076050 on Build GRCh38
rs367740781 Chr.7: 45075925-45075925 on Build GRCh38
rs759309544 Chr.7: 45075798-45075798 on Build GRCh38
rs764537037 Chr.7: 45076101-45076101 on Build GRCh38
rs542561285 Chr.7: 45075898-45075898 on Build GRCh38
rs760907540 Chr.7: 45076100-45076100 on Build GRCh38
rs1043711376 Chr.7: 45075963-45075963 on Build GRCh38
rs145579135 Chr.7: 45075783-45075783 on Build GRCh38
rs774015400 Chr.7: 45075857-45075857 on Build GRCh38
rs2304691 Chr.7: 45075982-45075982 on Build GRCh38
rs182764453 Chr.7: 45075720-45075720 on Build GRCh38
rs1019140961 Chr.7: 45075886-45075886 on Build GRCh38
rs769680088 Chr.7: 45076044-45076044 on Build GRCh38
rs144625146 Chr.7: 45075841-45075841 on Build GRCh38
rs755945799 Chr.7: 45076084-45076084 on Build GRCh38
rs201750934 Chr.7: 45075747-45075747 on Build GRCh38
rs199522537 Chr.7: 45075817-45075817 on Build GRCh38
rs768620293 Chr.7: 45076070-45076070 on Build GRCh38
rs925243805 Chr.7: 45076107-45076107 on Build GRCh38
rs887304366 Chr.7: 45075958-45075958 on Build GRCh38
rs770977163 Chr.7: 45076090-45076090 on Build GRCh38
rs767248510 Chr.7: 45075800-45075800 on Build GRCh38
rs773236463 Chr.7: 45075971-45075971 on Build GRCh38
rs772221792 Chr.7: 45076093-45076093 on Build GRCh38
rs144918172 Chr.7: 45076006-45076006 on Build GRCh38
rs778326011 Chr.7: 45075943-45075943 on Build GRCh38
rs577692551 Chr.7: 45075690-45075690 on Build GRCh38
rs1000120007 Chr.7: 45075818-45075818 on Build GRCh38
rs371371873 Chr.7: 45075854-45075854 on Build GRCh38
rs750632206 Chr.7: 45075915-45075915 on Build GRCh38
rs939543180 Chr.7: 45075942-45075942 on Build GRCh38
rs1018695949 Chr.7: 45076013-45076013 on Build GRCh38
rs771030259 Chr.7: 45076062-45076062 on Build GRCh38
rs775493426 Chr.7: 45075877-45075877 on Build GRCh38
rs778959221 Chr.7: 45076012-45076012 on Build GRCh38
rs116320627 Chr.7: 45076071-45076071 on Build GRCh38
rs771678360 Chr.7: 45075946-45075946 on Build GRCh38
rs141057892 Chr.7: 45075967-45075967 on Build GRCh38
rs139366651 Chr.7: 45075957-45075957 on Build GRCh38
rs755548359 Chr.7: 45075845-45075845 on Build GRCh38
rs375261158 Chr.7: 45076098-45076098 on Build GRCh38
rs755352606 Chr.7: 45075921-45075921 on Build GRCh38
rs150734280 Chr.7: 45075939-45075939 on Build GRCh38
rs1027238438 Chr.7: 45076024-45076024 on Build GRCh38
rs373929813 Chr.7: 45076076-45076076 on Build GRCh38
rs148192057 Chr.7: 45075856-45075856 on Build GRCh38

View Full Product Details