Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45891841-45892343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP1
Entrez Gene ID
3484
Gene Name
insulin like growth factor binding protein 1
Gene Aliases
AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1
UniGene
Hs.642938
Gene Chromosome Location
Chr.7: 45888360-45893668 on Build GRCh38
Transcripts
NM_000596.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281874 c.530G>A p.R177Q Chr.7: 45891942-45891942 on Build GRCh38
3881353 c.529C>T p.R177* Chr.7: 45891941-45891941 on Build GRCh38
3881354 c.535G>T p.E179* Chr.7: 45891947-45891947 on Build GRCh38
5543931 c.617G>T p.C206F Chr.7: 45892029-45892029 on Build GRCh38
1330600 c.575A>G p.E192G Chr.7: 45891987-45891987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004511412 Chr.7: 45892260-45892260 on Build GRCh38
rs369196046 Chr.7: 45891942-45891942 on Build GRCh38
rs1015863188 Chr.7: 45892315-45892315 on Build GRCh38
rs905963558 Chr.7: 45892178-45892178 on Build GRCh38
rs760281769 Chr.7: 45891936-45891936 on Build GRCh38
rs774512092 Chr.7: 45892082-45892082 on Build GRCh38
rs763174847 Chr.7: 45891956-45891956 on Build GRCh38
rs562582799 Chr.7: 45892118-45892118 on Build GRCh38
rs375773835 Chr.7: 45891913-45891913 on Build GRCh38
rs780481510 Chr.7: 45891900-45891900 on Build GRCh38
rs763075160 Chr.7: 45892003-45892003 on Build GRCh38
rs768523904 Chr.7: 45891935-45891935 on Build GRCh38
rs759646065 Chr.7: 45891974-45891974 on Build GRCh38
rs747950009 Chr.7: 45892046-45892046 on Build GRCh38
rs771885139 Chr.7: 45891899-45891899 on Build GRCh38
rs757935661 Chr.7: 45892019-45892019 on Build GRCh38
rs1065782 Chr.7: 45891959-45891959 on Build GRCh38
rs950958919 Chr.7: 45892219-45892219 on Build GRCh38
rs757704561 Chr.7: 45892099-45892099 on Build GRCh38
rs756491869 Chr.7: 45891992-45891992 on Build GRCh38
rs773322292 Chr.7: 45892054-45892054 on Build GRCh38
rs765237806 Chr.7: 45891991-45891991 on Build GRCh38
rs1053533634 Chr.7: 45892007-45892007 on Build GRCh38
rs751857594 Chr.7: 45891961-45891961 on Build GRCh38
rs373948510 Chr.7: 45892078-45892078 on Build GRCh38
rs1036273984 Chr.7: 45892211-45892211 on Build GRCh38
rs983556093 Chr.7: 45891872-45891872 on Build GRCh38
rs562004297 Chr.7: 45892075-45892075 on Build GRCh38
rs369532793 Chr.7: 45892279-45892279 on Build GRCh38
rs764586069 Chr.7: 45892256-45892256 on Build GRCh38
rs752943844 Chr.7: 45892086-45892086 on Build GRCh38
rs759811708 Chr.7: 45891921-45891922 on Build GRCh38
rs770139318 Chr.7: 45891927-45891927 on Build GRCh38
rs146202574 Chr.7: 45891962-45891962 on Build GRCh38
rs754365887 Chr.7: 45892097-45892097 on Build GRCh38
rs769609357 Chr.7: 45892053-45892053 on Build GRCh38
rs369361373 Chr.7: 45892022-45892022 on Build GRCh38
rs540843488 Chr.7: 45892042-45892042 on Build GRCh38
rs756480193 Chr.7: 45892091-45892091 on Build GRCh38
rs776233561 Chr.7: 45891941-45891941 on Build GRCh38
rs749767604 Chr.7: 45892018-45892018 on Build GRCh38
rs954654299 Chr.7: 45892184-45892184 on Build GRCh38
rs781367934 Chr.7: 45891919-45891919 on Build GRCh38
rs767692464 Chr.7: 45892085-45892085 on Build GRCh38
rs139788715 Chr.7: 45891955-45891955 on Build GRCh38
rs916430390 Chr.7: 45891943-45891943 on Build GRCh38
rs974487740 Chr.7: 45891990-45891990 on Build GRCh38
rs776400413 Chr.7: 45892083-45892083 on Build GRCh38
rs560655167 Chr.7: 45892312-45892312 on Build GRCh38
rs773088537 Chr.7: 45891921-45891921 on Build GRCh38
rs752931809 Chr.7: 45892000-45892000 on Build GRCh38
rs558612069 Chr.7: 45892125-45892125 on Build GRCh38
rs748709533 Chr.7: 45891923-45891923 on Build GRCh38
rs182253310 Chr.7: 45892152-45892152 on Build GRCh38
rs577810813 Chr.7: 45891876-45891876 on Build GRCh38
rs371271611 Chr.7: 45891952-45891952 on Build GRCh38
rs778342842 Chr.7: 45891934-45891934 on Build GRCh38
rs1011797601 Chr.7: 45892029-45892029 on Build GRCh38
rs376136318 Chr.7: 45891994-45891994 on Build GRCh38
rs9658219 Chr.7: 45892109-45892109 on Build GRCh38
rs767935469 Chr.7: 45891984-45891984 on Build GRCh38
rs948759128 Chr.7: 45891945-45891945 on Build GRCh38
rs544818427 Chr.7: 45891907-45891907 on Build GRCh38
rs765385012 Chr.7: 45892089-45892096 on Build GRCh38
rs376058917 Chr.7: 45892261-45892261 on Build GRCh38
rs768128135 Chr.7: 45892033-45892033 on Build GRCh38
rs973336136 Chr.7: 45892316-45892316 on Build GRCh38
rs1023023100 Chr.7: 45892133-45892133 on Build GRCh38
rs764405161 Chr.7: 45892093-45892093 on Build GRCh38

View Full Product Details