Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55365350-55365854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LANCL2
Entrez Gene ID
55915
Gene Name
LanC like 2
Gene Aliases
GPR69B, TASP
UniGene
Hs.595384
Gene Chromosome Location
Chr.7: 55365448-55433742 on Build GRCh38
Transcripts
NM_018697.3

SNP Detail

SNP ID Chromosome Location
rs1057266 Chr.7: 55365572-55365572 on Build GRCh38
rs369345106 Chr.7: 55365796-55365796 on Build GRCh38
rs976445219 Chr.7: 55365766-55365766 on Build GRCh38
rs570806490 Chr.7: 55365623-55365623 on Build GRCh38
rs1050136613 Chr.7: 55365615-55365615 on Build GRCh38
rs985815284 Chr.7: 55365776-55365776 on Build GRCh38
rs917186185 Chr.7: 55365532-55365532 on Build GRCh38
rs967274081 Chr.7: 55365689-55365689 on Build GRCh38
rs758918204 Chr.7: 55365558-55365558 on Build GRCh38
rs778341804 Chr.7: 55365729-55365729 on Build GRCh38
rs879713949 Chr.7: 55365399-55365400 on Build GRCh38
rs931747582 Chr.7: 55365601-55365601 on Build GRCh38
rs547546896 Chr.7: 55365515-55365515 on Build GRCh38
rs182929996 Chr.7: 55365828-55365828 on Build GRCh38
rs781594009 Chr.7: 55365701-55365701 on Build GRCh38
rs907724091 Chr.7: 55365742-55365742 on Build GRCh38
rs35026575 Chr.7: 55365539-55365540 on Build GRCh38
rs750922234 Chr.7: 55365771-55365771 on Build GRCh38
rs980214072 Chr.7: 55365489-55365489 on Build GRCh38
rs922922653 Chr.7: 55365769-55365769 on Build GRCh38
rs1010936436 Chr.7: 55365653-55365653 on Build GRCh38
rs529349689 Chr.7: 55365445-55365445 on Build GRCh38
rs552325987 Chr.7: 55365619-55365619 on Build GRCh38
rs932162467 Chr.7: 55365600-55365600 on Build GRCh38
rs556202413 Chr.7: 55365684-55365684 on Build GRCh38
rs922112118 Chr.7: 55365588-55365588 on Build GRCh38
rs538716085 Chr.7: 55365575-55365575 on Build GRCh38
rs892030773 Chr.7: 55365632-55365632 on Build GRCh38
rs568010143 Chr.7: 55365732-55365732 on Build GRCh38
rs927106199 Chr.7: 55365494-55365494 on Build GRCh38
rs1006338744 Chr.7: 55365837-55365837 on Build GRCh38
rs1015010588 Chr.7: 55365421-55365424 on Build GRCh38
rs553551242 Chr.7: 55365791-55365791 on Build GRCh38
rs1034203531 Chr.7: 55365452-55365452 on Build GRCh38
rs951860485 Chr.7: 55365503-55365503 on Build GRCh38
rs1024299619 Chr.7: 55365680-55365680 on Build GRCh38
rs977015317 Chr.7: 55365690-55365690 on Build GRCh38
rs1010258182 Chr.7: 55365407-55365407 on Build GRCh38
rs963503000 Chr.7: 55365432-55365432 on Build GRCh38
rs1024168050 Chr.7: 55365686-55365686 on Build GRCh38
rs78936413 Chr.7: 55365571-55365571 on Build GRCh38
rs11546656 Chr.7: 55365629-55365629 on Build GRCh38
rs912111186 Chr.7: 55365768-55365768 on Build GRCh38
rs961974193 Chr.7: 55365758-55365758 on Build GRCh38
rs995041722 Chr.7: 55365439-55365439 on Build GRCh38
rs967014958 Chr.7: 55365539-55365539 on Build GRCh38
rs946315206 Chr.7: 55365792-55365792 on Build GRCh38
rs1036368100 Chr.7: 55365622-55365622 on Build GRCh38
rs957292940 Chr.7: 55365699-55365699 on Build GRCh38
rs984670104 Chr.7: 55365553-55365553 on Build GRCh38
rs971495244 Chr.7: 55365431-55365431 on Build GRCh38
rs1034439741 Chr.7: 55365718-55365718 on Build GRCh38
rs3088171 Chr.7: 55365417-55365417 on Build GRCh38
rs1052014480 Chr.7: 55365609-55365609 on Build GRCh38
rs760386337 Chr.7: 55365812-55365813 on Build GRCh38
rs936450358 Chr.7: 55365786-55365786 on Build GRCh38
rs994943304 Chr.7: 55365626-55365628 on Build GRCh38
rs890443831 Chr.7: 55365409-55365409 on Build GRCh38
rs951829382 Chr.7: 55365727-55365727 on Build GRCh38
rs956207740 Chr.7: 55365393-55365393 on Build GRCh38
rs1054541198 Chr.7: 55365787-55365787 on Build GRCh38
rs987648347 Chr.7: 55365743-55365743 on Build GRCh38
rs138187979 Chr.7: 55365812-55365812 on Build GRCh38
rs1032692835 Chr.7: 55365697-55365697 on Build GRCh38
rs894598215 Chr.7: 55365820-55365820 on Build GRCh38
rs374910959 Chr.7: 55365722-55365722 on Build GRCh38
rs907166171 Chr.7: 55365808-55365808 on Build GRCh38
rs373858559 Chr.7: 55365783-55365783 on Build GRCh38
rs756733919 Chr.7: 55365725-55365725 on Build GRCh38
rs543454650 Chr.7: 55365380-55365380 on Build GRCh38
rs1001547003 Chr.7: 55365811-55365811 on Build GRCh38
rs371400750 Chr.7: 55365574-55365574 on Build GRCh38

View Full Product Details